Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76732572-76733150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2R
Entrez Gene ID
2149
Gene Name
coagulation factor II thrombin receptor
Gene Aliases
CF2R, HTR, PAR-1, PAR1, TR
UniGene
Hs.482562
Gene Chromosome Location
Chr.5: 76716043-76735780 on Build GRCh38
Transcripts
NM_001311313.1 NM_001992.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316319 c.707C>A p.P236H Chr.5: 76732932-76732932 on Build GRCh38
4550187 c.494G>A p.G165D Chr.5: 76732719-76732719 on Build GRCh38
1070231 c.598C>T p.R200W Chr.5: 76732823-76732823 on Build GRCh38
82127 c.879C>T p.I293I Chr.5: 76733104-76733104 on Build GRCh38
3429650 c.789C>T p.L263L Chr.5: 76733014-76733014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774046659 Chr.5: 76733105-76733105 on Build GRCh38
rs5894 Chr.5: 76732741-76732741 on Build GRCh38
rs368095355 Chr.5: 76732867-76732867 on Build GRCh38
rs756644144 Chr.5: 76732892-76732892 on Build GRCh38
rs1005319134 Chr.5: 76732808-76732808 on Build GRCh38
rs746379887 Chr.5: 76732787-76732787 on Build GRCh38
rs1055103 Chr.5: 76733027-76733027 on Build GRCh38
rs5893 Chr.5: 76732721-76732721 on Build GRCh38
rs368339604 Chr.5: 76732855-76732855 on Build GRCh38
rs754868290 Chr.5: 76732667-76732667 on Build GRCh38
rs753654575 Chr.5: 76732646-76732646 on Build GRCh38
rs772907284 Chr.5: 76733118-76733118 on Build GRCh38
rs777014666 Chr.5: 76732595-76732595 on Build GRCh38
rs5895 Chr.5: 76732610-76732610 on Build GRCh38
rs781662568 Chr.5: 76732786-76732786 on Build GRCh38
rs745475424 Chr.5: 76733089-76733089 on Build GRCh38
rs780737633 Chr.5: 76733086-76733086 on Build GRCh38
rs952663209 Chr.5: 76732783-76732783 on Build GRCh38
rs139001726 Chr.5: 76732903-76732903 on Build GRCh38
rs752508356 Chr.5: 76732865-76732865 on Build GRCh38
rs761939723 Chr.5: 76732851-76732851 on Build GRCh38
rs909217054 Chr.5: 76732663-76732663 on Build GRCh38
rs772218979 Chr.5: 76732940-76732940 on Build GRCh38
rs374309035 Chr.5: 76733064-76733064 on Build GRCh38
rs532063071 Chr.5: 76732728-76732728 on Build GRCh38
rs751241586 Chr.5: 76732868-76732868 on Build GRCh38
rs374709959 Chr.5: 76733114-76733114 on Build GRCh38
rs755299108 Chr.5: 76733043-76733043 on Build GRCh38
rs756689476 Chr.5: 76732869-76732869 on Build GRCh38
rs139787379 Chr.5: 76733066-76733066 on Build GRCh38
rs779156273 Chr.5: 76733050-76733050 on Build GRCh38
rs566159512 Chr.5: 76732716-76732716 on Build GRCh38
rs144447562 Chr.5: 76733022-76733022 on Build GRCh38
rs768959898 Chr.5: 76732908-76732908 on Build GRCh38
rs143038729 Chr.5: 76732642-76732642 on Build GRCh38
rs867136802 Chr.5: 76732904-76732904 on Build GRCh38
rs2227832 Chr.5: 76732994-76732994 on Build GRCh38
rs568954157 Chr.5: 76732751-76732751 on Build GRCh38
rs866890467 Chr.5: 76732899-76732899 on Build GRCh38
rs768450420 Chr.5: 76733095-76733095 on Build GRCh38
rs780695412 Chr.5: 76732876-76732876 on Build GRCh38
rs367893721 Chr.5: 76733041-76733041 on Build GRCh38
rs900744536 Chr.5: 76732738-76732738 on Build GRCh38
rs771440132 Chr.5: 76732693-76732693 on Build GRCh38
rs755668907 Chr.5: 76733014-76733014 on Build GRCh38
rs752467852 Chr.5: 76732817-76732817 on Build GRCh38
rs375693604 Chr.5: 76732718-76732718 on Build GRCh38
rs748320077 Chr.5: 76732937-76732937 on Build GRCh38
rs143981953 Chr.5: 76732944-76732944 on Build GRCh38
rs181326734 Chr.5: 76732633-76732633 on Build GRCh38
rs762806598 Chr.5: 76732966-76732966 on Build GRCh38
rs766397208 Chr.5: 76732745-76732745 on Build GRCh38
rs763800661 Chr.5: 76732982-76732982 on Build GRCh38
rs777334197 Chr.5: 76732757-76732757 on Build GRCh38
rs775856193 Chr.5: 76732812-76732812 on Build GRCh38
rs983917373 Chr.5: 76732660-76732660 on Build GRCh38
rs758373073 Chr.5: 76732890-76732890 on Build GRCh38
rs568758286 Chr.5: 76733001-76733001 on Build GRCh38
rs758102941 Chr.5: 76732756-76732756 on Build GRCh38
rs780601999 Chr.5: 76733126-76733126 on Build GRCh38
rs778691192 Chr.5: 76732669-76732669 on Build GRCh38
rs946825335 Chr.5: 76732615-76732615 on Build GRCh38
rs993867735 Chr.5: 76733020-76733020 on Build GRCh38
rs746222469 Chr.5: 76732717-76732717 on Build GRCh38
rs759556594 Chr.5: 76732859-76732859 on Build GRCh38
rs772339992 Chr.5: 76732852-76732852 on Build GRCh38
rs538007698 Chr.5: 76732824-76732824 on Build GRCh38
rs760173380 Chr.5: 76733128-76733128 on Build GRCh38
rs546007149 Chr.5: 76732598-76732598 on Build GRCh38
rs747075865 Chr.5: 76733084-76733084 on Build GRCh38
rs769834943 Chr.5: 76732961-76732961 on Build GRCh38
rs371207871 Chr.5: 76733012-76733012 on Build GRCh38
rs771205478 Chr.5: 76733085-76733085 on Build GRCh38
rs747183968 Chr.5: 76732791-76732792 on Build GRCh38
rs374957006 Chr.5: 76732622-76732622 on Build GRCh38
rs761638806 Chr.5: 76733109-76733109 on Build GRCh38
rs756097745 Chr.5: 76732636-76732636 on Build GRCh38
rs547401113 Chr.5: 76732924-76732924 on Build GRCh38
rs142662743 Chr.5: 76733088-76733088 on Build GRCh38
rs754812855 Chr.5: 76732749-76732749 on Build GRCh38
rs769598509 Chr.5: 76732626-76732626 on Build GRCh38
rs774484456 Chr.5: 76732849-76732849 on Build GRCh38
rs751167278 Chr.5: 76732992-76732992 on Build GRCh38
rs373886145 Chr.5: 76732866-76732866 on Build GRCh38
rs147817829 Chr.5: 76733059-76733059 on Build GRCh38
rs770087836 Chr.5: 76732788-76732788 on Build GRCh38
rs139693607 Chr.5: 76732963-76732963 on Build GRCh38
rs756780148 Chr.5: 76732775-76732775 on Build GRCh38
rs778192329 Chr.5: 76733077-76733077 on Build GRCh38
rs1039326870 Chr.5: 76732730-76732730 on Build GRCh38
rs994092360 Chr.5: 76732762-76732762 on Build GRCh38
rs546186677 Chr.5: 76732686-76732686 on Build GRCh38
rs760879718 Chr.5: 76732953-76732953 on Build GRCh38
rs915453516 Chr.5: 76732927-76732927 on Build GRCh38
rs780463885 Chr.5: 76732893-76732893 on Build GRCh38
rs752231723 Chr.5: 76732752-76732752 on Build GRCh38
rs2230849 Chr.5: 76732784-76732784 on Build GRCh38
rs529977516 Chr.5: 76732682-76732682 on Build GRCh38
rs774519321 Chr.5: 76732731-76732731 on Build GRCh38
rs766975443 Chr.5: 76733009-76733009 on Build GRCh38
rs560811435 Chr.5: 76732670-76732670 on Build GRCh38
rs746510136 Chr.5: 76732770-76732770 on Build GRCh38
rs138496271 Chr.5: 76732617-76732617 on Build GRCh38
rs1051926439 Chr.5: 76733015-76733015 on Build GRCh38
rs757777409 Chr.5: 76732679-76732679 on Build GRCh38
rs763252115 Chr.5: 76732725-76732725 on Build GRCh38
rs750271706 Chr.5: 76732635-76732635 on Build GRCh38
rs775623100 Chr.5: 76732722-76732722 on Build GRCh38
rs149484608 Chr.5: 76732933-76732933 on Build GRCh38
rs939416472 Chr.5: 76732968-76732968 on Build GRCh38
rs765073697 Chr.5: 76732863-76732863 on Build GRCh38
rs756717676 Chr.5: 76733023-76733023 on Build GRCh38

View Full Product Details