Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141653166-141653725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAP3
Entrez Gene ID
64411
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3
Gene Aliases
CENTD3, DRAG1
UniGene
Hs.726187
Gene Chromosome Location
Chr.5: 141653401-141682233 on Build GRCh38
Transcripts
NM_022481.5 XM_005268497.1 XM_005268498.1 XM_005268499.1 XM_006714792.1 XM_011537677.2

SNP Detail

SNP ID Chromosome Location
rs956030142 Chr.5: 141653415-141653415 on Build GRCh38
rs556884904 Chr.5: 141653646-141653646 on Build GRCh38
rs932925930 Chr.5: 141653567-141653567 on Build GRCh38
rs781486940 Chr.5: 141653289-141653289 on Build GRCh38
rs756404236 Chr.5: 141653416-141653416 on Build GRCh38
rs990528901 Chr.5: 141653279-141653279 on Build GRCh38
rs532987748 Chr.5: 141653352-141653352 on Build GRCh38
rs893916468 Chr.5: 141653443-141653443 on Build GRCh38
rs749629241 Chr.5: 141653495-141653495 on Build GRCh38
rs774784566 Chr.5: 141653567-141653567 on Build GRCh38
rs796286702 Chr.5: 141653467-141653467 on Build GRCh38
rs184224756 Chr.5: 141653679-141653679 on Build GRCh38
rs139650880 Chr.5: 141653288-141653288 on Build GRCh38
rs764744612 Chr.5: 141653414-141653414 on Build GRCh38
rs547836299 Chr.5: 141653496-141653496 on Build GRCh38
rs1016413944 Chr.5: 141653528-141653528 on Build GRCh38
rs777883699 Chr.5: 141653453-141653453 on Build GRCh38
rs1054279786 Chr.5: 141653436-141653436 on Build GRCh38
rs908179443 Chr.5: 141653519-141653519 on Build GRCh38
rs989359948 Chr.5: 141653651-141653651 on Build GRCh38
rs970387673 Chr.5: 141653574-141653574 on Build GRCh38
rs115301013 Chr.5: 141653225-141653225 on Build GRCh38
rs570394066 Chr.5: 141653672-141653672 on Build GRCh38
rs553351568 Chr.5: 141653694-141653694 on Build GRCh38
rs949259030 Chr.5: 141653308-141653308 on Build GRCh38
rs901200579 Chr.5: 141653474-141653474 on Build GRCh38
rs1039680800 Chr.5: 141653473-141653473 on Build GRCh38
rs963130710 Chr.5: 141653544-141653544 on Build GRCh38
rs567925151 Chr.5: 141653547-141653547 on Build GRCh38
rs1041670377 Chr.5: 141653551-141653551 on Build GRCh38
rs775105878 Chr.5: 141653668-141653668 on Build GRCh38
rs746433161 Chr.5: 141653619-141653619 on Build GRCh38
rs771291805 Chr.5: 141653506-141653506 on Build GRCh38
rs1000317841 Chr.5: 141653364-141653364 on Build GRCh38
rs923896488 Chr.5: 141653624-141653624 on Build GRCh38
rs891770259 Chr.5: 141653648-141653648 on Build GRCh38
rs1049915717 Chr.5: 141653579-141653579 on Build GRCh38
rs1041301899 Chr.5: 141653205-141653205 on Build GRCh38
rs998354310 Chr.5: 141653482-141653482 on Build GRCh38
rs372236677 Chr.5: 141653565-141653565 on Build GRCh38
rs755092091 Chr.5: 141653234-141653234 on Build GRCh38
rs1031675856 Chr.5: 141653378-141653378 on Build GRCh38
rs970941816 Chr.5: 141653472-141653472 on Build GRCh38
rs759083217 Chr.5: 141653391-141653391 on Build GRCh38
rs141725643 Chr.5: 141653358-141653358 on Build GRCh38
rs983874292 Chr.5: 141653514-141653514 on Build GRCh38
rs73794961 Chr.5: 141653631-141653631 on Build GRCh38

View Full Product Details