Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87505363-87505702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MINPP1
Entrez Gene ID
9562
Gene Name
multiple inositol-polyphosphate phosphatase 1
Gene Aliases
HIPER1, MINPP2, MIPP
UniGene
Hs.121260
Gene Chromosome Location
Chr.10: 87502792-87553461 on Build GRCh38
Transcripts
NM_001178117.1 NM_004897.4 XM_006718078.2 XM_011540379.2 XM_017016965.1 XM_017016966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5703159 c.552G>T p.K184N Chr.10: 87505467-87505467 on Build GRCh38
7242918 c.514G>C p.E172Q Chr.10: 87505429-87505429 on Build GRCh38
7401737 c.565G>C p.D189H Chr.10: 87505480-87505480 on Build GRCh38
5572122 c.596G>A p.W199* Chr.10: 87505511-87505511 on Build GRCh38
4988054 c.484G>A p.A162T Chr.10: 87505399-87505399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577883422 Chr.10: 87505625-87505625 on Build GRCh38
rs151188172 Chr.10: 87505500-87505500 on Build GRCh38
rs1030864515 Chr.10: 87505613-87505613 on Build GRCh38
rs776051431 Chr.10: 87505524-87505533 on Build GRCh38
rs142167582 Chr.10: 87505408-87505408 on Build GRCh38
rs748205648 Chr.10: 87505387-87505387 on Build GRCh38
rs923056042 Chr.10: 87505583-87505583 on Build GRCh38
rs753447221 Chr.10: 87505562-87505562 on Build GRCh38
rs1049163644 Chr.10: 87505404-87505404 on Build GRCh38
rs990092549 Chr.10: 87505494-87505494 on Build GRCh38
rs567369847 Chr.10: 87505537-87505537 on Build GRCh38
rs908883433 Chr.10: 87505650-87505650 on Build GRCh38
rs752107977 Chr.10: 87505579-87505579 on Build GRCh38
rs764579968 Chr.10: 87505561-87505562 on Build GRCh38
rs749403575 Chr.10: 87505584-87505584 on Build GRCh38
rs545606147 Chr.10: 87505663-87505663 on Build GRCh38
rs774184136 Chr.10: 87505413-87505413 on Build GRCh38
rs765012386 Chr.10: 87505567-87505567 on Build GRCh38
rs746148934 Chr.10: 87505603-87505603 on Build GRCh38
rs941639256 Chr.10: 87505426-87505426 on Build GRCh38
rs770686571 Chr.10: 87505411-87505411 on Build GRCh38
rs752550671 Chr.10: 87505442-87505442 on Build GRCh38
rs748198537 Chr.10: 87505478-87505478 on Build GRCh38
rs200029988 Chr.10: 87505410-87505410 on Build GRCh38
rs760117757 Chr.10: 87505435-87505435 on Build GRCh38
rs763480512 Chr.10: 87505561-87505561 on Build GRCh38
rs769988677 Chr.10: 87505391-87505391 on Build GRCh38
rs753536996 Chr.10: 87505441-87505441 on Build GRCh38
rs370593785 Chr.10: 87505542-87505542 on Build GRCh38
rs763719114 Chr.10: 87505438-87505438 on Build GRCh38
rs577079738 Chr.10: 87505470-87505470 on Build GRCh38
rs763506510 Chr.10: 87505556-87505557 on Build GRCh38
rs755500266 Chr.10: 87505466-87505466 on Build GRCh38
rs1022752935 Chr.10: 87505557-87505557 on Build GRCh38
rs370984163 Chr.10: 87505570-87505570 on Build GRCh38
rs769745870 Chr.10: 87505479-87505479 on Build GRCh38
rs759423253 Chr.10: 87505417-87505417 on Build GRCh38
rs767389525 Chr.10: 87505423-87505423 on Build GRCh38
rs760733066 Chr.10: 87505559-87505559 on Build GRCh38
rs777669215 Chr.10: 87505581-87505581 on Build GRCh38
rs371279085 Chr.10: 87505394-87505394 on Build GRCh38
rs757010654 Chr.10: 87505447-87505447 on Build GRCh38
rs367962697 Chr.10: 87505454-87505454 on Build GRCh38
rs781552268 Chr.10: 87505477-87505477 on Build GRCh38
rs910432787 Chr.10: 87505506-87505506 on Build GRCh38
rs780542979 Chr.10: 87505451-87505451 on Build GRCh38
rs749493424 Chr.10: 87505512-87505512 on Build GRCh38
rs753137200 Chr.10: 87505574-87505574 on Build GRCh38
rs941661899 Chr.10: 87505660-87505660 on Build GRCh38
rs1055397121 Chr.10: 87505674-87505674 on Build GRCh38
rs1047892591 Chr.10: 87505642-87505642 on Build GRCh38
rs759333132 Chr.10: 87505549-87505549 on Build GRCh38
rs775276659 Chr.10: 87505557-87505557 on Build GRCh38
rs975866020 Chr.10: 87505647-87505647 on Build GRCh38
rs749766225 Chr.10: 87505517-87505517 on Build GRCh38
rs779122692 Chr.10: 87505595-87505595 on Build GRCh38
rs751902526 Chr.10: 87505569-87505569 on Build GRCh38
rs757407631 Chr.10: 87505588-87505588 on Build GRCh38
rs556450511 Chr.10: 87505575-87505575 on Build GRCh38
rs1048772565 Chr.10: 87505593-87505593 on Build GRCh38
rs537804422 Chr.10: 87505572-87505572 on Build GRCh38
rs752187297 Chr.10: 87505434-87505434 on Build GRCh38
rs751994114 Chr.10: 87505453-87505453 on Build GRCh38

View Full Product Details