Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38738837-38739380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCGF2
Entrez Gene ID
7703
Gene Name
polycomb group ring finger 2
Gene Aliases
MEL-18, RNF110, ZNF144
UniGene
Hs.371617
Gene Chromosome Location
Chr.17: 38733897-38749817 on Build GRCh38
Transcripts
NM_007144.2 XM_005257640.2 XM_005257641.4 XM_005257642.3 XM_017025016.1

SNP Detail

SNP ID Chromosome Location
rs775215849 Chr.17: 38739165-38739165 on Build GRCh38
rs777304508 Chr.17: 38739019-38739019 on Build GRCh38
rs754174211 Chr.17: 38739245-38739245 on Build GRCh38
rs759394608 Chr.17: 38739073-38739073 on Build GRCh38
rs764992830 Chr.17: 38739080-38739080 on Build GRCh38
rs755754476 Chr.17: 38739161-38739162 on Build GRCh38
rs759437365 Chr.17: 38739159-38739159 on Build GRCh38
rs746360625 Chr.17: 38739100-38739100 on Build GRCh38
rs760633828 Chr.17: 38738883-38738883 on Build GRCh38
rs906535176 Chr.17: 38738948-38738948 on Build GRCh38
rs185769246 Chr.17: 38739305-38739305 on Build GRCh38
rs768663393 Chr.17: 38739059-38739059 on Build GRCh38
rs146768937 Chr.17: 38739263-38739263 on Build GRCh38
rs762525111 Chr.17: 38739169-38739169 on Build GRCh38
rs940142836 Chr.17: 38739125-38739125 on Build GRCh38
rs763533799 Chr.17: 38739085-38739085 on Build GRCh38
rs567662585 Chr.17: 38738981-38738981 on Build GRCh38
rs2075056 Chr.17: 38738953-38738953 on Build GRCh38
rs72819704 Chr.17: 38739261-38739261 on Build GRCh38
rs772136069 Chr.17: 38739153-38739153 on Build GRCh38
rs967179616 Chr.17: 38739162-38739162 on Build GRCh38
rs200480241 Chr.17: 38739101-38739101 on Build GRCh38
rs367547372 Chr.17: 38738891-38738891 on Build GRCh38
rs370499226 Chr.17: 38739272-38739272 on Build GRCh38
rs1002771757 Chr.17: 38738890-38738890 on Build GRCh38
rs1004433056 Chr.17: 38738871-38738871 on Build GRCh38
rs776609583 Chr.17: 38739155-38739155 on Build GRCh38
rs779455889 Chr.17: 38738942-38738942 on Build GRCh38
rs970896814 Chr.17: 38738875-38738875 on Build GRCh38
rs767116828 Chr.17: 38739190-38739190 on Build GRCh38
rs764805477 Chr.17: 38738895-38738895 on Build GRCh38
rs748192489 Chr.17: 38739143-38739143 on Build GRCh38
rs762948579 Chr.17: 38739361-38739361 on Build GRCh38
rs1025660565 Chr.17: 38738873-38738873 on Build GRCh38
rs1046420361 Chr.17: 38738944-38738944 on Build GRCh38
rs770081160 Chr.17: 38738860-38738860 on Build GRCh38
rs931344713 Chr.17: 38738968-38738968 on Build GRCh38
rs374299797 Chr.17: 38739300-38739300 on Build GRCh38
rs754776377 Chr.17: 38739127-38739128 on Build GRCh38
rs906538547 Chr.17: 38739349-38739349 on Build GRCh38
rs756653107 Chr.17: 38739198-38739198 on Build GRCh38
rs940088742 Chr.17: 38738939-38738939 on Build GRCh38
rs975642404 Chr.17: 38738959-38738959 on Build GRCh38
rs113061894 Chr.17: 38739299-38739299 on Build GRCh38
rs751313090 Chr.17: 38739035-38739036 on Build GRCh38
rs774454060 Chr.17: 38739061-38739061 on Build GRCh38
rs751009670 Chr.17: 38739193-38739193 on Build GRCh38
rs752652614 Chr.17: 38739145-38739146 on Build GRCh38
rs373711829 Chr.17: 38738865-38738865 on Build GRCh38
rs143648170 Chr.17: 38739163-38739163 on Build GRCh38
rs371972755 Chr.17: 38739304-38739304 on Build GRCh38
rs770275381 Chr.17: 38739054-38739054 on Build GRCh38
rs900891184 Chr.17: 38739293-38739293 on Build GRCh38
rs71384265 Chr.17: 38739286-38739286 on Build GRCh38
rs372347370 Chr.17: 38738952-38738952 on Build GRCh38
rs551103547 Chr.17: 38738978-38738978 on Build GRCh38
rs148178156 Chr.17: 38739174-38739174 on Build GRCh38
rs775149127 Chr.17: 38739291-38739291 on Build GRCh38
rs752343314 Chr.17: 38738908-38738908 on Build GRCh38
rs530992009 Chr.17: 38738977-38738977 on Build GRCh38
rs547034851 Chr.17: 38739022-38739022 on Build GRCh38
rs894023400 Chr.17: 38738964-38738964 on Build GRCh38
rs763189159 Chr.17: 38738882-38738882 on Build GRCh38
rs758029170 Chr.17: 38739084-38739084 on Build GRCh38
rs780536842 Chr.17: 38739103-38739103 on Build GRCh38
rs931146937 Chr.17: 38739275-38739275 on Build GRCh38
rs757730739 Chr.17: 38739042-38739042 on Build GRCh38
rs1011253528 Chr.17: 38739082-38739082 on Build GRCh38
rs749506365 Chr.17: 38739056-38739056 on Build GRCh38
rs776417863 Chr.17: 38739072-38739072 on Build GRCh38
rs367961651 Chr.17: 38739053-38739053 on Build GRCh38
rs766283137 Chr.17: 38739065-38739065 on Build GRCh38
rs1021214050 Chr.17: 38738918-38738918 on Build GRCh38
rs749508532 Chr.17: 38739104-38739104 on Build GRCh38
rs761441171 Chr.17: 38739180-38739180 on Build GRCh38
rs779263814 Chr.17: 38739258-38739258 on Build GRCh38
rs754208530 Chr.17: 38738888-38738888 on Build GRCh38
rs1046472821 Chr.17: 38739141-38739141 on Build GRCh38
rs371390028 Chr.17: 38738910-38738910 on Build GRCh38
rs940034499 Chr.17: 38739011-38739011 on Build GRCh38
rs751019545 Chr.17: 38739087-38739087 on Build GRCh38
rs371103302 Chr.17: 38739246-38739246 on Build GRCh38
rs142119022 Chr.17: 38738958-38738958 on Build GRCh38
rs757705562 Chr.17: 38739093-38739093 on Build GRCh38
rs752386015 Chr.17: 38739081-38739081 on Build GRCh38
rs144094243 Chr.17: 38739177-38739177 on Build GRCh38
rs1021526869 Chr.17: 38738975-38738975 on Build GRCh38
rs530159090 Chr.17: 38739003-38739003 on Build GRCh38
rs768320356 Chr.17: 38739298-38739298 on Build GRCh38
rs1046416363 Chr.17: 38739036-38739036 on Build GRCh38
rs374183574 Chr.17: 38739055-38739055 on Build GRCh38
rs772085100 Chr.17: 38738965-38738965 on Build GRCh38
rs574550228 Chr.17: 38738941-38738941 on Build GRCh38
rs780159639 Chr.17: 38739117-38739117 on Build GRCh38
rs755160782 Chr.17: 38739257-38739257 on Build GRCh38
rs965324493 Chr.17: 38738857-38738857 on Build GRCh38
rs374946636 Chr.17: 38739048-38739048 on Build GRCh38
rs887076588 Chr.17: 38738998-38738998 on Build GRCh38
rs781298870 Chr.17: 38739126-38739127 on Build GRCh38
rs762869680 Chr.17: 38739297-38739297 on Build GRCh38
rs912452623 Chr.17: 38738945-38738945 on Build GRCh38
rs148936414 Chr.17: 38738936-38738936 on Build GRCh38
rs780591825 Chr.17: 38739217-38739217 on Build GRCh38
rs565174834 Chr.17: 38738958-38738958 on Build GRCh38
rs772178610 Chr.17: 38739262-38739262 on Build GRCh38
rs781780710 Chr.17: 38739099-38739099 on Build GRCh38
rs760553044 Chr.17: 38739063-38739063 on Build GRCh38
rs768877758 Chr.17: 38739107-38739107 on Build GRCh38
rs753581185 Chr.17: 38738889-38738889 on Build GRCh38
rs565438724 Chr.17: 38739310-38739310 on Build GRCh38

View Full Product Details