Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 10471541-10472131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APCDD1
Entrez Gene ID
147495
Gene Name
APC down-regulated 1
Gene Aliases
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1
UniGene
Hs.293274
Gene Chromosome Location
Chr.18: 10454628-10488701 on Build GRCh38
Transcripts
NM_153000.4 XM_011525603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
708461 c.515C>T p.P172L Chr.18: 10471802-10471802 on Build GRCh38
6148280 c.662A>G p.H221R Chr.18: 10471949-10471949 on Build GRCh38
4071251 c.531C>T p.D177D Chr.18: 10471818-10471818 on Build GRCh38
417892 c.373C>T p.R125W Chr.18: 10471660-10471660 on Build GRCh38
4972848 c.538C>T p.P180S Chr.18: 10471825-10471825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758710972 Chr.18: 10471810-10471810 on Build GRCh38
rs759075518 Chr.18: 10471640-10471640 on Build GRCh38
rs768822914 Chr.18: 10471675-10471675 on Build GRCh38
rs745500767 Chr.18: 10471841-10471841 on Build GRCh38
rs770761989 Chr.18: 10472037-10472037 on Build GRCh38
rs116606798 Chr.18: 10471760-10471760 on Build GRCh38
rs763049478 Chr.18: 10471957-10471957 on Build GRCh38
rs149668434 Chr.18: 10471870-10471870 on Build GRCh38
rs761157830 Chr.18: 10471847-10471847 on Build GRCh38
rs918145531 Chr.18: 10471839-10471839 on Build GRCh38
rs150507107 Chr.18: 10471760-10471761 on Build GRCh38
rs766947286 Chr.18: 10471858-10471858 on Build GRCh38
rs117044576 Chr.18: 10471627-10471627 on Build GRCh38
rs201504676 Chr.18: 10471945-10471945 on Build GRCh38
rs537329264 Chr.18: 10471968-10471968 on Build GRCh38
rs3748415 Chr.18: 10471735-10471735 on Build GRCh38
rs750931054 Chr.18: 10471717-10471717 on Build GRCh38
rs374768897 Chr.18: 10471641-10471641 on Build GRCh38
rs755163934 Chr.18: 10471740-10471740 on Build GRCh38
rs372282253 Chr.18: 10472035-10472035 on Build GRCh38
rs897890762 Chr.18: 10472021-10472021 on Build GRCh38
rs774471766 Chr.18: 10472079-10472079 on Build GRCh38
rs771865619 Chr.18: 10471836-10471836 on Build GRCh38
rs780321900 Chr.18: 10471664-10471664 on Build GRCh38
rs1057289531 Chr.18: 10472026-10472026 on Build GRCh38
rs3748414 Chr.18: 10471767-10471767 on Build GRCh38
rs762629902 Chr.18: 10471651-10471651 on Build GRCh38
rs555203529 Chr.18: 10471678-10471678 on Build GRCh38
rs769615162 Chr.18: 10471906-10471906 on Build GRCh38
rs754806469 Chr.18: 10472006-10472006 on Build GRCh38
rs778943965 Chr.18: 10472013-10472013 on Build GRCh38
rs754683546 Chr.18: 10471561-10471570 on Build GRCh38
rs936793002 Chr.18: 10472072-10472072 on Build GRCh38
rs748706455 Chr.18: 10472024-10472024 on Build GRCh38
rs1007379968 Chr.18: 10471985-10471985 on Build GRCh38
rs150698120 Chr.18: 10471803-10471803 on Build GRCh38
rs751758151 Chr.18: 10471988-10471988 on Build GRCh38
rs956536247 Chr.18: 10471896-10471896 on Build GRCh38
rs1036419184 Chr.18: 10472005-10472005 on Build GRCh38
rs749572726 Chr.18: 10471616-10471616 on Build GRCh38
rs764251642 Chr.18: 10471963-10471963 on Build GRCh38
rs770157936 Chr.18: 10472063-10472063 on Build GRCh38
rs770865417 Chr.18: 10471692-10471692 on Build GRCh38
rs780068149 Chr.18: 10472082-10472082 on Build GRCh38
rs537794700 Chr.18: 10471700-10471700 on Build GRCh38
rs571030647 Chr.18: 10472085-10472085 on Build GRCh38
rs373747410 Chr.18: 10471699-10471699 on Build GRCh38
rs3748413 Chr.18: 10471776-10471776 on Build GRCh38
rs779865322 Chr.18: 10471900-10471900 on Build GRCh38
rs746711951 Chr.18: 10471925-10471925 on Build GRCh38
rs746650863 Chr.18: 10472028-10472028 on Build GRCh38
rs200141705 Chr.18: 10471629-10471629 on Build GRCh38
rs1030269928 Chr.18: 10471953-10471953 on Build GRCh38
rs368209332 Chr.18: 10471999-10471999 on Build GRCh38
rs777408847 Chr.18: 10472013-10472015 on Build GRCh38
rs775104994 Chr.18: 10471915-10471915 on Build GRCh38
rs757770933 Chr.18: 10471572-10471572 on Build GRCh38
rs780949022 Chr.18: 10471811-10471811 on Build GRCh38
rs753715344 Chr.18: 10471905-10471905 on Build GRCh38
rs376484896 Chr.18: 10471907-10471907 on Build GRCh38
rs372913393 Chr.18: 10471662-10471662 on Build GRCh38
rs758557404 Chr.18: 10471882-10471882 on Build GRCh38
rs776087296 Chr.18: 10472067-10472067 on Build GRCh38
rs751927102 Chr.18: 10471656-10471656 on Build GRCh38
rs376738259 Chr.18: 10471713-10471713 on Build GRCh38
rs866722203 Chr.18: 10471592-10471592 on Build GRCh38
rs767252274 Chr.18: 10471787-10471796 on Build GRCh38
rs541503972 Chr.18: 10471755-10471755 on Build GRCh38
rs549800578 Chr.18: 10472041-10472041 on Build GRCh38
rs781318162 Chr.18: 10471584-10471584 on Build GRCh38
rs771812570 Chr.18: 10471920-10471920 on Build GRCh38
rs563904768 Chr.18: 10471861-10471863 on Build GRCh38
rs371385019 Chr.18: 10471604-10471604 on Build GRCh38
rs762580615 Chr.18: 10471709-10471709 on Build GRCh38
rs145453912 Chr.18: 10471890-10471890 on Build GRCh38
rs773647477 Chr.18: 10471840-10471840 on Build GRCh38
rs775879731 Chr.18: 10471614-10471614 on Build GRCh38
rs773772057 Chr.18: 10471784-10471784 on Build GRCh38
rs777594465 Chr.18: 10471924-10471924 on Build GRCh38
rs776568911 Chr.18: 10471698-10471698 on Build GRCh38
rs377223916 Chr.18: 10471672-10471672 on Build GRCh38
rs558862972 Chr.18: 10471722-10471722 on Build GRCh38
rs777116492 Chr.18: 10471935-10471935 on Build GRCh38
rs768313678 Chr.18: 10471772-10471772 on Build GRCh38
rs762091049 Chr.18: 10471996-10471996 on Build GRCh38
rs979177506 Chr.18: 10471865-10471865 on Build GRCh38
rs767785802 Chr.18: 10472109-10472109 on Build GRCh38
rs370464025 Chr.18: 10471679-10471679 on Build GRCh38
rs550080767 Chr.18: 10471793-10471793 on Build GRCh38
rs775364287 Chr.18: 10471956-10471956 on Build GRCh38
rs781000361 Chr.18: 10471761-10471761 on Build GRCh38
rs774923279 Chr.18: 10471769-10471769 on Build GRCh38
rs755924933 Chr.18: 10471891-10471891 on Build GRCh38
rs775044078 Chr.18: 10471702-10471702 on Build GRCh38
rs149506112 Chr.18: 10471818-10471818 on Build GRCh38
rs369975485 Chr.18: 10471617-10471617 on Build GRCh38
rs1023294640 Chr.18: 10471831-10471831 on Build GRCh38
rs757722641 Chr.18: 10471659-10471659 on Build GRCh38
rs117942846 Chr.18: 10472077-10472077 on Build GRCh38
rs752102915 Chr.18: 10472023-10472023 on Build GRCh38
rs186616216 Chr.18: 10471661-10471661 on Build GRCh38
rs767992023 Chr.18: 10471780-10471780 on Build GRCh38
rs770344284 Chr.18: 10471948-10471948 on Build GRCh38
rs199591627 Chr.18: 10471861-10471861 on Build GRCh38
rs778044009 Chr.18: 10471562-10471562 on Build GRCh38
rs747745113 Chr.18: 10471824-10471824 on Build GRCh38
rs772950548 Chr.18: 10471626-10471626 on Build GRCh38
rs529777826 Chr.18: 10471869-10471869 on Build GRCh38
rs916340314 Chr.18: 10471660-10471660 on Build GRCh38
rs751883282 Chr.18: 10471714-10471714 on Build GRCh38
rs114674646 Chr.18: 10471859-10471859 on Build GRCh38
rs368134493 Chr.18: 10471819-10471819 on Build GRCh38
rs778230094 Chr.18: 10471912-10471912 on Build GRCh38
rs763499384 Chr.18: 10472076-10472076 on Build GRCh38
rs889016458 Chr.18: 10471959-10471959 on Build GRCh38
rs771387300 Chr.18: 10471927-10471927 on Build GRCh38
rs200405895 Chr.18: 10471792-10471792 on Build GRCh38
rs948231698 Chr.18: 10471851-10471851 on Build GRCh38
rs767000329 Chr.18: 10471796-10471796 on Build GRCh38
rs764514239 Chr.18: 10471653-10471653 on Build GRCh38
rs375515191 Chr.18: 10471607-10471607 on Build GRCh38
rs748750060 Chr.18: 10472096-10472096 on Build GRCh38
rs747515881 Chr.18: 10471568-10471568 on Build GRCh38
rs773854266 Chr.18: 10471773-10471773 on Build GRCh38
rs752826847 Chr.18: 10471805-10471805 on Build GRCh38
rs143954637 Chr.18: 10471802-10471802 on Build GRCh38
rs780268952 Chr.18: 10471734-10471734 on Build GRCh38
rs755754147 Chr.18: 10471981-10471982 on Build GRCh38
rs756242200 Chr.18: 10471663-10471663 on Build GRCh38
rs752320687 Chr.18: 10471647-10471647 on Build GRCh38
rs115383241 Chr.18: 10471813-10471813 on Build GRCh38
rs370392635 Chr.18: 10471797-10471797 on Build GRCh38
rs116328641 Chr.18: 10471768-10471768 on Build GRCh38
rs139232963 Chr.18: 10472001-10472001 on Build GRCh38
rs975887566 Chr.18: 10471903-10471903 on Build GRCh38
rs980981047 Chr.18: 10472061-10472061 on Build GRCh38

View Full Product Details