Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2038374-2038956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MKNK2
Entrez Gene ID
2872
Gene Name
MAP kinase interacting serine/threonine kinase 2
Gene Aliases
GPRK7, MNK2
UniGene
Hs.515032
Gene Chromosome Location
Chr.19: 2037471-2051244 on Build GRCh38
Transcripts
NM_017572.3 NM_199054.2

SNP Detail

SNP ID Chromosome Location
rs904463719 Chr.19: 2038673-2038673 on Build GRCh38
rs576851473 Chr.19: 2038472-2038472 on Build GRCh38
rs1013872173 Chr.19: 2038532-2038532 on Build GRCh38
rs754805029 Chr.19: 2038629-2038630 on Build GRCh38
rs1013924727 Chr.19: 2038687-2038687 on Build GRCh38
rs576448332 Chr.19: 2038721-2038721 on Build GRCh38
rs943378207 Chr.19: 2038661-2038661 on Build GRCh38
rs2287002 Chr.19: 2038820-2038820 on Build GRCh38
rs113813463 Chr.19: 2038827-2038827 on Build GRCh38
rs551323245 Chr.19: 2038912-2038912 on Build GRCh38
rs763447149 Chr.19: 2038632-2038632 on Build GRCh38
rs969704115 Chr.19: 2038591-2038591 on Build GRCh38
rs752648309 Chr.19: 2038641-2038641 on Build GRCh38
rs933907265 Chr.19: 2038937-2038937 on Build GRCh38
rs200666864 Chr.19: 2038818-2038821 on Build GRCh38
rs543562122 Chr.19: 2038804-2038804 on Build GRCh38
rs904327297 Chr.19: 2038524-2038524 on Build GRCh38
rs12151339 Chr.19: 2038460-2038460 on Build GRCh38
rs918318521 Chr.19: 2038540-2038540 on Build GRCh38
rs1048363032 Chr.19: 2038412-2038412 on Build GRCh38
rs1052973353 Chr.19: 2038826-2038826 on Build GRCh38
rs987558431 Chr.19: 2038643-2038643 on Build GRCh38
rs994124399 Chr.19: 2038934-2038934 on Build GRCh38
rs929695457 Chr.19: 2038676-2038676 on Build GRCh38
rs895599194 Chr.19: 2038529-2038529 on Build GRCh38
rs896822568 Chr.19: 2038905-2038905 on Build GRCh38
rs988743733 Chr.19: 2038478-2038478 on Build GRCh38
rs557496665 Chr.19: 2038531-2038531 on Build GRCh38
rs568179112 Chr.19: 2038450-2038450 on Build GRCh38
rs954869331 Chr.19: 2038801-2038801 on Build GRCh38
rs866126643 Chr.19: 2038871-2038871 on Build GRCh38
rs965197945 Chr.19: 2038459-2038459 on Build GRCh38
rs556605202 Chr.19: 2038691-2038691 on Build GRCh38
rs34824515 Chr.19: 2038690-2038690 on Build GRCh38
rs536760294 Chr.19: 2038633-2038633 on Build GRCh38
rs556674656 Chr.19: 2038645-2038645 on Build GRCh38
rs891919620 Chr.19: 2038796-2038796 on Build GRCh38
rs181895797 Chr.19: 2038701-2038701 on Build GRCh38
rs1053400479 Chr.19: 2038683-2038683 on Build GRCh38
rs927496279 Chr.19: 2038668-2038668 on Build GRCh38
rs534749032 Chr.19: 2038642-2038642 on Build GRCh38
rs559664322 Chr.19: 2038864-2038864 on Build GRCh38
rs981017624 Chr.19: 2038598-2038598 on Build GRCh38
rs993680604 Chr.19: 2038755-2038755 on Build GRCh38
rs531744334 Chr.19: 2038399-2038399 on Build GRCh38
rs910658921 Chr.19: 2038805-2038805 on Build GRCh38
rs546159408 Chr.19: 2038692-2038692 on Build GRCh38
rs1038484621 Chr.19: 2038904-2038904 on Build GRCh38
rs905635443 Chr.19: 2038868-2038868 on Build GRCh38
rs1026446680 Chr.19: 2038759-2038759 on Build GRCh38
rs949905070 Chr.19: 2038857-2038857 on Build GRCh38
rs143739660 Chr.19: 2038501-2038501 on Build GRCh38
rs143988124 Chr.19: 2038623-2038623 on Build GRCh38
rs963557598 Chr.19: 2038605-2038605 on Build GRCh38
rs377433648 Chr.19: 2038502-2038502 on Build GRCh38
rs73916836 Chr.19: 2038811-2038811 on Build GRCh38
rs1016501445 Chr.19: 2038710-2038710 on Build GRCh38
rs375302836 Chr.19: 2038865-2038865 on Build GRCh38
rs1031401699 Chr.19: 2038528-2038528 on Build GRCh38
rs569556609 Chr.19: 2038914-2038914 on Build GRCh38
rs901056045 Chr.19: 2038777-2038777 on Build GRCh38
rs888390566 Chr.19: 2038601-2038601 on Build GRCh38
rs139727878 Chr.19: 2038499-2038499 on Build GRCh38
rs529161642 Chr.19: 2038451-2038451 on Build GRCh38
rs999113110 Chr.19: 2038525-2038525 on Build GRCh38
rs1016944831 Chr.19: 2038604-2038604 on Build GRCh38
rs917051962 Chr.19: 2038854-2038854 on Build GRCh38
rs191074909 Chr.19: 2038674-2038674 on Build GRCh38
rs933293347 Chr.19: 2038484-2038484 on Build GRCh38
rs993022196 Chr.19: 2038836-2038836 on Build GRCh38
rs1022546274 Chr.19: 2038535-2038535 on Build GRCh38
rs1051800546 Chr.19: 2038486-2038486 on Build GRCh38
rs979200923 Chr.19: 2038819-2038819 on Build GRCh38
rs549920236 Chr.19: 2038430-2038430 on Build GRCh38
rs1009645115 Chr.19: 2038897-2038916 on Build GRCh38
rs748498645 Chr.19: 2038473-2038473 on Build GRCh38
rs547719195 Chr.19: 2038468-2038468 on Build GRCh38
rs1018089699 Chr.19: 2038454-2038454 on Build GRCh38
rs981433218 Chr.19: 2038428-2038428 on Build GRCh38
rs1052201657 Chr.19: 2038795-2038795 on Build GRCh38
rs75579352 Chr.19: 2038821-2038821 on Build GRCh38
rs963999180 Chr.19: 2038737-2038737 on Build GRCh38
rs933867039 Chr.19: 2038786-2038786 on Build GRCh38
rs1021577152 Chr.19: 2038657-2038657 on Build GRCh38
rs922567132 Chr.19: 2038932-2038932 on Build GRCh38
rs1039628071 Chr.19: 2038704-2038704 on Build GRCh38
rs533287196 Chr.19: 2038908-2038908 on Build GRCh38
rs773584498 Chr.19: 2038602-2038602 on Build GRCh38
rs112045798 Chr.19: 2038780-2038780 on Build GRCh38
rs929643294 Chr.19: 2038590-2038590 on Build GRCh38
rs998388422 Chr.19: 2038637-2038637 on Build GRCh38
rs142187689 Chr.19: 2038684-2038684 on Build GRCh38
rs574633835 Chr.19: 2038672-2038672 on Build GRCh38
rs770387310 Chr.19: 2038517-2038517 on Build GRCh38
rs148663343 Chr.19: 2038662-2038662 on Build GRCh38

View Full Product Details