Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45494386-45494967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTN2
Entrez Gene ID
6253
Gene Name
reticulon 2
Gene Aliases
NSP2, NSPL1, NSPLI, SPG12
UniGene
Hs.47517
Gene Chromosome Location
Chr.19: 45485288-45497055 on Build GRCh38
Transcripts
NM_005619.4 NM_206900.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6026227 c.351A>G p.Q117Q Chr.19: 45494734-45494734 on Build GRCh38
3389140 c.201C>T p.I67I Chr.19: 45494884-45494884 on Build GRCh38
5464629 c.150G>T p.E50D Chr.19: 45494935-45494935 on Build GRCh38
6870591 c.122G>A p.R41Q Chr.19: 45494963-45494963 on Build GRCh38
4427915 c.277G>T p.V93F Chr.19: 45494808-45494808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776994539 Chr.19: 45494711-45494711 on Build GRCh38
rs750150381 Chr.19: 45494826-45494826 on Build GRCh38
rs763387041 Chr.19: 45494906-45494907 on Build GRCh38
rs142118512 Chr.19: 45494725-45494725 on Build GRCh38
rs755957152 Chr.19: 45494718-45494718 on Build GRCh38
rs771059263 Chr.19: 45494855-45494855 on Build GRCh38
rs763920905 Chr.19: 45494729-45494729 on Build GRCh38
rs768375312 Chr.19: 45494431-45494431 on Build GRCh38
rs759898844 Chr.19: 45494617-45494617 on Build GRCh38
rs767779372 Chr.19: 45494409-45494409 on Build GRCh38
rs764402044 Chr.19: 45494518-45494518 on Build GRCh38
rs769157154 Chr.19: 45494766-45494766 on Build GRCh38
rs770628974 Chr.19: 45494603-45494603 on Build GRCh38
rs773521863 Chr.19: 45494784-45494784 on Build GRCh38
rs895596505 Chr.19: 45494804-45494804 on Build GRCh38
rs747332877 Chr.19: 45494699-45494699 on Build GRCh38
rs768686847 Chr.19: 45494871-45494871 on Build GRCh38
rs144028452 Chr.19: 45494734-45494734 on Build GRCh38
rs755706051 Chr.19: 45494945-45494947 on Build GRCh38
rs149153041 Chr.19: 45494813-45494813 on Build GRCh38
rs765729301 Chr.19: 45494450-45494451 on Build GRCh38
rs532731946 Chr.19: 45494687-45494687 on Build GRCh38
rs769517671 Chr.19: 45494443-45494443 on Build GRCh38
rs760772669 Chr.19: 45494515-45494515 on Build GRCh38
rs560858936 Chr.19: 45494580-45494580 on Build GRCh38
rs774445918 Chr.19: 45494862-45494862 on Build GRCh38
rs876657982 Chr.19: 45494421-45494421 on Build GRCh38
rs370068938 Chr.19: 45494427-45494427 on Build GRCh38
rs753115199 Chr.19: 45494503-45494503 on Build GRCh38
rs570062973 Chr.19: 45494682-45494682 on Build GRCh38
rs765475323 Chr.19: 45494712-45494712 on Build GRCh38
rs779963520 Chr.19: 45494531-45494531 on Build GRCh38
rs866720252 Chr.19: 45494540-45494540 on Build GRCh38
rs761153006 Chr.19: 45494624-45494624 on Build GRCh38
rs753894861 Chr.19: 45494519-45494519 on Build GRCh38
rs755974615 Chr.19: 45494679-45494679 on Build GRCh38
rs139232850 Chr.19: 45494641-45494641 on Build GRCh38
rs754128612 Chr.19: 45494633-45494633 on Build GRCh38
rs913184423 Chr.19: 45494737-45494737 on Build GRCh38
rs762572203 Chr.19: 45494464-45494464 on Build GRCh38
rs752411639 Chr.19: 45494661-45494661 on Build GRCh38
rs762935871 Chr.19: 45494788-45494788 on Build GRCh38
rs780784322 Chr.19: 45494550-45494550 on Build GRCh38
rs771540486 Chr.19: 45494466-45494466 on Build GRCh38
rs764272171 Chr.19: 45494629-45494629 on Build GRCh38
rs947260810 Chr.19: 45494462-45494462 on Build GRCh38
rs751046301 Chr.19: 45494922-45494922 on Build GRCh38
rs866666196 Chr.19: 45494791-45494791 on Build GRCh38
rs752906428 Chr.19: 45494416-45494416 on Build GRCh38
rs779322568 Chr.19: 45494946-45494946 on Build GRCh38
rs750191809 Chr.19: 45494890-45494890 on Build GRCh38
rs776780195 Chr.19: 45494883-45494883 on Build GRCh38
rs780022596 Chr.19: 45494429-45494429 on Build GRCh38
rs761869759 Chr.19: 45494634-45494634 on Build GRCh38
rs146004780 Chr.19: 45494800-45494800 on Build GRCh38
rs1057465223 Chr.19: 45494914-45494914 on Build GRCh38
rs774304208 Chr.19: 45494796-45494796 on Build GRCh38
rs140389710 Chr.19: 45494714-45494714 on Build GRCh38
rs747706464 Chr.19: 45494435-45494435 on Build GRCh38
rs113581535 Chr.19: 45494790-45494790 on Build GRCh38
rs748534400 Chr.19: 45494769-45494769 on Build GRCh38
rs766535005 Chr.19: 45494795-45494795 on Build GRCh38
rs751101450 Chr.19: 45494832-45494832 on Build GRCh38
rs772669907 Chr.19: 45494622-45494622 on Build GRCh38
rs372925067 Chr.19: 45494901-45494901 on Build GRCh38
rs756925105 Chr.19: 45494731-45494731 on Build GRCh38
rs751557271 Chr.19: 45494536-45494536 on Build GRCh38
rs775947644 Chr.19: 45494904-45494904 on Build GRCh38
rs769980951 Chr.19: 45494777-45494777 on Build GRCh38
rs750521507 Chr.19: 45494639-45494639 on Build GRCh38
rs561232438 Chr.19: 45494549-45494549 on Build GRCh38
rs150654638 Chr.19: 45494728-45494728 on Build GRCh38
rs750037372 Chr.19: 45494936-45494944 on Build GRCh38
rs775672769 Chr.19: 45494623-45494623 on Build GRCh38
rs757996406 Chr.19: 45494748-45494748 on Build GRCh38
rs767826865 Chr.19: 45494490-45494490 on Build GRCh38
rs765970940 Chr.19: 45494915-45494915 on Build GRCh38
rs77577960 Chr.19: 45494926-45494926 on Build GRCh38
rs746800975 Chr.19: 45494430-45494430 on Build GRCh38
rs1020504637 Chr.19: 45494823-45494823 on Build GRCh38
rs772248018 Chr.19: 45494559-45494559 on Build GRCh38
rs769280878 Chr.19: 45494581-45494581 on Build GRCh38
rs373315487 Chr.19: 45494799-45494799 on Build GRCh38
rs753666898 Chr.19: 45494816-45494816 on Build GRCh38
rs149317318 Chr.19: 45494530-45494530 on Build GRCh38
rs1042862083 Chr.19: 45494509-45494509 on Build GRCh38
rs541422215 Chr.19: 45494434-45494434 on Build GRCh38
rs767238040 Chr.19: 45494935-45494937 on Build GRCh38
rs144198343 Chr.19: 45494758-45494758 on Build GRCh38
rs745342926 Chr.19: 45494747-45494747 on Build GRCh38
rs775035353 Chr.19: 45494481-45494481 on Build GRCh38
rs550254965 Chr.19: 45494648-45494648 on Build GRCh38
rs371978046 Chr.19: 45494658-45494658 on Build GRCh38
rs200744554 Chr.19: 45494428-45494428 on Build GRCh38
rs749642332 Chr.19: 45494938-45494938 on Build GRCh38
rs367898848 Chr.19: 45494637-45494637 on Build GRCh38
rs368336082 Chr.19: 45494715-45494715 on Build GRCh38
rs753687079 Chr.19: 45494730-45494730 on Build GRCh38
rs377571193 Chr.19: 45494848-45494848 on Build GRCh38
rs375715266 Chr.19: 45494943-45494943 on Build GRCh38
rs778128536 Chr.19: 45494932-45494932 on Build GRCh38
rs758002007 Chr.19: 45494828-45494828 on Build GRCh38
rs1025906520 Chr.19: 45494668-45494668 on Build GRCh38
rs1000458460 Chr.19: 45494886-45494886 on Build GRCh38
rs554856462 Chr.19: 45494884-45494884 on Build GRCh38
rs138223572 Chr.19: 45494686-45494686 on Build GRCh38
rs143394159 Chr.19: 45494697-45494697 on Build GRCh38
rs772058826 Chr.19: 45494860-45494860 on Build GRCh38
rs774982121 Chr.19: 45494616-45494616 on Build GRCh38
rs748006309 Chr.19: 45494556-45494556 on Build GRCh38
rs143302045 Chr.19: 45494852-45494852 on Build GRCh38
rs375788260 Chr.19: 45494698-45494698 on Build GRCh38
rs777614999 Chr.19: 45494584-45494584 on Build GRCh38
rs866686676 Chr.19: 45494830-45494830 on Build GRCh38
rs759936921 Chr.19: 45494484-45494484 on Build GRCh38
rs922734559 Chr.19: 45494642-45494642 on Build GRCh38
rs559597401 Chr.19: 45494417-45494417 on Build GRCh38
rs147152016 Chr.19: 45494700-45494700 on Build GRCh38
rs781700125 Chr.19: 45494835-45494835 on Build GRCh38
rs752745242 Chr.19: 45494717-45494717 on Build GRCh38
rs1020453743 Chr.19: 45494555-45494555 on Build GRCh38
rs745409162 Chr.19: 45494696-45494696 on Build GRCh38
rs753259137 Chr.19: 45494930-45494930 on Build GRCh38
rs778531345 Chr.19: 45494695-45494695 on Build GRCh38
rs1018513566 Chr.19: 45494888-45494888 on Build GRCh38
rs759743423 Chr.19: 45494866-45494866 on Build GRCh38
rs755428380 Chr.19: 45494833-45494833 on Build GRCh38
rs776941256 Chr.19: 45494767-45494767 on Build GRCh38
rs770208488 Chr.19: 45494850-45494850 on Build GRCh38
rs777670948 Chr.19: 45494418-45494418 on Build GRCh38
rs199928834 Chr.19: 45494911-45494911 on Build GRCh38
rs1009984474 Chr.19: 45494814-45494814 on Build GRCh38
rs968993826 Chr.19: 45494575-45494575 on Build GRCh38
rs907709997 Chr.19: 45494705-45494705 on Build GRCh38
rs772783579 Chr.19: 45494461-45494461 on Build GRCh38
rs556975707 Chr.19: 45494927-45494927 on Build GRCh38

View Full Product Details