Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 177032860-177033268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBL1XR1
Entrez Gene ID
79718
Gene Name
transducin (beta)-like 1 X-linked receptor 1
Gene Aliases
C21, DC42, IRA1, MRD41, TBLR1
UniGene
Hs.714201
Gene Chromosome Location
Chr.3: 177019355-177197357 on Build GRCh38
Transcripts
NM_001321193.1 NM_001321194.1 NM_001321195.1 NM_024665.5 XM_005247775.2 XM_006713746.1 XM_011513141.1 XM_011513142.2 XM_011513143.2 XM_017007185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5045073 c.1267A>C p.T423P Chr.3: 177033120-177033120 on Build GRCh38
7341032 c.1381T>C p.S461P Chr.3: 177033006-177033006 on Build GRCh38
5045122 c.1337A>G p.Y446C Chr.3: 177033050-177033050 on Build GRCh38
7104020 c.1290C>T p.D430D Chr.3: 177033097-177033097 on Build GRCh38
6385481 c.1399C>T p.H467Y Chr.3: 177032988-177032988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779947196 Chr.3: 177033146-177033146 on Build GRCh38
rs144060569 Chr.3: 177032957-177032957 on Build GRCh38
rs781533983 Chr.3: 177032940-177032940 on Build GRCh38
rs77709343 Chr.3: 177033245-177033245 on Build GRCh38
rs969982584 Chr.3: 177033225-177033225 on Build GRCh38
rs16827730 Chr.3: 177032946-177032946 on Build GRCh38
rs942517908 Chr.3: 177033238-177033238 on Build GRCh38
rs755423086 Chr.3: 177032931-177032931 on Build GRCh38
rs776138830 Chr.3: 177032995-177032995 on Build GRCh38
rs749343046 Chr.3: 177032952-177032952 on Build GRCh38
rs62298928 Chr.3: 177033067-177033067 on Build GRCh38
rs200972803 Chr.3: 177033166-177033166 on Build GRCh38
rs762389924 Chr.3: 177033058-177033058 on Build GRCh38
rs112987834 Chr.3: 177033095-177033095 on Build GRCh38
rs766951138 Chr.3: 177033076-177033076 on Build GRCh38
rs749887135 Chr.3: 177033037-177033037 on Build GRCh38
rs771949829 Chr.3: 177032955-177032955 on Build GRCh38
rs748450161 Chr.3: 177032944-177032944 on Build GRCh38
rs768431229 Chr.3: 177033151-177033151 on Build GRCh38
rs1022845331 Chr.3: 177033201-177033201 on Build GRCh38
rs745346385 Chr.3: 177033167-177033168 on Build GRCh38
rs776417431 Chr.3: 177033154-177033154 on Build GRCh38
rs768492558 Chr.3: 177032974-177032974 on Build GRCh38
rs752151434 Chr.3: 177033085-177033085 on Build GRCh38
rs761590111 Chr.3: 177033023-177033023 on Build GRCh38
rs764679582 Chr.3: 177033149-177033149 on Build GRCh38
rs755371824 Chr.3: 177033096-177033096 on Build GRCh38
rs34880754 Chr.3: 177033007-177033007 on Build GRCh38
rs1050780080 Chr.3: 177033024-177033024 on Build GRCh38
rs978912378 Chr.3: 177032900-177032900 on Build GRCh38
rs373306105 Chr.3: 177033121-177033121 on Build GRCh38
rs148654942 Chr.3: 177033144-177033144 on Build GRCh38
rs34878071 Chr.3: 177033072-177033072 on Build GRCh38
rs778600858 Chr.3: 177033143-177033146 on Build GRCh38
rs756302181 Chr.3: 177033120-177033120 on Build GRCh38
rs990612971 Chr.3: 177032951-177032951 on Build GRCh38
rs189143133 Chr.3: 177033199-177033199 on Build GRCh38
rs768051548 Chr.3: 177033115-177033115 on Build GRCh38
rs1003997273 Chr.3: 177033088-177033088 on Build GRCh38
rs764798364 Chr.3: 177033029-177033029 on Build GRCh38
rs772769069 Chr.3: 177033178-177033178 on Build GRCh38
rs769760072 Chr.3: 177032945-177032945 on Build GRCh38
rs564159813 Chr.3: 177033241-177033242 on Build GRCh38
rs766034723 Chr.3: 177033182-177033182 on Build GRCh38
rs185031120 Chr.3: 177032904-177032904 on Build GRCh38
rs759954654 Chr.3: 177032913-177032913 on Build GRCh38
rs878854402 Chr.3: 177033050-177033050 on Build GRCh38
rs1048278295 Chr.3: 177033194-177033194 on Build GRCh38
rs749349045 Chr.3: 177033128-177033128 on Build GRCh38
rs753008616 Chr.3: 177033117-177033117 on Build GRCh38
rs900044554 Chr.3: 177032948-177032948 on Build GRCh38
rs914102124 Chr.3: 177032911-177032911 on Build GRCh38
rs760434778 Chr.3: 177032967-177032967 on Build GRCh38
rs747906715 Chr.3: 177033156-177033156 on Build GRCh38
rs1052929848 Chr.3: 177032907-177032907 on Build GRCh38
rs762519548 Chr.3: 177033181-177033181 on Build GRCh38

View Full Product Details