Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1268222-1268759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TERT
Entrez Gene ID
7015
Gene Name
telomerase reverse transcriptase
Gene Aliases
CMM9, DKCA2, DKCB4, EST2, PFBMFT1, TCS1, TP2, TRT, hEST2, hTRT
UniGene
Hs.492203
Gene Chromosome Location
Chr.5: 1253167-1295626 on Build GRCh38
Transcripts
NM_001193376.1 NM_198253.2 XM_017009796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6546005 c.2567G>A p.G856E Chr.5: 1268535-1268535 on Build GRCh38
6300281 c.2541C>T p.G847G Chr.5: 1268561-1268561 on Build GRCh38
3918554 c.2508C>A p.I836I Chr.5: 1268594-1268594 on Build GRCh38
3918556 c.2472C>T p.S824S Chr.5: 1268630-1268630 on Build GRCh38
6950676 c.2553C>A p.N851K Chr.5: 1268549-1268549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759744531 Chr.5: 1268630-1268630 on Build GRCh38
rs373917530 Chr.5: 1268661-1268661 on Build GRCh38
rs749279645 Chr.5: 1268547-1268547 on Build GRCh38
rs536444159 Chr.5: 1268740-1268740 on Build GRCh38
rs144310369 Chr.5: 1268582-1268582 on Build GRCh38
rs753034642 Chr.5: 1268640-1268640 on Build GRCh38
rs34923115 Chr.5: 1268721-1268721 on Build GRCh38
rs556029118 Chr.5: 1268296-1268296 on Build GRCh38
rs772747367 Chr.5: 1268643-1268643 on Build GRCh38
rs369807360 Chr.5: 1268655-1268655 on Build GRCh38
rs376988566 Chr.5: 1268449-1268449 on Build GRCh38
rs1005469712 Chr.5: 1268310-1268310 on Build GRCh38
rs538359641 Chr.5: 1268698-1268698 on Build GRCh38
rs766810802 Chr.5: 1268500-1268500 on Build GRCh38
rs993409760 Chr.5: 1268686-1268686 on Build GRCh38
rs1040679853 Chr.5: 1268464-1268464 on Build GRCh38
rs993277585 Chr.5: 1268431-1268431 on Build GRCh38
rs375042320 Chr.5: 1268545-1268545 on Build GRCh38
rs897272953 Chr.5: 1268725-1268725 on Build GRCh38
rs138170971 Chr.5: 1268330-1268330 on Build GRCh38
rs1040182158 Chr.5: 1268253-1268253 on Build GRCh38
rs763896942 Chr.5: 1268641-1268642 on Build GRCh38
rs33988305 Chr.5: 1268465-1268465 on Build GRCh38
rs924748176 Chr.5: 1268334-1268334 on Build GRCh38
rs374940572 Chr.5: 1268591-1268591 on Build GRCh38
rs763470933 Chr.5: 1268606-1268606 on Build GRCh38
rs900314049 Chr.5: 1268298-1268298 on Build GRCh38
rs776894859 Chr.5: 1268482-1268482 on Build GRCh38
rs373636164 Chr.5: 1268433-1268433 on Build GRCh38
rs757292450 Chr.5: 1268652-1268652 on Build GRCh38
rs775014633 Chr.5: 1268538-1268538 on Build GRCh38
rs34881188 Chr.5: 1268481-1268482 on Build GRCh38
rs868693799 Chr.5: 1268554-1268554 on Build GRCh38
rs766415474 Chr.5: 1268513-1268513 on Build GRCh38
rs1029677700 Chr.5: 1268584-1268584 on Build GRCh38
rs200350772 Chr.5: 1268491-1268491 on Build GRCh38
rs915422897 Chr.5: 1268621-1268621 on Build GRCh38
rs991503502 Chr.5: 1268302-1268302 on Build GRCh38
rs755942852 Chr.5: 1268641-1268641 on Build GRCh38
rs914992494 Chr.5: 1268384-1268384 on Build GRCh38
rs760293541 Chr.5: 1268672-1268672 on Build GRCh38
rs199422302 Chr.5: 1268565-1268565 on Build GRCh38
rs996408914 Chr.5: 1268488-1268488 on Build GRCh38
rs755183437 Chr.5: 1268526-1268526 on Build GRCh38
rs367785298 Chr.5: 1268684-1268684 on Build GRCh38
rs770091645 Chr.5: 1268685-1268685 on Build GRCh38
rs532660823 Chr.5: 1268452-1268452 on Build GRCh38
rs764029536 Chr.5: 1268642-1268642 on Build GRCh38
rs770543108 Chr.5: 1268561-1268561 on Build GRCh38
rs753781409 Chr.5: 1268645-1268645 on Build GRCh38
rs1006130536 Chr.5: 1268272-1268272 on Build GRCh38
rs933107181 Chr.5: 1268368-1268368 on Build GRCh38
rs149583527 Chr.5: 1268503-1268503 on Build GRCh38
rs751708357 Chr.5: 1268632-1268633 on Build GRCh38
rs780624815 Chr.5: 1268463-1268463 on Build GRCh38
rs876658018 Chr.5: 1268527-1268527 on Build GRCh38
rs1003575407 Chr.5: 1268560-1268560 on Build GRCh38
rs773683541 Chr.5: 1268596-1268596 on Build GRCh38
rs922798759 Chr.5: 1268572-1268572 on Build GRCh38
rs150829248 Chr.5: 1268299-1268299 on Build GRCh38
rs180675821 Chr.5: 1268509-1268509 on Build GRCh38
rs553433898 Chr.5: 1268728-1268728 on Build GRCh38
rs757988387 Chr.5: 1268662-1268662 on Build GRCh38
rs748503447 Chr.5: 1268586-1268586 on Build GRCh38
rs779572012 Chr.5: 1268668-1268668 on Build GRCh38
rs1037065793 Chr.5: 1268724-1268724 on Build GRCh38
rs1006056887 Chr.5: 1268383-1268383 on Build GRCh38
rs6899038 Chr.5: 1268732-1268732 on Build GRCh38
rs1042044844 Chr.5: 1268391-1268391 on Build GRCh38
rs778309923 Chr.5: 1268564-1268564 on Build GRCh38
rs571126114 Chr.5: 1268688-1268688 on Build GRCh38
rs952836416 Chr.5: 1268309-1268309 on Build GRCh38
rs1016018898 Chr.5: 1268273-1268273 on Build GRCh38
rs1048851621 Chr.5: 1268696-1268696 on Build GRCh38
rs144779807 Chr.5: 1268529-1268529 on Build GRCh38
rs770039670 Chr.5: 1268357-1268357 on Build GRCh38
rs565086421 Chr.5: 1268389-1268389 on Build GRCh38
rs765749413 Chr.5: 1268498-1268498 on Build GRCh38
rs34181584 Chr.5: 1268331-1268331 on Build GRCh38
rs542946344 Chr.5: 1268343-1268343 on Build GRCh38
rs764952566 Chr.5: 1268735-1268735 on Build GRCh38
rs895640774 Chr.5: 1268711-1268711 on Build GRCh38
rs766001220 Chr.5: 1268704-1268704 on Build GRCh38
rs576404979 Chr.5: 1268371-1268371 on Build GRCh38
rs753852444 Chr.5: 1268644-1268647 on Build GRCh38
rs140124989 Chr.5: 1268585-1268585 on Build GRCh38
rs756282971 Chr.5: 1268637-1268638 on Build GRCh38
rs972100142 Chr.5: 1268282-1268282 on Build GRCh38
rs538306685 Chr.5: 1268300-1268300 on Build GRCh38
rs961857435 Chr.5: 1268281-1268281 on Build GRCh38
rs368353833 Chr.5: 1268699-1268699 on Build GRCh38
rs543790721 Chr.5: 1268372-1268372 on Build GRCh38
rs141382603 Chr.5: 1268627-1268627 on Build GRCh38
rs35243220 Chr.5: 1268349-1268349 on Build GRCh38
rs767620636 Chr.5: 1268638-1268638 on Build GRCh38
rs759896390 Chr.5: 1268636-1268637 on Build GRCh38
rs572269760 Chr.5: 1268340-1268340 on Build GRCh38
rs991990236 Chr.5: 1268494-1268494 on Build GRCh38
rs771514853 Chr.5: 1268618-1268618 on Build GRCh38
rs751752830 Chr.5: 1268522-1268522 on Build GRCh38
rs897634462 Chr.5: 1268388-1268388 on Build GRCh38
rs959793423 Chr.5: 1268367-1268367 on Build GRCh38
rs769077049 Chr.5: 1268537-1268537 on Build GRCh38

View Full Product Details