Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151055536-151056006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5
Entrez Gene ID
1020
Gene Name
cyclin dependent kinase 5
Gene Aliases
LIS7, PSSALRE
UniGene
Hs.647078
Gene Chromosome Location
Chr.7: 151053812-151057965 on Build GRCh38
Transcripts
NM_001164410.2 NM_004935.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5915637 c.313-8C>T p.? Chr.7: 151055856-151055856 on Build GRCh38
3261433 c.358C>T p.R120C Chr.7: 151055803-151055803 on Build GRCh38
3261430 c.413G>A p.G138E Chr.7: 151055602-151055602 on Build GRCh38
1200564 c.404A>G p.N135S Chr.7: 151055757-151055757 on Build GRCh38
5762045 c.464delT p.V155fs*58 Chr.7: 151055551-151055551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781580336 Chr.7: 151055563-151055563 on Build GRCh38
rs370641796 Chr.7: 151055880-151055880 on Build GRCh38
rs760567025 Chr.7: 151055653-151055653 on Build GRCh38
rs984235218 Chr.7: 151055913-151055913 on Build GRCh38
rs768036739 Chr.7: 151055713-151055713 on Build GRCh38
rs951349142 Chr.7: 151055908-151055908 on Build GRCh38
rs563686216 Chr.7: 151055944-151055944 on Build GRCh38
rs1034801827 Chr.7: 151055847-151055847 on Build GRCh38
rs887032429 Chr.7: 151055674-151055674 on Build GRCh38
rs778524903 Chr.7: 151055746-151055746 on Build GRCh38
rs768225731 Chr.7: 151055631-151055631 on Build GRCh38
rs773188858 Chr.7: 151055649-151055649 on Build GRCh38
rs2069455 Chr.7: 151055733-151055733 on Build GRCh38
rs777882765 Chr.7: 151055774-151055774 on Build GRCh38
rs773110149 Chr.7: 151055952-151055952 on Build GRCh38
rs770097615 Chr.7: 151055703-151055703 on Build GRCh38
rs750272338 Chr.7: 151055597-151055597 on Build GRCh38
rs373178311 Chr.7: 151055803-151055803 on Build GRCh38
rs750749537 Chr.7: 151055719-151055719 on Build GRCh38
rs747917302 Chr.7: 151055629-151055629 on Build GRCh38
rs779714023 Chr.7: 151055617-151055617 on Build GRCh38
rs2069454 Chr.7: 151055895-151055895 on Build GRCh38
rs370113778 Chr.7: 151055768-151055768 on Build GRCh38
rs746730482 Chr.7: 151055799-151055799 on Build GRCh38
rs748013524 Chr.7: 151055749-151055749 on Build GRCh38
rs942782201 Chr.7: 151055939-151055939 on Build GRCh38
rs998425634 Chr.7: 151055664-151055664 on Build GRCh38
rs774444285 Chr.7: 151055845-151055845 on Build GRCh38
rs1047388762 Chr.7: 151055668-151055668 on Build GRCh38
rs369911390 Chr.7: 151055722-151055722 on Build GRCh38
rs753399279 Chr.7: 151055883-151055883 on Build GRCh38
rs892136456 Chr.7: 151055707-151055707 on Build GRCh38
rs376208556 Chr.7: 151055704-151055704 on Build GRCh38
rs567702898 Chr.7: 151055601-151055601 on Build GRCh38
rs749567698 Chr.7: 151055807-151055807 on Build GRCh38
rs753584743 Chr.7: 151055741-151055741 on Build GRCh38
rs146929805 Chr.7: 151055950-151055950 on Build GRCh38
rs754707102 Chr.7: 151055886-151055886 on Build GRCh38
rs757562417 Chr.7: 151055562-151055562 on Build GRCh38
rs771961587 Chr.7: 151055634-151055634 on Build GRCh38
rs907178017 Chr.7: 151055630-151055630 on Build GRCh38
rs971951393 Chr.7: 151055889-151055889 on Build GRCh38
rs1002467832 Chr.7: 151055819-151055819 on Build GRCh38
rs538525166 Chr.7: 151055938-151055938 on Build GRCh38
rs1057766 Chr.7: 151055565-151055565 on Build GRCh38
rs377444178 Chr.7: 151055858-151055858 on Build GRCh38
rs773722553 Chr.7: 151055879-151055879 on Build GRCh38
rs762360856 Chr.7: 151055711-151055711 on Build GRCh38
rs142719014 Chr.7: 151055909-151055909 on Build GRCh38
rs1016715345 Chr.7: 151055716-151055716 on Build GRCh38
rs543004593 Chr.7: 151055914-151055914 on Build GRCh38
rs755717863 Chr.7: 151055604-151055604 on Build GRCh38
rs768598460 Chr.7: 151055822-151055822 on Build GRCh38
rs377502490 Chr.7: 151055744-151055744 on Build GRCh38
rs993525174 Chr.7: 151055662-151055662 on Build GRCh38
rs879355011 Chr.7: 151055724-151055724 on Build GRCh38
rs774693411 Chr.7: 151055708-151055708 on Build GRCh38
rs770858106 Chr.7: 151055802-151055802 on Build GRCh38
rs922758590 Chr.7: 151055960-151055960 on Build GRCh38
rs778704035 Chr.7: 151055632-151055632 on Build GRCh38

View Full Product Details