Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 76704623-76705220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFHX4
Entrez Gene ID
79776
Gene Name
zinc finger homeobox 4
Gene Aliases
ZFH4, ZHF4
UniGene
Hs.458973
Gene Chromosome Location
Chr.8: 76681219-76867285 on Build GRCh38
Transcripts
NM_024721.4 XM_011517592.2 XM_011517593.2 XM_011517594.2 XM_011517595.2 XM_011517596.2 XM_011517597.2 XM_017013845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7403240 c.644C>T p.S215L Chr.8: 76704732-76704732 on Build GRCh38
7249501 c.683G>T p.R228L Chr.8: 76704771-76704771 on Build GRCh38
4902244 c.1102C>T p.H368Y Chr.8: 76705190-76705190 on Build GRCh38
1458250 c.1128G>A p.P376P Chr.8: 76705216-76705216 on Build GRCh38
5677964 c.904C>G p.R302G Chr.8: 76704992-76704992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766666781 Chr.8: 76705125-76705125 on Build GRCh38
rs768463983 Chr.8: 76704680-76704680 on Build GRCh38
rs774436983 Chr.8: 76704807-76704807 on Build GRCh38
rs753750852 Chr.8: 76705163-76705163 on Build GRCh38
rs776733747 Chr.8: 76705111-76705111 on Build GRCh38
rs771303721 Chr.8: 76705195-76705195 on Build GRCh38
rs780418234 Chr.8: 76705079-76705079 on Build GRCh38
rs917060953 Chr.8: 76705173-76705173 on Build GRCh38
rs1006955039 Chr.8: 76705100-76705100 on Build GRCh38
rs768552335 Chr.8: 76704785-76704785 on Build GRCh38
rs745459061 Chr.8: 76704744-76704744 on Build GRCh38
rs754222202 Chr.8: 76704716-76704716 on Build GRCh38
rs753265770 Chr.8: 76704709-76704709 on Build GRCh38
rs554249315 Chr.8: 76704846-76704846 on Build GRCh38
rs559571105 Chr.8: 76704984-76704984 on Build GRCh38
rs545258517 Chr.8: 76705157-76705157 on Build GRCh38
rs866261853 Chr.8: 76704986-76704986 on Build GRCh38
rs751723912 Chr.8: 76704697-76704697 on Build GRCh38
rs747161654 Chr.8: 76705097-76705097 on Build GRCh38
rs370619364 Chr.8: 76705198-76705198 on Build GRCh38
rs778416005 Chr.8: 76704742-76704742 on Build GRCh38
rs575948315 Chr.8: 76704687-76704687 on Build GRCh38
rs753830045 Chr.8: 76705063-76705063 on Build GRCh38
rs934470343 Chr.8: 76704883-76704883 on Build GRCh38
rs772479291 Chr.8: 76705076-76705076 on Build GRCh38
rs372636732 Chr.8: 76704914-76704914 on Build GRCh38
rs767563060 Chr.8: 76704701-76704701 on Build GRCh38
rs776461133 Chr.8: 76704786-76704786 on Build GRCh38
rs267601996 Chr.8: 76705110-76705110 on Build GRCh38
rs979842713 Chr.8: 76705191-76705191 on Build GRCh38
rs76780980 Chr.8: 76704704-76704704 on Build GRCh38
rs746982156 Chr.8: 76704772-76704772 on Build GRCh38
rs779492595 Chr.8: 76704749-76704749 on Build GRCh38
rs772104280 Chr.8: 76704646-76704646 on Build GRCh38
rs1044466296 Chr.8: 76704933-76704933 on Build GRCh38
rs759742407 Chr.8: 76704811-76704811 on Build GRCh38
rs1031589143 Chr.8: 76705162-76705162 on Build GRCh38
rs758031067 Chr.8: 76704879-76704879 on Build GRCh38
rs762267675 Chr.8: 76705112-76705112 on Build GRCh38
rs746502584 Chr.8: 76704893-76704893 on Build GRCh38
rs968040047 Chr.8: 76705138-76705138 on Build GRCh38
rs201163377 Chr.8: 76705136-76705136 on Build GRCh38
rs191883247 Chr.8: 76704685-76704685 on Build GRCh38
rs373343766 Chr.8: 76705120-76705120 on Build GRCh38
rs763397631 Chr.8: 76705192-76705192 on Build GRCh38
rs370381615 Chr.8: 76705128-76705128 on Build GRCh38
rs375151254 Chr.8: 76704866-76704866 on Build GRCh38
rs978707598 Chr.8: 76705142-76705142 on Build GRCh38
rs369214100 Chr.8: 76704898-76704898 on Build GRCh38
rs748054601 Chr.8: 76705054-76705054 on Build GRCh38
rs903722943 Chr.8: 76704998-76704998 on Build GRCh38
rs759670574 Chr.8: 76705024-76705024 on Build GRCh38
rs757558402 Chr.8: 76704864-76704864 on Build GRCh38
rs969040934 Chr.8: 76705149-76705149 on Build GRCh38
rs773777266 Chr.8: 76705186-76705186 on Build GRCh38
rs376626064 Chr.8: 76705164-76705164 on Build GRCh38
rs774655361 Chr.8: 76705197-76705197 on Build GRCh38
rs748247880 Chr.8: 76705172-76705172 on Build GRCh38
rs767725738 Chr.8: 76704826-76704826 on Build GRCh38
rs747266189 Chr.8: 76705169-76705169 on Build GRCh38
rs199800319 Chr.8: 76705127-76705127 on Build GRCh38
rs770274276 Chr.8: 76705183-76705183 on Build GRCh38
rs760824197 Chr.8: 76704951-76704951 on Build GRCh38
rs1024589117 Chr.8: 76704649-76704649 on Build GRCh38
rs773007933 Chr.8: 76704908-76704908 on Build GRCh38
rs924445995 Chr.8: 76704770-76704770 on Build GRCh38
rs1037238862 Chr.8: 76705005-76705005 on Build GRCh38
rs771496482 Chr.8: 76704746-76704746 on Build GRCh38
rs751140518 Chr.8: 76705025-76705025 on Build GRCh38
rs775412271 Chr.8: 76704651-76704651 on Build GRCh38
rs954096599 Chr.8: 76704691-76704691 on Build GRCh38
rs774274741 Chr.8: 76705102-76705102 on Build GRCh38
rs768970618 Chr.8: 76705170-76705170 on Build GRCh38
rs779580830 Chr.8: 76704889-76704889 on Build GRCh38
rs998291994 Chr.8: 76705060-76705060 on Build GRCh38
rs866957030 Chr.8: 76704900-76704900 on Build GRCh38
rs760549285 Chr.8: 76704662-76704662 on Build GRCh38
rs781179725 Chr.8: 76705171-76705171 on Build GRCh38
rs1015717748 Chr.8: 76704658-76704658 on Build GRCh38
rs751230347 Chr.8: 76705126-76705126 on Build GRCh38
rs766005588 Chr.8: 76705009-76705009 on Build GRCh38
rs1050302678 Chr.8: 76704943-76704943 on Build GRCh38
rs941624915 Chr.8: 76704978-76704978 on Build GRCh38
rs961492645 Chr.8: 76704659-76704659 on Build GRCh38
rs754970156 Chr.8: 76704897-76704897 on Build GRCh38
rs778310765 Chr.8: 76704710-76704710 on Build GRCh38
rs776908219 Chr.8: 76704790-76704790 on Build GRCh38
rs555478805 Chr.8: 76704836-76704836 on Build GRCh38
rs761303149 Chr.8: 76704830-76704830 on Build GRCh38
rs573775710 Chr.8: 76704840-76704840 on Build GRCh38
rs746105394 Chr.8: 76704913-76704913 on Build GRCh38
rs756035851 Chr.8: 76705057-76705057 on Build GRCh38
rs998259416 Chr.8: 76704666-76704666 on Build GRCh38
rs370676988 Chr.8: 76705137-76705137 on Build GRCh38
rs992614485 Chr.8: 76704884-76704884 on Build GRCh38
rs771255702 Chr.8: 76704804-76704804 on Build GRCh38
rs749086451 Chr.8: 76705074-76705074 on Build GRCh38
rs888095047 Chr.8: 76704934-76704934 on Build GRCh38
rs752217978 Chr.8: 76705156-76705156 on Build GRCh38
rs777703837 Chr.8: 76704799-76704799 on Build GRCh38
rs748055530 Chr.8: 76704899-76704899 on Build GRCh38
rs759534883 Chr.8: 76704700-76704700 on Build GRCh38
rs374762945 Chr.8: 76705154-76705154 on Build GRCh38
rs763305879 Chr.8: 76705124-76705124 on Build GRCh38
rs375654092 Chr.8: 76704915-76704915 on Build GRCh38
rs527520302 Chr.8: 76705180-76705180 on Build GRCh38
rs756983466 Chr.8: 76704741-76704741 on Build GRCh38
rs770313841 Chr.8: 76705118-76705118 on Build GRCh38
rs369925965 Chr.8: 76704954-76704954 on Build GRCh38
rs35627561 Chr.8: 76705056-76705057 on Build GRCh38
rs536758211 Chr.8: 76704828-76704828 on Build GRCh38
rs370419358 Chr.8: 76705045-76705045 on Build GRCh38
rs754492746 Chr.8: 76705039-76705039 on Build GRCh38
rs764765016 Chr.8: 76704953-76704953 on Build GRCh38
rs367546818 Chr.8: 76704988-76704988 on Build GRCh38
rs865979348 Chr.8: 76705190-76705190 on Build GRCh38

View Full Product Details