Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32786326-32786895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM215
Entrez Gene ID
401498
Gene Name
transmembrane protein 215
Gene Aliases
-
UniGene
Hs.522063
Gene Chromosome Location
Chr.9: 32783499-32789201 on Build GRCh38
Transcripts
NM_212558.2

SNP Detail

SNP ID Chromosome Location
rs964854509 Chr.9: 32786749-32786749 on Build GRCh38
rs551210222 Chr.9: 32786838-32786838 on Build GRCh38
rs111522808 Chr.9: 32786728-32786728 on Build GRCh38
rs913163393 Chr.9: 32786783-32786783 on Build GRCh38
rs1026569051 Chr.9: 32786691-32786691 on Build GRCh38
rs916967095 Chr.9: 32786800-32786800 on Build GRCh38
rs1017843460 Chr.9: 32786399-32786399 on Build GRCh38
rs145148180 Chr.9: 32786846-32786846 on Build GRCh38
rs1044052631 Chr.9: 32786572-32786572 on Build GRCh38
rs926971880 Chr.9: 32786872-32786872 on Build GRCh38
rs565356157 Chr.9: 32786720-32786720 on Build GRCh38
rs300639 Chr.9: 32786555-32786555 on Build GRCh38
rs1054814060 Chr.9: 32786630-32786630 on Build GRCh38
rs950980375 Chr.9: 32786479-32786479 on Build GRCh38
rs751290581 Chr.9: 32786832-32786832 on Build GRCh38
rs546409212 Chr.9: 32786752-32786752 on Build GRCh38
rs1013881618 Chr.9: 32786667-32786667 on Build GRCh38
rs148395569 Chr.9: 32786764-32786764 on Build GRCh38
rs954676997 Chr.9: 32786385-32786385 on Build GRCh38
rs538568361 Chr.9: 32786357-32786357 on Build GRCh38
rs543189438 Chr.9: 32786524-32786524 on Build GRCh38
rs923169904 Chr.9: 32786436-32786436 on Build GRCh38
rs1021081928 Chr.9: 32786381-32786381 on Build GRCh38
rs947062925 Chr.9: 32786563-32786563 on Build GRCh38
rs920769293 Chr.9: 32786420-32786420 on Build GRCh38
rs781195392 Chr.9: 32786830-32786830 on Build GRCh38
rs374805713 Chr.9: 32786450-32786450 on Build GRCh38
rs964879405 Chr.9: 32786387-32786387 on Build GRCh38
rs1042474362 Chr.9: 32786658-32786658 on Build GRCh38
rs983601439 Chr.9: 32786790-32786790 on Build GRCh38
rs891615856 Chr.9: 32786633-32786633 on Build GRCh38
rs1042753220 Chr.9: 32786793-32786793 on Build GRCh38
rs111724809 Chr.9: 32786857-32786857 on Build GRCh38
rs531165444 Chr.9: 32786784-32786784 on Build GRCh38
rs1055326649 Chr.9: 32786624-32786624 on Build GRCh38
rs988655328 Chr.9: 32786778-32786778 on Build GRCh38
rs577459778 Chr.9: 32786515-32786515 on Build GRCh38
rs542711879 Chr.9: 32786623-32786623 on Build GRCh38
rs300638 Chr.9: 32786588-32786588 on Build GRCh38
rs780505478 Chr.9: 32786373-32786373 on Build GRCh38
rs1009646466 Chr.9: 32786354-32786354 on Build GRCh38
rs180756640 Chr.9: 32786415-32786415 on Build GRCh38
rs300637 Chr.9: 32786655-32786655 on Build GRCh38
rs1017617936 Chr.9: 32786732-32786732 on Build GRCh38
rs950659373 Chr.9: 32786706-32786706 on Build GRCh38
rs765422990 Chr.9: 32786616-32786616 on Build GRCh38
rs923147727 Chr.9: 32786760-32786760 on Build GRCh38
rs758021130 Chr.9: 32786473-32786473 on Build GRCh38
rs995089703 Chr.9: 32786736-32786736 on Build GRCh38
rs974630156 Chr.9: 32786408-32786408 on Build GRCh38
rs890559461 Chr.9: 32786711-32786711 on Build GRCh38
rs372198004 Chr.9: 32786367-32786367 on Build GRCh38

View Full Product Details