Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4379576-4380149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF23
Entrez Gene ID
8074
Gene Name
fibroblast growth factor 23
Gene Aliases
ADHR, FGFN, HPDR2, HYPF, PHPTC
UniGene
Hs.287370
Gene Chromosome Location
Chr.12: 4368227-4379728 on Build GRCh38
Transcripts
NM_020638.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
128000 c.6G>A p.L2L Chr.12: 4379577-4379577 on Build GRCh38
7368203 c.6G>T p.L2F Chr.12: 4379577-4379577 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs917006987 Chr.12: 4379691-4379691 on Build GRCh38
rs759380521 Chr.12: 4379628-4379628 on Build GRCh38
rs1031607763 Chr.12: 4379997-4379997 on Build GRCh38
rs1006961323 Chr.12: 4379913-4379913 on Build GRCh38
rs766642577 Chr.12: 4380101-4380101 on Build GRCh38
rs897882392 Chr.12: 4380029-4380029 on Build GRCh38
rs777736455 Chr.12: 4379606-4379606 on Build GRCh38
rs573078156 Chr.12: 4379755-4379755 on Build GRCh38
rs187035769 Chr.12: 4379635-4379635 on Build GRCh38
rs775119039 Chr.12: 4379864-4379864 on Build GRCh38
rs998326485 Chr.12: 4379850-4379850 on Build GRCh38
rs184719536 Chr.12: 4379910-4379910 on Build GRCh38
rs937877100 Chr.12: 4379961-4379961 on Build GRCh38
rs762575831 Chr.12: 4379869-4379869 on Build GRCh38
rs535686020 Chr.12: 4379808-4379808 on Build GRCh38
rs954136723 Chr.12: 4379891-4379891 on Build GRCh38
rs992671573 Chr.12: 4380030-4380030 on Build GRCh38
rs1056880193 Chr.12: 4379658-4379658 on Build GRCh38
rs3812822 Chr.12: 4380069-4380069 on Build GRCh38
rs942024111 Chr.12: 4379997-4379997 on Build GRCh38
rs768100347 Chr.12: 4379960-4379960 on Build GRCh38
rs1017187228 Chr.12: 4379846-4379846 on Build GRCh38
rs769556270 Chr.12: 4379698-4379698 on Build GRCh38
rs994009819 Chr.12: 4380042-4380042 on Build GRCh38
rs978738335 Chr.12: 4379622-4379622 on Build GRCh38
rs1037316124 Chr.12: 4379828-4379828 on Build GRCh38
rs1037664999 Chr.12: 4380108-4380108 on Build GRCh38
rs760895385 Chr.12: 4379637-4379637 on Build GRCh38
rs13312754 Chr.12: 4380097-4380097 on Build GRCh38
rs543839665 Chr.12: 4379911-4379911 on Build GRCh38
rs966889679 Chr.12: 4379912-4379912 on Build GRCh38
rs749504028 Chr.12: 4379619-4379619 on Build GRCh38
rs750971449 Chr.12: 4380015-4380015 on Build GRCh38
rs570732681 Chr.12: 4379708-4379708 on Build GRCh38
rs540530099 Chr.12: 4380039-4380039 on Build GRCh38
rs1007713970 Chr.12: 4380087-4380087 on Build GRCh38
rs1055780740 Chr.12: 4379660-4379660 on Build GRCh38
rs771094044 Chr.12: 4379620-4379620 on Build GRCh38
rs748668033 Chr.12: 4379596-4379596 on Build GRCh38
rs192182279 Chr.12: 4379890-4379890 on Build GRCh38
rs1037788654 Chr.12: 4379725-4379725 on Build GRCh38
rs577878876 Chr.12: 4379992-4379992 on Build GRCh38
rs553161255 Chr.12: 4379728-4379728 on Build GRCh38
rs934539125 Chr.12: 4379657-4379657 on Build GRCh38
rs909527260 Chr.12: 4380099-4380099 on Build GRCh38
rs968098842 Chr.12: 4379985-4379985 on Build GRCh38
rs748956554 Chr.12: 4379990-4379990 on Build GRCh38
rs954937693 Chr.12: 4379752-4379752 on Build GRCh38
rs769953313 Chr.12: 4379605-4379605 on Build GRCh38
rs764389755 Chr.12: 4380067-4380067 on Build GRCh38
rs771865871 Chr.12: 4379634-4379634 on Build GRCh38
rs1030471564 Chr.12: 4379750-4379750 on Build GRCh38
rs917123412 Chr.12: 4380048-4380048 on Build GRCh38
rs1006669222 Chr.12: 4379720-4379720 on Build GRCh38
rs897286243 Chr.12: 4379842-4379842 on Build GRCh38
rs539759806 Chr.12: 4379727-4379727 on Build GRCh38
rs761457252 Chr.12: 4379603-4379603 on Build GRCh38
rs372434012 Chr.12: 4379614-4379614 on Build GRCh38
rs190210195 Chr.12: 4379955-4379955 on Build GRCh38

View Full Product Details