Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43967645-43968089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT3
Entrez Gene ID
56894
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases
LPAAT-GAMMA1, LPAAT3
UniGene
Hs.248785
Gene Chromosome Location
Chr.21: 43865160-43987594 on Build GRCh38
Transcripts
NM_001037553.1 NM_020132.4 XM_005261160.4 XM_006724029.3 XM_006724030.3 XM_006724031.3 XM_011529660.2 XM_011529661.2 XM_011529662.2 XM_011529663.2 XM_011529664.2 XM_011529665.2 XM_017028408.1 XM_017028409.1 XM_017028410.1 XM_017028411.1 XM_017028412.1 XM_017028413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1307856 c.214G>C p.E72Q Chr.21: 43967981-43967981 on Build GRCh38
725909 c.212C>T p.T71M Chr.21: 43967979-43967979 on Build GRCh38
6596379 c.231G>A p.T77T Chr.21: 43967998-43967998 on Build GRCh38
4480307 c.240C>T p.A80A Chr.21: 43968007-43968007 on Build GRCh38
5614842 c.280C>A p.L94I Chr.21: 43968047-43968047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766655290 Chr.21: 43967720-43967720 on Build GRCh38
rs999423840 Chr.21: 43967801-43967801 on Build GRCh38
rs377075984 Chr.21: 43967926-43967926 on Build GRCh38
rs543357783 Chr.21: 43967767-43967767 on Build GRCh38
rs920395454 Chr.21: 43967713-43967713 on Build GRCh38
rs929139540 Chr.21: 43967719-43967719 on Build GRCh38
rs929024925 Chr.21: 43967700-43967700 on Build GRCh38
rs773959465 Chr.21: 43967927-43967927 on Build GRCh38
rs536360387 Chr.21: 43968037-43968037 on Build GRCh38
rs751767483 Chr.21: 43967822-43967822 on Build GRCh38
rs918718152 Chr.21: 43967860-43967860 on Build GRCh38
rs1022611220 Chr.21: 43967690-43967690 on Build GRCh38
rs115237511 Chr.21: 43967765-43967765 on Build GRCh38
rs973290114 Chr.21: 43967689-43967689 on Build GRCh38
rs186182916 Chr.21: 43967681-43967681 on Build GRCh38
rs748991482 Chr.21: 43967915-43967915 on Build GRCh38
rs896039402 Chr.21: 43967845-43967845 on Build GRCh38
rs759836461 Chr.21: 43967950-43967950 on Build GRCh38
rs528894771 Chr.21: 43967790-43967790 on Build GRCh38
rs758890874 Chr.21: 43967965-43967965 on Build GRCh38
rs749250492 Chr.21: 43967787-43967787 on Build GRCh38
rs553072371 Chr.21: 43967701-43967701 on Build GRCh38
rs984498012 Chr.21: 43967882-43967882 on Build GRCh38
rs747789869 Chr.21: 43968001-43968001 on Build GRCh38
rs747457775 Chr.21: 43967901-43967901 on Build GRCh38
rs115397760 Chr.21: 43968034-43968034 on Build GRCh38
rs755653262 Chr.21: 43967909-43967909 on Build GRCh38
rs1031869455 Chr.21: 43967836-43967836 on Build GRCh38
rs755344084 Chr.21: 43967850-43967850 on Build GRCh38
rs1035925253 Chr.21: 43967819-43967819 on Build GRCh38
rs1022557866 Chr.21: 43967905-43967905 on Build GRCh38
rs911785243 Chr.21: 43967738-43967738 on Build GRCh38
rs990161565 Chr.21: 43967664-43967664 on Build GRCh38
rs148475122 Chr.21: 43967766-43967766 on Build GRCh38
rs765234039 Chr.21: 43967670-43967670 on Build GRCh38
rs374261184 Chr.21: 43967912-43967912 on Build GRCh38
rs778591240 Chr.21: 43967998-43967998 on Build GRCh38
rs766666360 Chr.21: 43967937-43967937 on Build GRCh38
rs746992911 Chr.21: 43968011-43968011 on Build GRCh38
rs139706000 Chr.21: 43967719-43967720 on Build GRCh38
rs180749705 Chr.21: 43967671-43967671 on Build GRCh38
rs776526529 Chr.21: 43967941-43967941 on Build GRCh38
rs944661819 Chr.21: 43967743-43967743 on Build GRCh38
rs369212862 Chr.21: 43967959-43967959 on Build GRCh38
rs547485631 Chr.21: 43967872-43967872 on Build GRCh38
rs932638780 Chr.21: 43968060-43968060 on Build GRCh38
rs545580537 Chr.21: 43967751-43967751 on Build GRCh38
rs757654759 Chr.21: 43967988-43967988 on Build GRCh38
rs559466626 Chr.21: 43967883-43967883 on Build GRCh38
rs960355705 Chr.21: 43967679-43967679 on Build GRCh38
rs776602425 Chr.21: 43968018-43968018 on Build GRCh38
rs557448008 Chr.21: 43967742-43967742 on Build GRCh38
rs11910903 Chr.21: 43967675-43967675 on Build GRCh38
rs938848141 Chr.21: 43967684-43967684 on Build GRCh38
rs373523124 Chr.21: 43967980-43967980 on Build GRCh38
rs766497804 Chr.21: 43967940-43967940 on Build GRCh38
rs901697484 Chr.21: 43967732-43967732 on Build GRCh38
rs142672171 Chr.21: 43967890-43967890 on Build GRCh38
rs771837390 Chr.21: 43967932-43967932 on Build GRCh38
rs763209819 Chr.21: 43968067-43968067 on Build GRCh38
rs769981978 Chr.21: 43968035-43968035 on Build GRCh38
rs752119997 Chr.21: 43967986-43967986 on Build GRCh38
rs765597753 Chr.21: 43967954-43967954 on Build GRCh38
rs1031893463 Chr.21: 43967756-43967756 on Build GRCh38
rs960542576 Chr.21: 43967827-43967827 on Build GRCh38
rs1048717705 Chr.21: 43967702-43967702 on Build GRCh38
rs943032410 Chr.21: 43967884-43967884 on Build GRCh38
rs372562616 Chr.21: 43968061-43968061 on Build GRCh38
rs779644216 Chr.21: 43967990-43967990 on Build GRCh38
rs58345413 Chr.21: 43967784-43967784 on Build GRCh38
rs577743228 Chr.21: 43967710-43967710 on Build GRCh38
rs200808478 Chr.21: 43967979-43967979 on Build GRCh38
rs754459666 Chr.21: 43967997-43967997 on Build GRCh38
rs370497262 Chr.21: 43968017-43968017 on Build GRCh38
rs139644030 Chr.21: 43968009-43968009 on Build GRCh38
rs372444956 Chr.21: 43967930-43967930 on Build GRCh38
rs773204898 Chr.21: 43967935-43967935 on Build GRCh38
rs1021672652 Chr.21: 43967783-43967783 on Build GRCh38
rs748786236 Chr.21: 43967992-43967992 on Build GRCh38
rs973140772 Chr.21: 43967711-43967711 on Build GRCh38
rs867927001 Chr.21: 43967795-43967795 on Build GRCh38
rs200264871 Chr.21: 43968010-43968010 on Build GRCh38

View Full Product Details