Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71475572-71476161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF19
Entrez Gene ID
7567
Gene Name
zinc finger protein 19
Gene Aliases
KOX12
UniGene
Hs.660076
Gene Chromosome Location
Chr.16: 71474073-71489351 on Build GRCh38
Transcripts
NM_006961.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5385309 c.483C>T p.F161F Chr.16: 71476064-71476064 on Build GRCh38
3511834 c.414C>T p.H138H Chr.16: 71476133-71476133 on Build GRCh38
5435798 c.533G>T p.R178I Chr.16: 71476014-71476014 on Build GRCh38
3402455 c.774C>T p.S258S Chr.16: 71475773-71475773 on Build GRCh38
4967200 c.433A>G p.K145E Chr.16: 71476114-71476114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747832537 Chr.16: 71475701-71475701 on Build GRCh38
rs770646320 Chr.16: 71475788-71475788 on Build GRCh38
rs757485950 Chr.16: 71475652-71475652 on Build GRCh38
rs955927438 Chr.16: 71475636-71475636 on Build GRCh38
rs866652931 Chr.16: 71475948-71475948 on Build GRCh38
rs878952563 Chr.16: 71475677-71475677 on Build GRCh38
rs932274597 Chr.16: 71476131-71476131 on Build GRCh38
rs867416862 Chr.16: 71475669-71475669 on Build GRCh38
rs2288489 Chr.16: 71475782-71475782 on Build GRCh38
rs2288487 Chr.16: 71475685-71475685 on Build GRCh38
rs751178083 Chr.16: 71475858-71475858 on Build GRCh38
rs781184447 Chr.16: 71476081-71476081 on Build GRCh38
rs750092610 Chr.16: 71475949-71475949 on Build GRCh38
rs776806070 Chr.16: 71475902-71475902 on Build GRCh38
rs8050871 Chr.16: 71475893-71475893 on Build GRCh38
rs759876859 Chr.16: 71475849-71475849 on Build GRCh38
rs200860470 Chr.16: 71475772-71475772 on Build GRCh38
rs190278786 Chr.16: 71476059-71476059 on Build GRCh38
rs753159829 Chr.16: 71475921-71475921 on Build GRCh38
rs199884411 Chr.16: 71475931-71475931 on Build GRCh38
rs868731730 Chr.16: 71475987-71475987 on Build GRCh38
rs778331518 Chr.16: 71475700-71475700 on Build GRCh38
rs137874422 Chr.16: 71475600-71475600 on Build GRCh38
rs778482017 Chr.16: 71475827-71475827 on Build GRCh38
rs201608341 Chr.16: 71475909-71475909 on Build GRCh38
rs1009772638 Chr.16: 71475649-71475649 on Build GRCh38
rs767137185 Chr.16: 71476138-71476138 on Build GRCh38
rs754155369 Chr.16: 71475837-71475837 on Build GRCh38
rs751030484 Chr.16: 71475938-71475938 on Build GRCh38
rs147699542 Chr.16: 71475856-71475856 on Build GRCh38
rs958379164 Chr.16: 71475708-71475708 on Build GRCh38
rs749701548 Chr.16: 71476090-71476090 on Build GRCh38
rs200632530 Chr.16: 71475998-71475998 on Build GRCh38
rs776896425 Chr.16: 71475989-71475989 on Build GRCh38
rs750623723 Chr.16: 71475747-71475747 on Build GRCh38
rs745846950 Chr.16: 71475939-71475941 on Build GRCh38
rs140092802 Chr.16: 71475791-71475791 on Build GRCh38
rs199580690 Chr.16: 71476003-71476003 on Build GRCh38
rs764709458 Chr.16: 71476009-71476009 on Build GRCh38
rs749184710 Chr.16: 71475970-71475970 on Build GRCh38
rs756500579 Chr.16: 71475749-71475749 on Build GRCh38
rs778421522 Chr.16: 71475773-71475773 on Build GRCh38
rs779933459 Chr.16: 71475958-71475958 on Build GRCh38
rs992457642 Chr.16: 71475725-71475725 on Build GRCh38
rs773305576 Chr.16: 71475896-71475896 on Build GRCh38
rs768591637 Chr.16: 71475971-71475971 on Build GRCh38
rs774532473 Chr.16: 71475744-71475744 on Build GRCh38
rs375143736 Chr.16: 71475794-71475794 on Build GRCh38
rs2288488 Chr.16: 71475731-71475731 on Build GRCh38
rs147578256 Chr.16: 71476073-71476073 on Build GRCh38
rs759490984 Chr.16: 71475733-71475733 on Build GRCh38
rs746820113 Chr.16: 71475729-71475729 on Build GRCh38
rs763451561 Chr.16: 71475933-71475933 on Build GRCh38
rs760688769 Chr.16: 71475613-71475613 on Build GRCh38
rs762347896 Chr.16: 71476127-71476127 on Build GRCh38
rs575955916 Chr.16: 71475877-71475877 on Build GRCh38
rs759697187 Chr.16: 71475992-71475992 on Build GRCh38
rs772054712 Chr.16: 71475894-71475894 on Build GRCh38
rs761836042 Chr.16: 71475817-71475817 on Build GRCh38
rs371975799 Chr.16: 71475628-71475628 on Build GRCh38
rs772241696 Chr.16: 71475848-71475848 on Build GRCh38
rs771976566 Chr.16: 71476020-71476022 on Build GRCh38
rs369036617 Chr.16: 71475934-71475934 on Build GRCh38
rs542310581 Chr.16: 71476020-71476020 on Build GRCh38
rs768898558 Chr.16: 71476098-71476098 on Build GRCh38
rs780202978 Chr.16: 71475884-71475884 on Build GRCh38
rs763390532 Chr.16: 71476000-71476000 on Build GRCh38
rs985892718 Chr.16: 71476065-71476065 on Build GRCh38
rs746122425 Chr.16: 71476083-71476083 on Build GRCh38
rs780461558 Chr.16: 71475687-71475687 on Build GRCh38
rs562940678 Chr.16: 71475936-71475936 on Build GRCh38
rs769806047 Chr.16: 71475795-71475795 on Build GRCh38
rs770410989 Chr.16: 71475814-71475815 on Build GRCh38
rs369556832 Chr.16: 71475808-71475808 on Build GRCh38
rs774689136 Chr.16: 71476015-71476015 on Build GRCh38
rs141601110 Chr.16: 71475738-71475738 on Build GRCh38
rs867090026 Chr.16: 71475727-71475727 on Build GRCh38
rs377052556 Chr.16: 71475950-71475950 on Build GRCh38
rs754537907 Chr.16: 71475695-71475695 on Build GRCh38
rs370498796 Chr.16: 71475975-71475975 on Build GRCh38
rs1002351725 Chr.16: 71475915-71475915 on Build GRCh38
rs774976831 Chr.16: 71476111-71476111 on Build GRCh38
rs773361881 Chr.16: 71475832-71475832 on Build GRCh38
rs763195162 Chr.16: 71475638-71475638 on Build GRCh38
rs552689733 Chr.16: 71476087-71476087 on Build GRCh38
rs767955822 Chr.16: 71475678-71475678 on Build GRCh38
rs778696228 Chr.16: 71475974-71475974 on Build GRCh38
rs766749424 Chr.16: 71475759-71475759 on Build GRCh38
rs766112653 Chr.16: 71476064-71476064 on Build GRCh38
rs749787976 Chr.16: 71475688-71475688 on Build GRCh38
rs778980582 Chr.16: 71476070-71476070 on Build GRCh38
rs145400322 Chr.16: 71475836-71475836 on Build GRCh38
rs962014957 Chr.16: 71476026-71476026 on Build GRCh38
rs767118912 Chr.16: 71476023-71476023 on Build GRCh38
rs748916253 Chr.16: 71475722-71475723 on Build GRCh38
rs771580209 Chr.16: 71475715-71475715 on Build GRCh38
rs767626355 Chr.16: 71475828-71475828 on Build GRCh38
rs55802324 Chr.16: 71475597-71475597 on Build GRCh38
rs868420487 Chr.16: 71475952-71475952 on Build GRCh38
rs201660319 Chr.16: 71475712-71475712 on Build GRCh38
rs149461299 Chr.16: 71475679-71475679 on Build GRCh38
rs149111762 Chr.16: 71476047-71476047 on Build GRCh38
rs765694698 Chr.16: 71476011-71476011 on Build GRCh38
rs867926196 Chr.16: 71475960-71475960 on Build GRCh38
rs199702128 Chr.16: 71475901-71475901 on Build GRCh38
rs769676661 Chr.16: 71475883-71475883 on Build GRCh38
rs371577988 Chr.16: 71475997-71475997 on Build GRCh38
rs764258156 Chr.16: 71475644-71475644 on Build GRCh38
rs368490940 Chr.16: 71475996-71475996 on Build GRCh38
rs765777803 Chr.16: 71475850-71475850 on Build GRCh38
rs571914805 Chr.16: 71475834-71475834 on Build GRCh38
rs759960933 Chr.16: 71475905-71475905 on Build GRCh38
rs751843531 Chr.16: 71475645-71475645 on Build GRCh38
rs145725096 Chr.16: 71475939-71475939 on Build GRCh38
rs372256168 Chr.16: 71475763-71475763 on Build GRCh38
rs781031409 Chr.16: 71475942-71475942 on Build GRCh38
rs10500557 Chr.16: 71475876-71475876 on Build GRCh38
rs766276470 Chr.16: 71475614-71475614 on Build GRCh38
rs1034073175 Chr.16: 71475705-71475705 on Build GRCh38
rs772902248 Chr.16: 71475718-71475718 on Build GRCh38
rs775552227 Chr.16: 71476089-71476089 on Build GRCh38
rs746661649 Chr.16: 71475785-71475785 on Build GRCh38

View Full Product Details