Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2087275-2087850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMG6
Entrez Gene ID
23293
Gene Name
SMG6, nonsense mediated mRNA decay factor
Gene Aliases
C17orf31, EST1A, SMG-6, hSMG5/7a
UniGene
Hs.448342
Gene Chromosome Location
Chr.17: 2059839-2303836 on Build GRCh38
Transcripts
NM_001256827.1 NM_001256828.1 NM_001282326.1 NM_017575.4 XM_005256569.3 XM_005256571.4 XM_011523769.2 XM_011523775.2 XM_017024399.1

SNP Detail

SNP ID Chromosome Location
rs897450171 Chr.17: 2087777-2087777 on Build GRCh38
rs1044966215 Chr.17: 2087508-2087508 on Build GRCh38
rs752142479 Chr.17: 2087722-2087722 on Build GRCh38
rs765395120 Chr.17: 2087584-2087584 on Build GRCh38
rs1007403992 Chr.17: 2087814-2087814 on Build GRCh38
rs976388253 Chr.17: 2087641-2087641 on Build GRCh38
rs548752339 Chr.17: 2087621-2087621 on Build GRCh38
rs779804553 Chr.17: 2087501-2087502 on Build GRCh38
rs552746219 Chr.17: 2087719-2087719 on Build GRCh38
rs967602263 Chr.17: 2087410-2087410 on Build GRCh38
rs958660851 Chr.17: 2087731-2087731 on Build GRCh38
rs1035452357 Chr.17: 2087704-2087704 on Build GRCh38
rs3744274 Chr.17: 2087665-2087665 on Build GRCh38
rs908098474 Chr.17: 2087789-2087789 on Build GRCh38
rs560872391 Chr.17: 2087540-2087540 on Build GRCh38
rs750086953 Chr.17: 2087373-2087373 on Build GRCh38
rs916837175 Chr.17: 2087506-2087506 on Build GRCh38
rs938890754 Chr.17: 2087754-2087754 on Build GRCh38
rs184062793 Chr.17: 2087598-2087598 on Build GRCh38
rs1052791128 Chr.17: 2087447-2087447 on Build GRCh38
rs981652092 Chr.17: 2087697-2087697 on Build GRCh38
rs569508031 Chr.17: 2087363-2087363 on Build GRCh38
rs548210295 Chr.17: 2087376-2087376 on Build GRCh38
rs972792597 Chr.17: 2087356-2087356 on Build GRCh38
rs991118865 Chr.17: 2087495-2087495 on Build GRCh38
rs1011475452 Chr.17: 2087666-2087666 on Build GRCh38
rs940884745 Chr.17: 2087790-2087790 on Build GRCh38
rs1017868566 Chr.17: 2087601-2087601 on Build GRCh38
rs112206049 Chr.17: 2087796-2087796 on Build GRCh38
rs763665136 Chr.17: 2087496-2087496 on Build GRCh38
rs544388420 Chr.17: 2087533-2087533 on Build GRCh38
rs914286854 Chr.17: 2087682-2087682 on Build GRCh38
rs34048072 Chr.17: 2087619-2087620 on Build GRCh38
rs181066361 Chr.17: 2087514-2087514 on Build GRCh38
rs572480684 Chr.17: 2087471-2087471 on Build GRCh38
rs530011076 Chr.17: 2087551-2087551 on Build GRCh38
rs1005003723 Chr.17: 2087348-2087348 on Build GRCh38
rs188183150 Chr.17: 2087468-2087468 on Build GRCh38
rs117232118 Chr.17: 2087677-2087677 on Build GRCh38
rs149062441 Chr.17: 2087494-2087494 on Build GRCh38
rs1003552961 Chr.17: 2087526-2087526 on Build GRCh38
rs183455574 Chr.17: 2087442-2087442 on Build GRCh38
rs761503611 Chr.17: 2087780-2087780 on Build GRCh38
rs188923603 Chr.17: 2087319-2087319 on Build GRCh38
rs1057386388 Chr.17: 2087755-2087755 on Build GRCh38
rs957253768 Chr.17: 2087372-2087372 on Build GRCh38
rs368911335 Chr.17: 2087416-2087416 on Build GRCh38
rs1028987611 Chr.17: 2087634-2087634 on Build GRCh38
rs531662263 Chr.17: 2087751-2087751 on Build GRCh38
rs994203616 Chr.17: 2087554-2087554 on Build GRCh38
rs1049334522 Chr.17: 2087517-2087517 on Build GRCh38
rs533919258 Chr.17: 2087528-2087528 on Build GRCh38
rs952776279 Chr.17: 2087586-2087586 on Build GRCh38
rs552682867 Chr.17: 2087355-2087355 on Build GRCh38
rs192702085 Chr.17: 2087424-2087424 on Build GRCh38
rs996143415 Chr.17: 2087611-2087611 on Build GRCh38
rs569370983 Chr.17: 2087738-2087738 on Build GRCh38
rs755393930 Chr.17: 2087750-2087750 on Build GRCh38
rs964637560 Chr.17: 2087633-2087633 on Build GRCh38
rs2232482 Chr.17: 2087803-2087803 on Build GRCh38
rs117908996 Chr.17: 2087384-2087384 on Build GRCh38
rs147554048 Chr.17: 2087326-2087326 on Build GRCh38
rs934351991 Chr.17: 2087425-2087425 on Build GRCh38
rs544729182 Chr.17: 2087483-2087483 on Build GRCh38
rs548775333 Chr.17: 2087766-2087766 on Build GRCh38
rs934321358 Chr.17: 2087674-2087674 on Build GRCh38
rs759966393 Chr.17: 2087349-2087349 on Build GRCh38
rs538425222 Chr.17: 2087366-2087366 on Build GRCh38
rs910432530 Chr.17: 2087374-2087374 on Build GRCh38
rs930953335 Chr.17: 2087779-2087779 on Build GRCh38
rs1036114300 Chr.17: 2087328-2087328 on Build GRCh38
rs1049220692 Chr.17: 2087325-2087325 on Build GRCh38

View Full Product Details