Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51758054-51758378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS12
Entrez Gene ID
79758
Gene Name
dehydrogenase/reductase 12
Gene Aliases
SDR40C1
UniGene
Hs.266728
Gene Chromosome Location
Chr.13: 51754856-51804206 on Build GRCh38
Transcripts
XM_017020750.1 XM_017020751.1 NM_052950.3 XM_011534914.1 XM_011534915.2 XM_017020386.1

Gene Symbol
WDFY2
Entrez Gene ID
115825
Gene Name
WD repeat and FYVE domain containing 2
Gene Aliases
PROF, WDF2, ZFYVE22
UniGene
Hs.208550
Gene Chromosome Location
Chr.13: 51584194-51766799 on Build GRCh38
Transcripts
XM_017020750.1 XM_017020751.1 NM_052950.3 XM_011534914.1 XM_011534915.2 XM_017020386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947976 c.1122C>T p.F374F Chr.13: 51758249-51758249 on Build GRCh38
6500864 c.1076C>T p.T359I Chr.13: 51758203-51758203 on Build GRCh38
3384869 c.1088A>G p.H363R Chr.13: 51758215-51758215 on Build GRCh38
432485 c.1066C>T p.R356C Chr.13: 51758193-51758193 on Build GRCh38
4622845 c.1162A>G p.K388E Chr.13: 51758289-51758289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368920902 Chr.13: 51758194-51758194 on Build GRCh38
rs756604674 Chr.13: 51758179-51758179 on Build GRCh38
rs775222071 Chr.13: 51758173-51758176 on Build GRCh38
rs373154740 Chr.13: 51758305-51758305 on Build GRCh38
rs376165492 Chr.13: 51758324-51758324 on Build GRCh38
rs767757269 Chr.13: 51758349-51758349 on Build GRCh38
rs746370087 Chr.13: 51758171-51758171 on Build GRCh38
rs747778835 Chr.13: 51758196-51758196 on Build GRCh38
rs886176962 Chr.13: 51758133-51758134 on Build GRCh38
rs772735112 Chr.13: 51758199-51758199 on Build GRCh38
rs768372007 Chr.13: 51758133-51758136 on Build GRCh38
rs543299262 Chr.13: 51758225-51758225 on Build GRCh38
rs755406495 Chr.13: 51758251-51758251 on Build GRCh38
rs757273060 Chr.13: 51758167-51758167 on Build GRCh38
rs980435716 Chr.13: 51758087-51758087 on Build GRCh38
rs760177170 Chr.13: 51758343-51758344 on Build GRCh38
rs746494123 Chr.13: 51758289-51758289 on Build GRCh38
rs767526434 Chr.13: 51758239-51758239 on Build GRCh38
rs766279668 Chr.13: 51758203-51758203 on Build GRCh38
rs377337483 Chr.13: 51758352-51758352 on Build GRCh38
rs751731978 Chr.13: 51758163-51758163 on Build GRCh38
rs776446892 Chr.13: 51758204-51758204 on Build GRCh38
rs781280039 Chr.13: 51758170-51758170 on Build GRCh38
rs759325063 Chr.13: 51758213-51758213 on Build GRCh38
rs909276084 Chr.13: 51758316-51758316 on Build GRCh38
rs780677694 Chr.13: 51758249-51758249 on Build GRCh38
rs369342196 Chr.13: 51758219-51758219 on Build GRCh38
rs779270096 Chr.13: 51758252-51758252 on Build GRCh38
rs747786344 Chr.13: 51758270-51758270 on Build GRCh38
rs890733974 Chr.13: 51758254-51758254 on Build GRCh38
rs35409673 Chr.13: 51758193-51758193 on Build GRCh38
rs940453833 Chr.13: 51758137-51758137 on Build GRCh38
rs776540045 Chr.13: 51758298-51758298 on Build GRCh38
rs764360064 Chr.13: 51758216-51758216 on Build GRCh38
rs750467137 Chr.13: 51758241-51758241 on Build GRCh38
rs761964873 Chr.13: 51758337-51758337 on Build GRCh38
rs554316811 Chr.13: 51758188-51758188 on Build GRCh38
rs141650653 Chr.13: 51758182-51758182 on Build GRCh38
rs1050771719 Chr.13: 51758148-51758148 on Build GRCh38
rs749727071 Chr.13: 51758250-51758250 on Build GRCh38
rs373067803 Chr.13: 51758200-51758200 on Build GRCh38
rs771703477 Chr.13: 51758273-51758273 on Build GRCh38
rs148391704 Chr.13: 51758095-51758095 on Build GRCh38
rs772642435 Chr.13: 51758288-51758288 on Build GRCh38
rs969329194 Chr.13: 51758276-51758276 on Build GRCh38
rs879402819 Chr.13: 51758120-51758120 on Build GRCh38
rs764072257 Chr.13: 51758315-51758315 on Build GRCh38
rs771486346 Chr.13: 51758198-51758198 on Build GRCh38
rs766963244 Chr.13: 51758145-51758146 on Build GRCh38
rs770919533 Chr.13: 51758290-51758290 on Build GRCh38
rs765025291 Chr.13: 51758311-51758311 on Build GRCh38
rs571768187 Chr.13: 51758102-51758102 on Build GRCh38
rs916615818 Chr.13: 51758259-51758259 on Build GRCh38
rs927248789 Chr.13: 51758098-51758098 on Build GRCh38
rs1056749305 Chr.13: 51758119-51758119 on Build GRCh38

View Full Product Details