Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97994514-97994828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANP32B
Entrez Gene ID
10541
Gene Name
acidic nuclear phosphoprotein 32 family member B
Gene Aliases
APRIL, PHAPI2, SSP29
UniGene
Hs.730654
Gene Chromosome Location
Chr.9: 97983207-98015943 on Build GRCh38
Transcripts
NM_006401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6312125 c.107G>T p.G36V Chr.9: 97994683-97994683 on Build GRCh38
1624345 c.69C>A p.V23V Chr.9: 97994645-97994645 on Build GRCh38
3432694 c.58C>T p.R20* Chr.9: 97994634-97994634 on Build GRCh38
752335 c.86C>G p.S29* Chr.9: 97994662-97994662 on Build GRCh38
1464343 c.179T>G p.L60R Chr.9: 97994755-97994755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562853890 Chr.9: 97994607-97994608 on Build GRCh38
rs779462378 Chr.9: 97994798-97994798 on Build GRCh38
rs776354657 Chr.9: 97994635-97994635 on Build GRCh38
rs745863418 Chr.9: 97994621-97994621 on Build GRCh38
rs767568260 Chr.9: 97994716-97994716 on Build GRCh38
rs765124629 Chr.9: 97994678-97994678 on Build GRCh38
rs771679420 Chr.9: 97994598-97994598 on Build GRCh38
rs756222170 Chr.9: 97994730-97994730 on Build GRCh38
rs1044321383 Chr.9: 97994741-97994741 on Build GRCh38
rs397803685 Chr.9: 97994615-97994616 on Build GRCh38
rs986087086 Chr.9: 97994547-97994547 on Build GRCh38
rs754112809 Chr.9: 97994771-97994771 on Build GRCh38
rs973099581 Chr.9: 97994718-97994718 on Build GRCh38
rs930015470 Chr.9: 97994654-97994654 on Build GRCh38
rs188640709 Chr.9: 97994693-97994693 on Build GRCh38
rs764181693 Chr.9: 97994767-97994767 on Build GRCh38
rs768138478 Chr.9: 97994628-97994628 on Build GRCh38
rs921804556 Chr.9: 97994765-97994765 on Build GRCh38
rs911503126 Chr.9: 97994567-97994567 on Build GRCh38
rs142118802 Chr.9: 97994675-97994675 on Build GRCh38
rs746427879 Chr.9: 97994591-97994591 on Build GRCh38
rs771358121 Chr.9: 97994581-97994581 on Build GRCh38
rs376602943 Chr.9: 97994677-97994677 on Build GRCh38
rs777286270 Chr.9: 97994597-97994598 on Build GRCh38
rs372434240 Chr.9: 97994578-97994578 on Build GRCh38
rs375200191 Chr.9: 97994803-97994803 on Build GRCh38
rs931935482 Chr.9: 97994791-97994791 on Build GRCh38
rs748208608 Chr.9: 97994689-97994689 on Build GRCh38
rs759034393 Chr.9: 97994804-97994804 on Build GRCh38
rs779513805 Chr.9: 97994588-97994588 on Build GRCh38
rs146174352 Chr.9: 97994702-97994702 on Build GRCh38
rs3215934 Chr.9: 97994609-97994610 on Build GRCh38
rs372216408 Chr.9: 97994797-97994797 on Build GRCh38
rs962830490 Chr.9: 97994575-97994575 on Build GRCh38
rs760365777 Chr.9: 97994625-97994625 on Build GRCh38
rs1000022416 Chr.9: 97994801-97994801 on Build GRCh38
rs752713432 Chr.9: 97994720-97994720 on Build GRCh38
rs974249120 Chr.9: 97994609-97994609 on Build GRCh38

View Full Product Details