Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 103097119-103097691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIN1
Entrez Gene ID
54826
Gene Name
gypsy retrotransposon integrase 1
Gene Aliases
GIN-1, TGIN1, ZH2C2
UniGene
Hs.24088
Gene Chromosome Location
Chr.5: 103086000-103120149 on Build GRCh38
Transcripts
NM_001317954.1 NM_017676.2 NR_134299.1 XM_005272028.3 XM_017009599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1059281 c.806C>T p.S269L Chr.5: 103097615-103097615 on Build GRCh38
5441741 c.949C>G p.L317V Chr.5: 103097373-103097373 on Build GRCh38
1059280 c.907C>A p.H303N Chr.5: 103097415-103097415 on Build GRCh38
4585319 c.896C>A p.S299* Chr.5: 103097426-103097426 on Build GRCh38
6546383 c.981G>A p.M327I Chr.5: 103097341-103097341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145309875 Chr.5: 103097511-103097511 on Build GRCh38
rs781894981 Chr.5: 103097585-103097585 on Build GRCh38
rs572875187 Chr.5: 103097172-103097172 on Build GRCh38
rs918375957 Chr.5: 103097268-103097268 on Build GRCh38
rs34806575 Chr.5: 103097287-103097288 on Build GRCh38
rs570010718 Chr.5: 103097662-103097662 on Build GRCh38
rs782192581 Chr.5: 103097524-103097525 on Build GRCh38
rs182983870 Chr.5: 103097264-103097264 on Build GRCh38
rs186181946 Chr.5: 103097212-103097212 on Build GRCh38
rs782269685 Chr.5: 103097341-103097341 on Build GRCh38
rs781966228 Chr.5: 103097642-103097642 on Build GRCh38
rs962409014 Chr.5: 103097249-103097249 on Build GRCh38
rs781896105 Chr.5: 103097431-103097431 on Build GRCh38
rs782785900 Chr.5: 103097663-103097663 on Build GRCh38
rs782261502 Chr.5: 103097472-103097472 on Build GRCh38
rs79506806 Chr.5: 103097394-103097394 on Build GRCh38
rs782417984 Chr.5: 103097626-103097626 on Build GRCh38
rs782589545 Chr.5: 103097305-103097306 on Build GRCh38
rs782695201 Chr.5: 103097430-103097430 on Build GRCh38
rs781926180 Chr.5: 103097392-103097392 on Build GRCh38
rs782813041 Chr.5: 103097583-103097583 on Build GRCh38
rs545089031 Chr.5: 103097199-103097199 on Build GRCh38
rs1000366570 Chr.5: 103097180-103097180 on Build GRCh38
rs782061834 Chr.5: 103097422-103097422 on Build GRCh38
rs782393563 Chr.5: 103097352-103097352 on Build GRCh38
rs551096117 Chr.5: 103097440-103097440 on Build GRCh38
rs199639404 Chr.5: 103097510-103097510 on Build GRCh38
rs202047535 Chr.5: 103097600-103097600 on Build GRCh38
rs782759061 Chr.5: 103097534-103097534 on Build GRCh38
rs781805394 Chr.5: 103097318-103097318 on Build GRCh38
rs538975183 Chr.5: 103097632-103097632 on Build GRCh38
rs536788313 Chr.5: 103097484-103097484 on Build GRCh38
rs958691648 Chr.5: 103097201-103097201 on Build GRCh38
rs782259917 Chr.5: 103097635-103097635 on Build GRCh38
rs782372737 Chr.5: 103097641-103097641 on Build GRCh38
rs782592648 Chr.5: 103097310-103097310 on Build GRCh38
rs191419143 Chr.5: 103097488-103097488 on Build GRCh38
rs1014221886 Chr.5: 103097636-103097636 on Build GRCh38
rs376339815 Chr.5: 103097592-103097592 on Build GRCh38
rs2270563 Chr.5: 103097300-103097300 on Build GRCh38
rs782688806 Chr.5: 103097323-103097323 on Build GRCh38
rs782186067 Chr.5: 103097459-103097459 on Build GRCh38
rs376398924 Chr.5: 103097272-103097272 on Build GRCh38
rs782766428 Chr.5: 103097442-103097442 on Build GRCh38
rs781999131 Chr.5: 103097512-103097512 on Build GRCh38
rs561503755 Chr.5: 103097265-103097265 on Build GRCh38
rs571318747 Chr.5: 103097461-103097461 on Build GRCh38
rs782437628 Chr.5: 103097575-103097575 on Build GRCh38
rs530910513 Chr.5: 103097242-103097242 on Build GRCh38
rs782034963 Chr.5: 103097661-103097661 on Build GRCh38
rs782119647 Chr.5: 103097404-103097404 on Build GRCh38
rs375806379 Chr.5: 103097475-103097475 on Build GRCh38
rs185820409 Chr.5: 103097378-103097378 on Build GRCh38
rs782066633 Chr.5: 103097266-103097266 on Build GRCh38
rs782111359 Chr.5: 103097514-103097514 on Build GRCh38
rs138420837 Chr.5: 103097645-103097645 on Build GRCh38
rs375232582 Chr.5: 103097587-103097587 on Build GRCh38
rs782478457 Chr.5: 103097451-103097451 on Build GRCh38
rs782778300 Chr.5: 103097288-103097288 on Build GRCh38
rs372385163 Chr.5: 103097470-103097470 on Build GRCh38
rs190509737 Chr.5: 103097220-103097220 on Build GRCh38
rs782426801 Chr.5: 103097319-103097319 on Build GRCh38
rs782634722 Chr.5: 103097376-103097376 on Build GRCh38
rs782070321 Chr.5: 103097213-103097214 on Build GRCh38
rs75433174 Chr.5: 103097171-103097171 on Build GRCh38
rs782752511 Chr.5: 103097408-103097408 on Build GRCh38
rs367914452 Chr.5: 103097603-103097603 on Build GRCh38
rs782204346 Chr.5: 103097502-103097502 on Build GRCh38
rs528050660 Chr.5: 103097418-103097418 on Build GRCh38
rs931284268 Chr.5: 103097324-103097324 on Build GRCh38
rs202036425 Chr.5: 103097391-103097391 on Build GRCh38
rs200357523 Chr.5: 103097399-103097399 on Build GRCh38
rs201680199 Chr.5: 103097366-103097366 on Build GRCh38
rs1034551423 Chr.5: 103097539-103097539 on Build GRCh38
rs370710680 Chr.5: 103097537-103097537 on Build GRCh38
rs782470928 Chr.5: 103097616-103097616 on Build GRCh38
rs781854688 Chr.5: 103097450-103097450 on Build GRCh38
rs782588762 Chr.5: 103097453-103097453 on Build GRCh38
rs781906407 Chr.5: 103097276-103097276 on Build GRCh38
rs782690279 Chr.5: 103097652-103097652 on Build GRCh38
rs782232747 Chr.5: 103097601-103097601 on Build GRCh38
rs782587694 Chr.5: 103097623-103097623 on Build GRCh38
rs374705868 Chr.5: 103097509-103097509 on Build GRCh38
rs984029078 Chr.5: 103097344-103097344 on Build GRCh38
rs782482153 Chr.5: 103097308-103097308 on Build GRCh38
rs200543823 Chr.5: 103097286-103097286 on Build GRCh38
rs782006808 Chr.5: 103097401-103097401 on Build GRCh38
rs542549105 Chr.5: 103097170-103097170 on Build GRCh38

View Full Product Details