Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109493880-109494444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK9
Entrez Gene ID
221264
Gene Name
adenylate kinase 9
Gene Aliases
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1
UniGene
Hs.205144
Gene Chromosome Location
Chr.6: 109492855-109691253 on Build GRCh38
Transcripts
NM_001145128.2 XM_006715376.3 XM_011535550.2 XM_011535552.2 XM_011535553.2 XM_011535554.2 XM_011535558.1 XM_017010385.1 XM_017010387.1 XM_017010389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3155365 c.678C>T p.P226P Chr.6: 109494033-109494033 on Build GRCh38
4499897 c.5529C>T p.L1843L Chr.6: 109493985-109493985 on Build GRCh38
6482065 c.640G>C p.A214P Chr.6: 109494071-109494071 on Build GRCh38
3857395 c.653C>T p.A218V Chr.6: 109494058-109494058 on Build GRCh38
6576013 c.730+5G>C p.? Chr.6: 109493976-109493976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886546276 Chr.6: 109494361-109494361 on Build GRCh38
rs56375804 Chr.6: 109494274-109494274 on Build GRCh38
rs768982477 Chr.6: 109494079-109494079 on Build GRCh38
rs369525950 Chr.6: 109494130-109494130 on Build GRCh38
rs757492331 Chr.6: 109494023-109494023 on Build GRCh38
rs767353050 Chr.6: 109494137-109494137 on Build GRCh38
rs761221097 Chr.6: 109494096-109494096 on Build GRCh38
rs942648993 Chr.6: 109494273-109494273 on Build GRCh38
rs749508310 Chr.6: 109493917-109493917 on Build GRCh38
rs551159902 Chr.6: 109494168-109494168 on Build GRCh38
rs142277030 Chr.6: 109493920-109493920 on Build GRCh38
rs955254601 Chr.6: 109494232-109494232 on Build GRCh38
rs920603376 Chr.6: 109494292-109494292 on Build GRCh38
rs761537156 Chr.6: 109493932-109493932 on Build GRCh38
rs1003619215 Chr.6: 109494380-109494380 on Build GRCh38
rs1039705884 Chr.6: 109494034-109494034 on Build GRCh38
rs930616895 Chr.6: 109494322-109494322 on Build GRCh38
rs895313866 Chr.6: 109494391-109494391 on Build GRCh38
rs750297547 Chr.6: 109493943-109493943 on Build GRCh38
rs1047770433 Chr.6: 109493926-109493926 on Build GRCh38
rs754949252 Chr.6: 109494131-109494131 on Build GRCh38
rs772585505 Chr.6: 109494086-109494086 on Build GRCh38
rs758174718 Chr.6: 109493953-109493953 on Build GRCh38
rs116326624 Chr.6: 109494359-109494359 on Build GRCh38
rs771253648 Chr.6: 109494051-109494051 on Build GRCh38
rs956085027 Chr.6: 109494275-109494275 on Build GRCh38
rs764995702 Chr.6: 109494097-109494097 on Build GRCh38
rs776042227 Chr.6: 109494101-109494101 on Build GRCh38
rs1023999268 Chr.6: 109494194-109494194 on Build GRCh38
rs543075643 Chr.6: 109494112-109494112 on Build GRCh38
rs1051506431 Chr.6: 109494103-109494103 on Build GRCh38
rs988876011 Chr.6: 109494314-109494314 on Build GRCh38
rs192902118 Chr.6: 109494057-109494057 on Build GRCh38
rs541453415 Chr.6: 109494416-109494416 on Build GRCh38
rs923657997 Chr.6: 109494222-109494222 on Build GRCh38
rs1057070008 Chr.6: 109494387-109494387 on Build GRCh38
rs575654381 Chr.6: 109494400-109494400 on Build GRCh38
rs563681269 Chr.6: 109493978-109493978 on Build GRCh38
rs886622374 Chr.6: 109494055-109494055 on Build GRCh38
rs764726864 Chr.6: 109493939-109493939 on Build GRCh38
rs143217016 Chr.6: 109494393-109494393 on Build GRCh38
rs7755941 Chr.6: 109494148-109494148 on Build GRCh38
rs7773950 Chr.6: 109494237-109494237 on Build GRCh38
rs571151568 Chr.6: 109494211-109494211 on Build GRCh38
rs777922099 Chr.6: 109494003-109494003 on Build GRCh38
rs372465779 Chr.6: 109494058-109494058 on Build GRCh38
rs567225386 Chr.6: 109494301-109494301 on Build GRCh38
rs1022009641 Chr.6: 109494129-109494129 on Build GRCh38
rs747512776 Chr.6: 109494102-109494102 on Build GRCh38
rs911048771 Chr.6: 109494272-109494272 on Build GRCh38
rs377123845 Chr.6: 109494064-109494064 on Build GRCh38
rs754869300 Chr.6: 109493999-109493999 on Build GRCh38
rs1035178083 Chr.6: 109494090-109494090 on Build GRCh38
rs749260551 Chr.6: 109494025-109494025 on Build GRCh38
rs975003381 Chr.6: 109494401-109494401 on Build GRCh38

View Full Product Details