Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109497401-109497939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK9
Entrez Gene ID
221264
Gene Name
adenylate kinase 9
Gene Aliases
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1
UniGene
Hs.205144
Gene Chromosome Location
Chr.6: 109492855-109691253 on Build GRCh38
Transcripts
NM_001145128.2 XM_006715376.3 XM_011535550.2 XM_011535552.2 XM_011535553.2 XM_011535554.2 XM_011535558.1 XM_017010385.1 XM_017010387.1 XM_017010389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1754419 c.5223A>C p.E1741D Chr.6: 109497557-109497557 on Build GRCh38
6597379 c.506T>G p.F169C Chr.6: 109497471-109497471 on Build GRCh38
1697740 c.5152C>T p.R1718* Chr.6: 109497860-109497860 on Build GRCh38
1072012 c.433G>A p.G145S Chr.6: 109497544-109497544 on Build GRCh38
6478394 c.469A>G p.I157V Chr.6: 109497508-109497508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748847137 Chr.6: 109497747-109497747 on Build GRCh38
rs984873703 Chr.6: 109497447-109497447 on Build GRCh38
rs770285264 Chr.6: 109497428-109497428 on Build GRCh38
rs760547047 Chr.6: 109497525-109497525 on Build GRCh38
rs545753398 Chr.6: 109497705-109497705 on Build GRCh38
rs767002176 Chr.6: 109497599-109497599 on Build GRCh38
rs774082870 Chr.6: 109497823-109497823 on Build GRCh38
rs778297280 Chr.6: 109497896-109497896 on Build GRCh38
rs763162878 Chr.6: 109497815-109497815 on Build GRCh38
rs370370966 Chr.6: 109497531-109497531 on Build GRCh38
rs374592203 Chr.6: 109497829-109497829 on Build GRCh38
rs751607519 Chr.6: 109497833-109497833 on Build GRCh38
rs556435529 Chr.6: 109497731-109497731 on Build GRCh38
rs144863971 Chr.6: 109497697-109497697 on Build GRCh38
rs777270273 Chr.6: 109497449-109497449 on Build GRCh38
rs952076013 Chr.6: 109497716-109497716 on Build GRCh38
rs759690458 Chr.6: 109497839-109497839 on Build GRCh38
rs1049696489 Chr.6: 109497696-109497696 on Build GRCh38
rs926738509 Chr.6: 109497455-109497455 on Build GRCh38
rs754371911 Chr.6: 109497886-109497886 on Build GRCh38
rs75512843 Chr.6: 109497741-109497741 on Build GRCh38
rs751698093 Chr.6: 109497586-109497586 on Build GRCh38
rs764966539 Chr.6: 109497501-109497502 on Build GRCh38
rs938172849 Chr.6: 109497457-109497457 on Build GRCh38
rs9487128 Chr.6: 109497503-109497503 on Build GRCh38
rs373953563 Chr.6: 109497753-109497753 on Build GRCh38
rs150270108 Chr.6: 109497510-109497510 on Build GRCh38
rs553993589 Chr.6: 109497596-109497596 on Build GRCh38
rs766754088 Chr.6: 109497581-109497581 on Build GRCh38
rs903502475 Chr.6: 109497553-109497553 on Build GRCh38
rs527373847 Chr.6: 109497779-109497779 on Build GRCh38
rs1032667040 Chr.6: 109497914-109497914 on Build GRCh38
rs748793685 Chr.6: 109497472-109497472 on Build GRCh38
rs778290881 Chr.6: 109497859-109497859 on Build GRCh38
rs757709048 Chr.6: 109497894-109497894 on Build GRCh38
rs771948940 Chr.6: 109497860-109497860 on Build GRCh38
rs775310575 Chr.6: 109497511-109497511 on Build GRCh38
rs1007982861 Chr.6: 109497677-109497677 on Build GRCh38
rs932629208 Chr.6: 109497666-109497666 on Build GRCh38
rs540758676 Chr.6: 109497808-109497808 on Build GRCh38
rs778239998 Chr.6: 109497612-109497612 on Build GRCh38
rs748806074 Chr.6: 109497623-109497623 on Build GRCh38
rs922615336 Chr.6: 109497588-109497588 on Build GRCh38
rs781431485 Chr.6: 109497598-109497598 on Build GRCh38
rs763109349 Chr.6: 109497561-109497561 on Build GRCh38
rs753047867 Chr.6: 109497603-109497603 on Build GRCh38
rs775157277 Chr.6: 109497773-109497773 on Build GRCh38
rs1019433783 Chr.6: 109497694-109497694 on Build GRCh38
rs756681184 Chr.6: 109497609-109497609 on Build GRCh38
rs748632941 Chr.6: 109497424-109497424 on Build GRCh38
rs1040254442 Chr.6: 109497573-109497573 on Build GRCh38
rs1049485618 Chr.6: 109497595-109497595 on Build GRCh38
rs534333328 Chr.6: 109497545-109497545 on Build GRCh38
rs754342987 Chr.6: 109497704-109497704 on Build GRCh38
rs752290352 Chr.6: 109497836-109497836 on Build GRCh38
rs753059975 Chr.6: 109497844-109497844 on Build GRCh38
rs888558889 Chr.6: 109497719-109497719 on Build GRCh38
rs189818350 Chr.6: 109497772-109497772 on Build GRCh38
rs1027339058 Chr.6: 109497708-109497708 on Build GRCh38
rs370853204 Chr.6: 109497843-109497843 on Build GRCh38
rs370727173 Chr.6: 109497814-109497814 on Build GRCh38
rs756447943 Chr.6: 109497855-109497855 on Build GRCh38
rs941444654 Chr.6: 109497725-109497725 on Build GRCh38
rs770629436 Chr.6: 109497496-109497496 on Build GRCh38
rs778356471 Chr.6: 109497754-109497754 on Build GRCh38
rs745532645 Chr.6: 109497771-109497771 on Build GRCh38

View Full Product Details