Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8534580-8535160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4E
Entrez Gene ID
26253
Gene Name
C-type lectin domain family 4 member E
Gene Aliases
CLECSF9, MINCLE
UniGene
Hs.236516
Gene Chromosome Location
Chr.12: 8533305-8541328 on Build GRCh38
Transcripts
NM_014358.3 XM_011520614.2 XM_011520615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3466188 c.647G>A p.G216E Chr.12: 8534651-8534651 on Build GRCh38
1737301 c.653C>A p.S218Y Chr.12: 8534645-8534645 on Build GRCh38
3466189 c.634C>T p.P212S Chr.12: 8534664-8534664 on Build GRCh38
4749628 c.589T>C p.C197R Chr.12: 8534709-8534709 on Build GRCh38
3466190 c.626G>A p.G209E Chr.12: 8534672-8534672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776432894 Chr.12: 8535112-8535112 on Build GRCh38
rs903555438 Chr.12: 8535093-8535093 on Build GRCh38
rs746789322 Chr.12: 8534832-8534832 on Build GRCh38
rs748557692 Chr.12: 8534653-8534653 on Build GRCh38
rs1039921075 Chr.12: 8535084-8535084 on Build GRCh38
rs748353394 Chr.12: 8534604-8534604 on Build GRCh38
rs770779385 Chr.12: 8534620-8534620 on Build GRCh38
rs776175940 Chr.12: 8534846-8534846 on Build GRCh38
rs760109138 Chr.12: 8534750-8534750 on Build GRCh38
rs777167306 Chr.12: 8534747-8534748 on Build GRCh38
rs369080920 Chr.12: 8534656-8534656 on Build GRCh38
rs759349567 Chr.12: 8534641-8534641 on Build GRCh38
rs1052504479 Chr.12: 8534991-8534991 on Build GRCh38
rs202073795 Chr.12: 8534639-8534639 on Build GRCh38
rs186242880 Chr.12: 8534917-8534917 on Build GRCh38
rs772849084 Chr.12: 8534860-8534860 on Build GRCh38
rs1050388343 Chr.12: 8534939-8534939 on Build GRCh38
rs748613322 Chr.12: 8534611-8534611 on Build GRCh38
rs373912455 Chr.12: 8534659-8534659 on Build GRCh38
rs976776472 Chr.12: 8534886-8534886 on Build GRCh38
rs768065710 Chr.12: 8534774-8534774 on Build GRCh38
rs753025511 Chr.12: 8534688-8534688 on Build GRCh38
rs1030692262 Chr.12: 8534711-8534711 on Build GRCh38
rs769970179 Chr.12: 8534607-8534607 on Build GRCh38
rs562475046 Chr.12: 8534893-8534893 on Build GRCh38
rs905076651 Chr.12: 8535081-8535081 on Build GRCh38
rs144868707 Chr.12: 8534811-8534811 on Build GRCh38
rs371106734 Chr.12: 8535115-8535115 on Build GRCh38
rs745861394 Chr.12: 8534824-8534824 on Build GRCh38
rs141145068 Chr.12: 8534803-8534803 on Build GRCh38
rs764046984 Chr.12: 8534958-8534958 on Build GRCh38
rs775019747 Chr.12: 8534986-8534986 on Build GRCh38
rs763221133 Chr.12: 8534676-8534676 on Build GRCh38
rs113510124 Chr.12: 8534633-8534633 on Build GRCh38
rs376973359 Chr.12: 8534856-8534856 on Build GRCh38
rs745524207 Chr.12: 8535018-8535018 on Build GRCh38
rs770996250 Chr.12: 8534698-8534698 on Build GRCh38
rs755780446 Chr.12: 8535011-8535011 on Build GRCh38
rs891293479 Chr.12: 8535019-8535019 on Build GRCh38
rs768952242 Chr.12: 8534646-8534647 on Build GRCh38
rs781519351 Chr.12: 8534606-8534606 on Build GRCh38
rs773578382 Chr.12: 8534807-8534808 on Build GRCh38
rs761318275 Chr.12: 8534657-8534657 on Build GRCh38
rs752500807 Chr.12: 8534977-8534977 on Build GRCh38
rs764797711 Chr.12: 8534744-8534744 on Build GRCh38
rs375803624 Chr.12: 8534642-8534642 on Build GRCh38
rs765787135 Chr.12: 8534793-8534793 on Build GRCh38
rs180999307 Chr.12: 8534775-8534775 on Build GRCh38
rs756443256 Chr.12: 8534799-8534799 on Build GRCh38
rs779896075 Chr.12: 8534826-8534826 on Build GRCh38
rs373339920 Chr.12: 8534650-8534650 on Build GRCh38
rs138191509 Chr.12: 8534965-8534965 on Build GRCh38
rs755437434 Chr.12: 8534683-8534683 on Build GRCh38
rs753773846 Chr.12: 8534918-8534918 on Build GRCh38
rs770254786 Chr.12: 8534646-8534646 on Build GRCh38
rs10841846 Chr.12: 8535108-8535108 on Build GRCh38
rs767089632 Chr.12: 8534993-8534993 on Build GRCh38
rs778940677 Chr.12: 8534821-8534821 on Build GRCh38
rs747214654 Chr.12: 8534639-8534640 on Build GRCh38
rs201811623 Chr.12: 8534781-8534781 on Build GRCh38
rs781676283 Chr.12: 8534684-8534684 on Build GRCh38
rs747817772 Chr.12: 8534851-8534851 on Build GRCh38
rs776285555 Chr.12: 8534730-8534730 on Build GRCh38
rs770452839 Chr.12: 8535095-8535095 on Build GRCh38
rs771901039 Chr.12: 8534703-8534703 on Build GRCh38
rs774218702 Chr.12: 8534628-8534628 on Build GRCh38
rs775162634 Chr.12: 8534645-8534645 on Build GRCh38
rs749317295 Chr.12: 8534618-8534618 on Build GRCh38
rs925375226 Chr.12: 8534954-8534956 on Build GRCh38
rs992406962 Chr.12: 8534794-8534794 on Build GRCh38
rs762017874 Chr.12: 8534766-8534766 on Build GRCh38
rs372995129 Chr.12: 8534728-8534728 on Build GRCh38
rs942991088 Chr.12: 8535037-8535037 on Build GRCh38
rs372099470 Chr.12: 8534614-8534614 on Build GRCh38
rs376271215 Chr.12: 8534825-8534825 on Build GRCh38
rs760441714 Chr.12: 8534710-8534710 on Build GRCh38
rs781583386 Chr.12: 8534599-8534599 on Build GRCh38
rs377061220 Chr.12: 8534691-8534691 on Build GRCh38
rs146546350 Chr.12: 8534690-8534690 on Build GRCh38
rs756454054 Chr.12: 8534720-8534720 on Build GRCh38
rs372963987 Chr.12: 8534831-8534831 on Build GRCh38
rs761249435 Chr.12: 8534737-8534737 on Build GRCh38
rs777477368 Chr.12: 8535000-8535000 on Build GRCh38
rs1013417035 Chr.12: 8535032-8535032 on Build GRCh38
rs768446474 Chr.12: 8534836-8534836 on Build GRCh38
rs752164006 Chr.12: 8534746-8534746 on Build GRCh38
rs775252361 Chr.12: 8534704-8534704 on Build GRCh38
rs746482562 Chr.12: 8535007-8535007 on Build GRCh38
rs369879941 Chr.12: 8534701-8534701 on Build GRCh38
rs778854931 Chr.12: 8534699-8534699 on Build GRCh38

View Full Product Details