Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78492919-78493517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTGFR
Entrez Gene ID
5737
Gene Name
prostaglandin F receptor
Gene Aliases
FP
UniGene
Hs.654365
Gene Chromosome Location
Chr.1: 78490974-78540701 on Build GRCh38
Transcripts
NM_000959.3 NM_001039585.1 XM_017001873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6881830 c.215G>A p.G72D Chr.1: 78492958-78492958 on Build GRCh38
5674767 c.656G>T p.C219F Chr.1: 78493399-78493399 on Build GRCh38
3985240 c.683T>C p.L228S Chr.1: 78493426-78493426 on Build GRCh38
414783 c.546G>A p.S182S Chr.1: 78493289-78493289 on Build GRCh38
3492570 c.365C>T p.P122L Chr.1: 78493108-78493108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547608444 Chr.1: 78493338-78493338 on Build GRCh38
rs772307440 Chr.1: 78493086-78493086 on Build GRCh38
rs745990444 Chr.1: 78493322-78493322 on Build GRCh38
rs775975756 Chr.1: 78493379-78493379 on Build GRCh38
rs1031401954 Chr.1: 78493386-78493386 on Build GRCh38
rs796106629 Chr.1: 78493006-78493006 on Build GRCh38
rs749120257 Chr.1: 78493233-78493233 on Build GRCh38
rs116532636 Chr.1: 78493476-78493476 on Build GRCh38
rs147607519 Chr.1: 78493318-78493318 on Build GRCh38
rs754839078 Chr.1: 78493323-78493323 on Build GRCh38
rs765817668 Chr.1: 78493289-78493289 on Build GRCh38
rs769693596 Chr.1: 78493140-78493140 on Build GRCh38
rs779341659 Chr.1: 78493082-78493082 on Build GRCh38
rs768658971 Chr.1: 78493240-78493240 on Build GRCh38
rs770979500 Chr.1: 78493227-78493227 on Build GRCh38
rs749974467 Chr.1: 78493059-78493059 on Build GRCh38
rs751711019 Chr.1: 78493315-78493315 on Build GRCh38
rs759423133 Chr.1: 78492976-78492976 on Build GRCh38
rs754233774 Chr.1: 78493189-78493189 on Build GRCh38
rs767075974 Chr.1: 78493054-78493054 on Build GRCh38
rs757511674 Chr.1: 78493317-78493317 on Build GRCh38
rs944226941 Chr.1: 78493436-78493436 on Build GRCh38
rs757835420 Chr.1: 78493226-78493226 on Build GRCh38
rs200749896 Chr.1: 78493068-78493068 on Build GRCh38
rs755367220 Chr.1: 78493071-78493071 on Build GRCh38
rs747952854 Chr.1: 78493330-78493330 on Build GRCh38
rs532815634 Chr.1: 78493040-78493040 on Build GRCh38
rs368886963 Chr.1: 78493486-78493486 on Build GRCh38
rs758433633 Chr.1: 78493490-78493490 on Build GRCh38
rs779100761 Chr.1: 78493196-78493196 on Build GRCh38
rs905232913 Chr.1: 78493176-78493176 on Build GRCh38
rs778022964 Chr.1: 78493218-78493218 on Build GRCh38
rs867042175 Chr.1: 78493195-78493195 on Build GRCh38
rs145225940 Chr.1: 78493445-78493445 on Build GRCh38
rs754322857 Chr.1: 78493257-78493257 on Build GRCh38
rs142013001 Chr.1: 78493177-78493177 on Build GRCh38
rs76806235 Chr.1: 78493085-78493085 on Build GRCh38
rs747135810 Chr.1: 78493131-78493131 on Build GRCh38
rs745393690 Chr.1: 78493195-78493196 on Build GRCh38
rs200068380 Chr.1: 78493267-78493267 on Build GRCh38
rs940551658 Chr.1: 78493336-78493336 on Build GRCh38
rs779042598 Chr.1: 78493329-78493329 on Build GRCh38
rs755551230 Chr.1: 78493407-78493407 on Build GRCh38
rs1045066876 Chr.1: 78493129-78493129 on Build GRCh38
rs144240090 Chr.1: 78493254-78493254 on Build GRCh38
rs775328895 Chr.1: 78493015-78493015 on Build GRCh38
rs775712841 Chr.1: 78493286-78493286 on Build GRCh38
rs753042266 Chr.1: 78493295-78493295 on Build GRCh38
rs200228266 Chr.1: 78493432-78493432 on Build GRCh38
rs780419713 Chr.1: 78493219-78493219 on Build GRCh38
rs988329081 Chr.1: 78493067-78493067 on Build GRCh38
rs762801817 Chr.1: 78493155-78493155 on Build GRCh38
rs774207493 Chr.1: 78493168-78493168 on Build GRCh38
rs372903767 Chr.1: 78493185-78493185 on Build GRCh38
rs12088246 Chr.1: 78493178-78493178 on Build GRCh38
rs758816165 Chr.1: 78493298-78493298 on Build GRCh38
rs778200974 Chr.1: 78492984-78492985 on Build GRCh38
rs770650859 Chr.1: 78493349-78493349 on Build GRCh38
rs765750868 Chr.1: 78493276-78493276 on Build GRCh38
rs756283790 Chr.1: 78493471-78493471 on Build GRCh38
rs760409524 Chr.1: 78493272-78493272 on Build GRCh38
rs146358187 Chr.1: 78493183-78493183 on Build GRCh38
rs867894526 Chr.1: 78493281-78493281 on Build GRCh38
rs769758684 Chr.1: 78492995-78492995 on Build GRCh38
rs753095259 Chr.1: 78493197-78493197 on Build GRCh38
rs757380076 Chr.1: 78493447-78493447 on Build GRCh38
rs775343965 Chr.1: 78493141-78493141 on Build GRCh38
rs768266962 Chr.1: 78493160-78493160 on Build GRCh38
rs771837091 Chr.1: 78493263-78493263 on Build GRCh38
rs751233611 Chr.1: 78493042-78493042 on Build GRCh38
rs776039904 Chr.1: 78493421-78493421 on Build GRCh38
rs982007917 Chr.1: 78493024-78493024 on Build GRCh38
rs922734190 Chr.1: 78492963-78492963 on Build GRCh38
rs765652939 Chr.1: 78493403-78493403 on Build GRCh38
rs371784163 Chr.1: 78493049-78493049 on Build GRCh38
rs375003751 Chr.1: 78493194-78493194 on Build GRCh38
rs879522256 Chr.1: 78493137-78493137 on Build GRCh38
rs778857364 Chr.1: 78493429-78493429 on Build GRCh38
rs766906575 Chr.1: 78493188-78493188 on Build GRCh38
rs115994459 Chr.1: 78493396-78493396 on Build GRCh38
rs777794939 Chr.1: 78493124-78493124 on Build GRCh38
rs773296843 Chr.1: 78492967-78492967 on Build GRCh38
rs747995888 Chr.1: 78493262-78493262 on Build GRCh38
rs116802591 Chr.1: 78493208-78493208 on Build GRCh38
rs887686684 Chr.1: 78493259-78493259 on Build GRCh38
rs142217993 Chr.1: 78493494-78493494 on Build GRCh38
rs772074577 Chr.1: 78492951-78492951 on Build GRCh38
rs369124989 Chr.1: 78493070-78493070 on Build GRCh38
rs571722411 Chr.1: 78492943-78492943 on Build GRCh38
rs750447377 Chr.1: 78493450-78493450 on Build GRCh38
rs767820312 Chr.1: 78493448-78493448 on Build GRCh38
rs975843052 Chr.1: 78492950-78492950 on Build GRCh38
rs762700474 Chr.1: 78493019-78493019 on Build GRCh38
rs866779560 Chr.1: 78493170-78493170 on Build GRCh38
rs764292482 Chr.1: 78493299-78493299 on Build GRCh38
rs771927042 Chr.1: 78493331-78493331 on Build GRCh38
rs781334182 Chr.1: 78493231-78493231 on Build GRCh38
rs144613797 Chr.1: 78493278-78493278 on Build GRCh38

View Full Product Details