Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35863231-35863820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF20
Entrez Gene ID
51230
Gene Name
PHD finger protein 20
Gene Aliases
C20orf104, GLEA2, HCA58, NZF, TDRD20A, TZP
UniGene
Hs.517044
Gene Chromosome Location
Chr.20: 35771999-35950377 on Build GRCh38
Transcripts
NM_016436.4 XM_005260421.2 XM_011528843.1 XM_011528844.1 XM_017027864.1 XM_017027865.1 XM_017027866.1 XM_017027867.1 XM_017027868.1 XM_017027869.1 XM_017027870.1 XM_017027871.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5930027 c.649G>A p.E217K Chr.20: 35863241-35863241 on Build GRCh38
5921730 c.641C>T p.P214L Chr.20: 35863233-35863233 on Build GRCh38
359684 c.759G>A p.L253L Chr.20: 35863351-35863351 on Build GRCh38
1262045 c.659A>G p.D220G Chr.20: 35863251-35863251 on Build GRCh38
5365789 c.808+6delT p.? Chr.20: 35863406-35863406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17093182 Chr.20: 35863618-35863618 on Build GRCh38
rs182242136 Chr.20: 35863267-35863267 on Build GRCh38
rs763124948 Chr.20: 35863446-35863446 on Build GRCh38
rs999638406 Chr.20: 35863484-35863484 on Build GRCh38
rs1023759359 Chr.20: 35863690-35863690 on Build GRCh38
rs761722923 Chr.20: 35863426-35863426 on Build GRCh38
rs750572738 Chr.20: 35863345-35863345 on Build GRCh38
rs928402360 Chr.20: 35863692-35863692 on Build GRCh38
rs761776074 Chr.20: 35863343-35863343 on Build GRCh38
rs750402610 Chr.20: 35863441-35863441 on Build GRCh38
rs985021929 Chr.20: 35863546-35863546 on Build GRCh38
rs947027087 Chr.20: 35863742-35863742 on Build GRCh38
rs1036262024 Chr.20: 35863669-35863669 on Build GRCh38
rs770492032 Chr.20: 35863364-35863364 on Build GRCh38
rs771515722 Chr.20: 35863488-35863488 on Build GRCh38
rs866371178 Chr.20: 35863436-35863436 on Build GRCh38
rs969447940 Chr.20: 35863445-35863445 on Build GRCh38
rs775226377 Chr.20: 35863411-35863411 on Build GRCh38
rs753749754 Chr.20: 35863342-35863342 on Build GRCh38
rs566574221 Chr.20: 35863584-35863584 on Build GRCh38
rs763967095 Chr.20: 35863333-35863333 on Build GRCh38
rs781724302 Chr.20: 35863355-35863355 on Build GRCh38
rs1034952592 Chr.20: 35863706-35863706 on Build GRCh38
rs148610392 Chr.20: 35863483-35863483 on Build GRCh38
rs771562037 Chr.20: 35863398-35863398 on Build GRCh38
rs867362238 Chr.20: 35863285-35863285 on Build GRCh38
rs373676967 Chr.20: 35863323-35863323 on Build GRCh38
rs745400759 Chr.20: 35863388-35863388 on Build GRCh38
rs763301798 Chr.20: 35863695-35863695 on Build GRCh38
rs775094069 Chr.20: 35863321-35863321 on Build GRCh38
rs768978383 Chr.20: 35863264-35863264 on Build GRCh38
rs946859009 Chr.20: 35863492-35863492 on Build GRCh38
rs767262328 Chr.20: 35863318-35863318 on Build GRCh38
rs118077118 Chr.20: 35863514-35863514 on Build GRCh38
rs182783791 Chr.20: 35863412-35863412 on Build GRCh38
rs865823589 Chr.20: 35863300-35863300 on Build GRCh38
rs921671613 Chr.20: 35863565-35863565 on Build GRCh38
rs755548587 Chr.20: 35863402-35863410 on Build GRCh38
rs940561221 Chr.20: 35863624-35863624 on Build GRCh38
rs557425287 Chr.20: 35863547-35863547 on Build GRCh38
rs1014731601 Chr.20: 35863296-35863296 on Build GRCh38
rs1048888503 Chr.20: 35863284-35863284 on Build GRCh38
rs952004312 Chr.20: 35863529-35863529 on Build GRCh38
rs376891422 Chr.20: 35863351-35863351 on Build GRCh38
rs982517239 Chr.20: 35863450-35863450 on Build GRCh38
rs866730577 Chr.20: 35863724-35863724 on Build GRCh38
rs765172410 Chr.20: 35863434-35863434 on Build GRCh38
rs768406385 Chr.20: 35863418-35863418 on Build GRCh38
rs936986778 Chr.20: 35863698-35863698 on Build GRCh38
rs568634168 Chr.20: 35863456-35863456 on Build GRCh38
rs772655513 Chr.20: 35863491-35863491 on Build GRCh38
rs754199635 Chr.20: 35863344-35863345 on Build GRCh38
rs1044263029 Chr.20: 35863795-35863795 on Build GRCh38
rs568355175 Chr.20: 35863485-35863485 on Build GRCh38
rs746322028 Chr.20: 35863670-35863670 on Build GRCh38
rs759109502 Chr.20: 35863313-35863313 on Build GRCh38
rs770545172 Chr.20: 35863278-35863278 on Build GRCh38
rs758491078 Chr.20: 35863348-35863348 on Build GRCh38
rs1003552336 Chr.20: 35863703-35863703 on Build GRCh38
rs17093184 Chr.20: 35863680-35863680 on Build GRCh38
rs185182541 Chr.20: 35863268-35863268 on Build GRCh38
rs371215042 Chr.20: 35863425-35863425 on Build GRCh38
rs895377093 Chr.20: 35863730-35863730 on Build GRCh38
rs2235991 Chr.20: 35863683-35863683 on Build GRCh38
rs780327307 Chr.20: 35863349-35863349 on Build GRCh38
rs188694651 Chr.20: 35863462-35863462 on Build GRCh38
rs34314319 Chr.20: 35863354-35863354 on Build GRCh38
rs143366294 Chr.20: 35863363-35863363 on Build GRCh38
rs1004721328 Chr.20: 35863292-35863292 on Build GRCh38
rs907583626 Chr.20: 35863286-35863286 on Build GRCh38
rs778556177 Chr.20: 35863366-35863366 on Build GRCh38
rs6119664 Chr.20: 35863314-35863314 on Build GRCh38
rs774047531 Chr.20: 35863297-35863297 on Build GRCh38
rs73095524 Chr.20: 35863471-35863471 on Build GRCh38
rs533619493 Chr.20: 35863608-35863608 on Build GRCh38
rs1051391757 Chr.20: 35863586-35863586 on Build GRCh38

View Full Product Details