Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174560330-174560909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WIPF1
Entrez Gene ID
7456
Gene Name
WAS/WASL interacting protein family member 1
Gene Aliases
PRPL-2, WAS2, WASPIP, WIP
UniGene
Hs.128067
Gene Chromosome Location
Chr.2: 174559574-174682899 on Build GRCh38
Transcripts
NM_001077269.1 NM_003387.4 XM_011511780.2

SNP Detail

SNP ID Chromosome Location
rs575489083 Chr.2: 174560879-174560879 on Build GRCh38
rs4972697 Chr.2: 174560748-174560748 on Build GRCh38
rs1035295573 Chr.2: 174560677-174560677 on Build GRCh38
rs547614207 Chr.2: 174560428-174560428 on Build GRCh38
rs10930680 Chr.2: 174560411-174560411 on Build GRCh38
rs914819766 Chr.2: 174560753-174560753 on Build GRCh38
rs964853163 Chr.2: 174560810-174560810 on Build GRCh38
rs887135544 Chr.2: 174560545-174560545 on Build GRCh38
rs746724365 Chr.2: 174560502-174560502 on Build GRCh38
rs367945942 Chr.2: 174560745-174560745 on Build GRCh38
rs181075821 Chr.2: 174560577-174560577 on Build GRCh38
rs183755408 Chr.2: 174560578-174560578 on Build GRCh38
rs1004108074 Chr.2: 174560585-174560585 on Build GRCh38
rs974923017 Chr.2: 174560828-174560828 on Build GRCh38
rs933389227 Chr.2: 174560862-174560862 on Build GRCh38
rs900584800 Chr.2: 174560515-174560515 on Build GRCh38
rs11555986 Chr.2: 174560586-174560586 on Build GRCh38
rs933426518 Chr.2: 174560562-174560562 on Build GRCh38
rs922124983 Chr.2: 174560832-174560832 on Build GRCh38
rs914705116 Chr.2: 174560491-174560491 on Build GRCh38
rs773759670 Chr.2: 174560615-174560615 on Build GRCh38
rs557021071 Chr.2: 174560819-174560819 on Build GRCh38
rs969814940 Chr.2: 174560626-174560626 on Build GRCh38
rs1038472228 Chr.2: 174560578-174560578 on Build GRCh38
rs955710319 Chr.2: 174560687-174560687 on Build GRCh38
rs983379383 Chr.2: 174560836-174560836 on Build GRCh38
rs896737964 Chr.2: 174560587-174560587 on Build GRCh38
rs868126233 Chr.2: 174560791-174560791 on Build GRCh38
rs974189733 Chr.2: 174560361-174560361 on Build GRCh38
rs940038578 Chr.2: 174560886-174560893 on Build GRCh38
rs1051076734 Chr.2: 174560510-174560510 on Build GRCh38
rs188683001 Chr.2: 174560597-174560597 on Build GRCh38
rs9287978 Chr.2: 174560401-174560401 on Build GRCh38
rs531896359 Chr.2: 174560873-174560873 on Build GRCh38
rs3205060 Chr.2: 174560618-174560618 on Build GRCh38
rs142015388 Chr.2: 174560656-174560656 on Build GRCh38
rs189382730 Chr.2: 174560429-174560429 on Build GRCh38
rs767692719 Chr.2: 174560451-174560451 on Build GRCh38
rs943220860 Chr.2: 174560423-174560423 on Build GRCh38
rs923176956 Chr.2: 174560432-174560432 on Build GRCh38
rs970244262 Chr.2: 174560382-174560382 on Build GRCh38
rs1022720581 Chr.2: 174560375-174560375 on Build GRCh38
rs954598153 Chr.2: 174560833-174560833 on Build GRCh38
rs551787602 Chr.2: 174560575-174560575 on Build GRCh38
rs543796467 Chr.2: 174560383-174560383 on Build GRCh38
rs529374411 Chr.2: 174560494-174560494 on Build GRCh38
rs947567803 Chr.2: 174560493-174560493 on Build GRCh38
rs995343125 Chr.2: 174560605-174560605 on Build GRCh38
rs1039645726 Chr.2: 174560505-174560505 on Build GRCh38
rs533307341 Chr.2: 174560474-174560474 on Build GRCh38
rs548948633 Chr.2: 174560581-174560581 on Build GRCh38

View Full Product Details