Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103560339-103560912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG5
Entrez Gene ID
9529
Gene Name
BCL2 associated athanogene 5
Gene Aliases
BAG-5
UniGene
Hs.5443
Gene Chromosome Location
Chr.14: 103556544-103562814 on Build GRCh38
Transcripts
NM_001015048.2 NM_001015049.2 NM_004873.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5847621 c.631delG p.E211fs*13 Chr.14: 103560534-103560534 on Build GRCh38
953646 c.754T>G p.L252V Chr.14: 103560411-103560411 on Build GRCh38
374771 c.490G>T p.E164* Chr.14: 103560675-103560675 on Build GRCh38
6075282 c.652G>A p.E218K Chr.14: 103560636-103560636 on Build GRCh38
953648 c.429C>A p.I143I Chr.14: 103560736-103560736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007408769 Chr.14: 103560372-103560372 on Build GRCh38
rs377474437 Chr.14: 103560773-103560773 on Build GRCh38
rs778094001 Chr.14: 103560535-103560537 on Build GRCh38
rs777921086 Chr.14: 103560663-103560663 on Build GRCh38
rs921759055 Chr.14: 103560370-103560370 on Build GRCh38
rs780494384 Chr.14: 103560776-103560776 on Build GRCh38
rs185009666 Chr.14: 103560771-103560771 on Build GRCh38
rs752767387 Chr.14: 103560653-103560653 on Build GRCh38
rs765048453 Chr.14: 103560844-103560844 on Build GRCh38
rs972727028 Chr.14: 103560416-103560416 on Build GRCh38
rs749416088 Chr.14: 103560600-103560600 on Build GRCh38
rs538974554 Chr.14: 103560876-103560876 on Build GRCh38
rs1019682000 Chr.14: 103560382-103560382 on Build GRCh38
rs141215432 Chr.14: 103560461-103560461 on Build GRCh38
rs151161046 Chr.14: 103560408-103560408 on Build GRCh38
rs759518377 Chr.14: 103560837-103560837 on Build GRCh38
rs17854644 Chr.14: 103560694-103560694 on Build GRCh38
rs150155575 Chr.14: 103560822-103560822 on Build GRCh38
rs201699944 Chr.14: 103560466-103560466 on Build GRCh38
rs774049157 Chr.14: 103560712-103560712 on Build GRCh38
rs750356043 Chr.14: 103560551-103560551 on Build GRCh38
rs187928827 Chr.14: 103560890-103560890 on Build GRCh38
rs774897735 Chr.14: 103560613-103560613 on Build GRCh38
rs768986787 Chr.14: 103560782-103560782 on Build GRCh38
rs756420106 Chr.14: 103560426-103560428 on Build GRCh38
rs571692426 Chr.14: 103560659-103560659 on Build GRCh38
rs928444706 Chr.14: 103560495-103560495 on Build GRCh38
rs367847266 Chr.14: 103560598-103560598 on Build GRCh38
rs748792134 Chr.14: 103560702-103560702 on Build GRCh38
rs758276272 Chr.14: 103560419-103560419 on Build GRCh38
rs147561053 Chr.14: 103560757-103560757 on Build GRCh38
rs772819438 Chr.14: 103560811-103560811 on Build GRCh38
rs749789273 Chr.14: 103560781-103560781 on Build GRCh38
rs947616026 Chr.14: 103560558-103560558 on Build GRCh38
rs1044569278 Chr.14: 103560499-103560499 on Build GRCh38
rs368579842 Chr.14: 103560602-103560602 on Build GRCh38
rs906014476 Chr.14: 103560576-103560576 on Build GRCh38
rs776044539 Chr.14: 103560697-103560697 on Build GRCh38
rs760450858 Chr.14: 103560814-103560814 on Build GRCh38
rs759181274 Chr.14: 103560486-103560486 on Build GRCh38
rs753807360 Chr.14: 103560758-103560758 on Build GRCh38
rs754111260 Chr.14: 103560891-103560891 on Build GRCh38
rs1007050187 Chr.14: 103560718-103560718 on Build GRCh38
rs777446271 Chr.14: 103560430-103560430 on Build GRCh38
rs192732420 Chr.14: 103560625-103560625 on Build GRCh38
rs142309294 Chr.14: 103560768-103560768 on Build GRCh38
rs893510968 Chr.14: 103560549-103560549 on Build GRCh38
rs370330698 Chr.14: 103560810-103560810 on Build GRCh38
rs146938781 Chr.14: 103560748-103560748 on Build GRCh38
rs773010949 Chr.14: 103560733-103560733 on Build GRCh38
rs895904484 Chr.14: 103560618-103560618 on Build GRCh38
rs751304561 Chr.14: 103560432-103560432 on Build GRCh38
rs748217159 Chr.14: 103560575-103560575 on Build GRCh38
rs914892996 Chr.14: 103560861-103560861 on Build GRCh38
rs751626879 Chr.14: 103560769-103560769 on Build GRCh38
rs747608201 Chr.14: 103560805-103560805 on Build GRCh38
rs766289322 Chr.14: 103560650-103560650 on Build GRCh38
rs751335526 Chr.14: 103560505-103560505 on Build GRCh38
rs766951129 Chr.14: 103560374-103560374 on Build GRCh38
rs763704552 Chr.14: 103560623-103560623 on Build GRCh38
rs768693021 Chr.14: 103560465-103560465 on Build GRCh38
rs1052725417 Chr.14: 103560562-103560562 on Build GRCh38
rs777243034 Chr.14: 103560484-103560484 on Build GRCh38
rs747216872 Chr.14: 103560666-103560666 on Build GRCh38
rs770229250 Chr.14: 103560678-103560678 on Build GRCh38
rs750671414 Chr.14: 103560859-103560859 on Build GRCh38
rs767484038 Chr.14: 103560550-103560550 on Build GRCh38
rs201101341 Chr.14: 103560560-103560560 on Build GRCh38
rs778425552 Chr.14: 103560789-103560789 on Build GRCh38
rs909120395 Chr.14: 103560863-103560863 on Build GRCh38
rs144367794 Chr.14: 103560892-103560892 on Build GRCh38
rs762520743 Chr.14: 103560503-103560503 on Build GRCh38
rs751715026 Chr.14: 103560852-103560852 on Build GRCh38
rs778861866 Chr.14: 103560570-103560570 on Build GRCh38
rs771028859 Chr.14: 103560691-103560691 on Build GRCh38
rs889979870 Chr.14: 103560700-103560700 on Build GRCh38
rs372348583 Chr.14: 103560441-103560441 on Build GRCh38
rs372338173 Chr.14: 103560400-103560400 on Build GRCh38
rs770954774 Chr.14: 103560773-103560775 on Build GRCh38
rs748131386 Chr.14: 103560619-103560619 on Build GRCh38
rs911610105 Chr.14: 103560828-103560828 on Build GRCh38
rs776227351 Chr.14: 103560835-103560835 on Build GRCh38
rs756494455 Chr.14: 103560881-103560881 on Build GRCh38
rs756076256 Chr.14: 103560647-103560647 on Build GRCh38
rs753418462 Chr.14: 103560376-103560376 on Build GRCh38
rs772269638 Chr.14: 103560473-103560473 on Build GRCh38
rs773478504 Chr.14: 103560474-103560474 on Build GRCh38
rs368946901 Chr.14: 103560710-103560710 on Build GRCh38
rs1052109162 Chr.14: 103560498-103560498 on Build GRCh38
rs1043673900 Chr.14: 103560893-103560893 on Build GRCh38
rs371767275 Chr.14: 103560600-103560600 on Build GRCh38
rs375755201 Chr.14: 103560676-103560676 on Build GRCh38
rs767287868 Chr.14: 103560636-103560636 on Build GRCh38
rs757025881 Chr.14: 103560536-103560536 on Build GRCh38
rs759091597 Chr.14: 103560403-103560403 on Build GRCh38
rs757492335 Chr.14: 103560675-103560675 on Build GRCh38
rs761048937 Chr.14: 103560475-103560475 on Build GRCh38
rs375395190 Chr.14: 103560857-103560857 on Build GRCh38
rs373142750 Chr.14: 103560760-103560760 on Build GRCh38
rs767116401 Chr.14: 103560729-103560729 on Build GRCh38
rs761965282 Chr.14: 103560855-103560855 on Build GRCh38
rs750151456 Chr.14: 103560643-103560643 on Build GRCh38
rs750869599 Chr.14: 103560772-103560772 on Build GRCh38
rs778032103 Chr.14: 103560577-103560577 on Build GRCh38
rs761663464 Chr.14: 103560627-103560627 on Build GRCh38
rs560380530 Chr.14: 103560493-103560493 on Build GRCh38
rs909058318 Chr.14: 103560480-103560480 on Build GRCh38
rs761458604 Chr.14: 103560716-103560716 on Build GRCh38
rs771612201 Chr.14: 103560482-103560482 on Build GRCh38
rs748332347 Chr.14: 103560471-103560471 on Build GRCh38
rs770361362 Chr.14: 103560601-103560601 on Build GRCh38
rs757588826 Chr.14: 103560770-103560770 on Build GRCh38
rs1043340180 Chr.14: 103560561-103560562 on Build GRCh38
rs752283078 Chr.14: 103560415-103560415 on Build GRCh38
rs764822568 Chr.14: 103560492-103560492 on Build GRCh38
rs771413898 Chr.14: 103560580-103560580 on Build GRCh38
rs749420089 Chr.14: 103560462-103560462 on Build GRCh38

View Full Product Details