Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 129084100-129084653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM132D
Entrez Gene ID
121256
Gene Name
transmembrane protein 132D
Gene Aliases
MOLT, PPP1R153
UniGene
Hs.507268
Gene Chromosome Location
Chr.12: 129071726-129903667 on Build GRCh38
Transcripts
NM_133448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4955943 c.194A>G p.Q65R Chr.12: 129084566-129084566 on Build GRCh38
6803316 c.150C>A p.S50R Chr.12: 129084610-129084610 on Build GRCh38
936935 c.1504G>A p.V502M Chr.12: 129084642-129084642 on Build GRCh38
7331445 c.1618G>T p.G540C Chr.12: 129084528-129084528 on Build GRCh38
315979 c.1494C>A p.G498G Chr.12: 129084652-129084652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567219472 Chr.12: 129084337-129084337 on Build GRCh38
rs936182224 Chr.12: 129084218-129084218 on Build GRCh38
rs1024279628 Chr.12: 129084411-129084411 on Build GRCh38
rs770909071 Chr.12: 129084589-129084589 on Build GRCh38
rs867767964 Chr.12: 129084500-129084500 on Build GRCh38
rs778043494 Chr.12: 129084458-129084458 on Build GRCh38
rs990316526 Chr.12: 129084540-129084540 on Build GRCh38
rs555068393 Chr.12: 129084490-129084490 on Build GRCh38
rs565021234 Chr.12: 129084562-129084562 on Build GRCh38
rs766366335 Chr.12: 129084582-129084582 on Build GRCh38
rs145427504 Chr.12: 129084233-129084233 on Build GRCh38
rs190886410 Chr.12: 129084124-129084124 on Build GRCh38
rs952256664 Chr.12: 129084164-129084164 on Build GRCh38
rs376143160 Chr.12: 129084553-129084553 on Build GRCh38
rs550676571 Chr.12: 129084334-129084334 on Build GRCh38
rs10847755 Chr.12: 129084415-129084415 on Build GRCh38
rs752992861 Chr.12: 129084603-129084603 on Build GRCh38
rs746779283 Chr.12: 129084588-129084588 on Build GRCh38
rs778232035 Chr.12: 129084569-129084569 on Build GRCh38
rs544258949 Chr.12: 129084449-129084449 on Build GRCh38
rs929464545 Chr.12: 129084518-129084518 on Build GRCh38
rs545364344 Chr.12: 129084552-129084552 on Build GRCh38
rs546136196 Chr.12: 129084320-129084321 on Build GRCh38
rs760242454 Chr.12: 129084180-129084180 on Build GRCh38
rs764027287 Chr.12: 129084317-129084317 on Build GRCh38
rs1006194416 Chr.12: 129084166-129084166 on Build GRCh38
rs765508753 Chr.12: 129084606-129084606 on Build GRCh38
rs752106236 Chr.12: 129084626-129084626 on Build GRCh38
rs773097005 Chr.12: 129084511-129084511 on Build GRCh38
rs770309581 Chr.12: 129084232-129084232 on Build GRCh38
rs1026990741 Chr.12: 129084160-129084160 on Build GRCh38
rs763124843 Chr.12: 129084610-129084610 on Build GRCh38
rs79067456 Chr.12: 129084544-129084544 on Build GRCh38
rs571270830 Chr.12: 129084327-129084327 on Build GRCh38
rs753203016 Chr.12: 129084450-129084450 on Build GRCh38
rs889570730 Chr.12: 129084377-129084377 on Build GRCh38
rs199883487 Chr.12: 129084585-129084585 on Build GRCh38
rs746255800 Chr.12: 129084195-129084195 on Build GRCh38
rs528356197 Chr.12: 129084248-129084248 on Build GRCh38
rs994101124 Chr.12: 129084410-129084410 on Build GRCh38
rs553108684 Chr.12: 129084378-129084378 on Build GRCh38
rs767567527 Chr.12: 129084486-129084487 on Build GRCh38
rs768097547 Chr.12: 129084492-129084492 on Build GRCh38
rs542855447 Chr.12: 129084510-129084510 on Build GRCh38
rs372786001 Chr.12: 129084592-129084592 on Build GRCh38
rs369239393 Chr.12: 129084578-129084578 on Build GRCh38
rs980220213 Chr.12: 129084153-129084153 on Build GRCh38
rs752825227 Chr.12: 129084546-129084546 on Build GRCh38
rs144267550 Chr.12: 129084579-129084579 on Build GRCh38
rs927416023 Chr.12: 129084177-129084177 on Build GRCh38
rs753933036 Chr.12: 129084446-129084446 on Build GRCh38
rs549170842 Chr.12: 129084196-129084196 on Build GRCh38
rs764607310 Chr.12: 129084550-129084550 on Build GRCh38
rs779654730 Chr.12: 129084557-129084557 on Build GRCh38
rs183433591 Chr.12: 129084386-129084386 on Build GRCh38
rs981490222 Chr.12: 129084326-129084326 on Build GRCh38
rs766678401 Chr.12: 129084516-129084516 on Build GRCh38
rs755052788 Chr.12: 129084448-129084448 on Build GRCh38
rs575628199 Chr.12: 129084439-129084439 on Build GRCh38
rs112565551 Chr.12: 129084397-129084397 on Build GRCh38
rs528294635 Chr.12: 129084547-129084547 on Build GRCh38
rs748949588 Chr.12: 129084561-129084561 on Build GRCh38
rs763644523 Chr.12: 129084451-129084451 on Build GRCh38
rs769386298 Chr.12: 129084484-129084484 on Build GRCh38
rs946324797 Chr.12: 129084262-129084262 on Build GRCh38
rs551401954 Chr.12: 129084298-129084298 on Build GRCh38
rs536634970 Chr.12: 129084333-129084333 on Build GRCh38
rs774329506 Chr.12: 129084494-129084494 on Build GRCh38
rs574953184 Chr.12: 129084508-129084508 on Build GRCh38
rs775382777 Chr.12: 129084607-129084607 on Build GRCh38
rs536462848 Chr.12: 129084351-129084351 on Build GRCh38
rs913826761 Chr.12: 129084379-129084379 on Build GRCh38
rs760581231 Chr.12: 129084514-129084514 on Build GRCh38
rs747387094 Chr.12: 129084472-129084472 on Build GRCh38
rs140357606 Chr.12: 129084165-129084165 on Build GRCh38
rs993798502 Chr.12: 129084151-129084151 on Build GRCh38
rs376007280 Chr.12: 129084593-129084593 on Build GRCh38
rs754162885 Chr.12: 129084526-129084526 on Build GRCh38
rs764550024 Chr.12: 129084613-129084613 on Build GRCh38
rs1007304373 Chr.12: 129084122-129084122 on Build GRCh38

View Full Product Details