Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20222607-20223186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJB6
Entrez Gene ID
10804
Gene Name
gap junction protein beta 6
Gene Aliases
CX30, DFNA3, DFNA3B, DFNB1B, ECTD2, ED2, EDH, HED, HED2
UniGene
Hs.511757
Gene Chromosome Location
Chr.13: 20221962-20232395 on Build GRCh38
Transcripts
NM_001110219.2 NM_001110220.2 NM_001110221.2 NM_006783.4 XM_017020357.1 XM_017020358.1 XM_017020359.1 XM_017020360.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1718952 c.439G>A p.E147K Chr.13: 20223042-20223042 on Build GRCh38
6671235 c.689delA p.N230fs*7 Chr.13: 20222792-20222792 on Build GRCh38
6671234 c.633C>T p.C211C Chr.13: 20222848-20222848 on Build GRCh38
6451307 c.635A>G p.Y212C Chr.13: 20222846-20222846 on Build GRCh38
1706506 c.323G>A p.R108Q Chr.13: 20223158-20223158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756650660 Chr.13: 20222904-20222904 on Build GRCh38
rs761590601 Chr.13: 20222977-20222977 on Build GRCh38
rs779020990 Chr.13: 20222914-20222914 on Build GRCh38
rs138571061 Chr.13: 20222815-20222815 on Build GRCh38
rs749296000 Chr.13: 20223054-20223054 on Build GRCh38
rs776569668 Chr.13: 20222773-20222773 on Build GRCh38
rs754202125 Chr.13: 20223101-20223101 on Build GRCh38
rs1008818567 Chr.13: 20222684-20222684 on Build GRCh38
rs759639082 Chr.13: 20222677-20222677 on Build GRCh38
rs765642698 Chr.13: 20223122-20223122 on Build GRCh38
rs866019060 Chr.13: 20223003-20223003 on Build GRCh38
rs771392368 Chr.13: 20222742-20222742 on Build GRCh38
rs771466506 Chr.13: 20222669-20222669 on Build GRCh38
rs769932318 Chr.13: 20222687-20222687 on Build GRCh38
rs1004674545 Chr.13: 20222961-20222961 on Build GRCh38
rs758113938 Chr.13: 20222732-20222732 on Build GRCh38
rs548806574 Chr.13: 20222665-20222665 on Build GRCh38
rs955247134 Chr.13: 20223120-20223120 on Build GRCh38
rs752585451 Chr.13: 20222946-20222946 on Build GRCh38
rs1014466160 Chr.13: 20222777-20222777 on Build GRCh38
rs921827020 Chr.13: 20222877-20222877 on Build GRCh38
rs754273614 Chr.13: 20223002-20223002 on Build GRCh38
rs778441412 Chr.13: 20222656-20222656 on Build GRCh38
rs773950908 Chr.13: 20223078-20223078 on Build GRCh38
rs757539461 Chr.13: 20223132-20223132 on Build GRCh38
rs201783640 Chr.13: 20223053-20223053 on Build GRCh38
rs972404271 Chr.13: 20222839-20222839 on Build GRCh38
rs571030455 Chr.13: 20222643-20222643 on Build GRCh38
rs779119807 Chr.13: 20223023-20223023 on Build GRCh38
rs755468835 Chr.13: 20223012-20223012 on Build GRCh38
rs530181250 Chr.13: 20223128-20223128 on Build GRCh38
rs143766955 Chr.13: 20223142-20223142 on Build GRCh38
rs536309664 Chr.13: 20222632-20222632 on Build GRCh38
rs748558021 Chr.13: 20222652-20222652 on Build GRCh38
rs767980664 Chr.13: 20222896-20222896 on Build GRCh38
rs200035713 Chr.13: 20222766-20222766 on Build GRCh38
rs771265693 Chr.13: 20222736-20222737 on Build GRCh38
rs772862268 Chr.13: 20223159-20223159 on Build GRCh38
rs781689203 Chr.13: 20222745-20222745 on Build GRCh38
rs150583213 Chr.13: 20222965-20222965 on Build GRCh38
rs189971962 Chr.13: 20223085-20223085 on Build GRCh38
rs769645915 Chr.13: 20223038-20223038 on Build GRCh38
rs1001540278 Chr.13: 20223021-20223021 on Build GRCh38
rs968538869 Chr.13: 20223094-20223094 on Build GRCh38
rs773027260 Chr.13: 20222760-20222760 on Build GRCh38
rs146231737 Chr.13: 20222862-20222862 on Build GRCh38
rs145762940 Chr.13: 20223001-20223001 on Build GRCh38
rs757602049 Chr.13: 20222800-20222800 on Build GRCh38
rs771412904 Chr.13: 20223158-20223158 on Build GRCh38
rs1030054222 Chr.13: 20222634-20222634 on Build GRCh38
rs758698687 Chr.13: 20223025-20223025 on Build GRCh38
rs556168075 Chr.13: 20222758-20222758 on Build GRCh38
rs868439836 Chr.13: 20222910-20222910 on Build GRCh38
rs778717409 Chr.13: 20222726-20222726 on Build GRCh38
rs200881320 Chr.13: 20222888-20222888 on Build GRCh38
rs777143831 Chr.13: 20222897-20222897 on Build GRCh38
rs139711065 Chr.13: 20222987-20222987 on Build GRCh38
rs752343828 Chr.13: 20222728-20222728 on Build GRCh38
rs749567285 Chr.13: 20222754-20222754 on Build GRCh38
rs111033338 Chr.13: 20222886-20222886 on Build GRCh38
rs144174802 Chr.13: 20222913-20222913 on Build GRCh38
rs763870146 Chr.13: 20222887-20222887 on Build GRCh38
rs767044398 Chr.13: 20222983-20222983 on Build GRCh38
rs754990415 Chr.13: 20223141-20223141 on Build GRCh38
rs758167011 Chr.13: 20222787-20222787 on Build GRCh38
rs749058656 Chr.13: 20223138-20223138 on Build GRCh38
rs143962007 Chr.13: 20222700-20222700 on Build GRCh38
rs745637886 Chr.13: 20222952-20222952 on Build GRCh38
rs749587391 Chr.13: 20222728-20222731 on Build GRCh38
rs769845331 Chr.13: 20222751-20222751 on Build GRCh38
rs778351708 Chr.13: 20222733-20222733 on Build GRCh38
rs145438428 Chr.13: 20223076-20223076 on Build GRCh38
rs913100819 Chr.13: 20222854-20222854 on Build GRCh38
rs760492705 Chr.13: 20222764-20222764 on Build GRCh38
rs372607790 Chr.13: 20222794-20222794 on Build GRCh38
rs746807651 Chr.13: 20222845-20222845 on Build GRCh38
rs199790650 Chr.13: 20222801-20222801 on Build GRCh38
rs1005406189 Chr.13: 20223082-20223082 on Build GRCh38
rs756597598 Chr.13: 20222809-20222809 on Build GRCh38
rs775669210 Chr.13: 20222995-20222996 on Build GRCh38
rs777414019 Chr.13: 20222724-20222725 on Build GRCh38
rs750204590 Chr.13: 20222706-20222706 on Build GRCh38
rs766791253 Chr.13: 20223099-20223099 on Build GRCh38
rs772501788 Chr.13: 20222654-20222654 on Build GRCh38
rs200480676 Chr.13: 20222963-20222963 on Build GRCh38
rs768912194 Chr.13: 20222829-20222829 on Build GRCh38
rs766245553 Chr.13: 20222770-20222770 on Build GRCh38
rs868640784 Chr.13: 20223114-20223114 on Build GRCh38
rs755713634 Chr.13: 20222686-20222686 on Build GRCh38
rs761202742 Chr.13: 20223083-20223083 on Build GRCh38
rs770543700 Chr.13: 20223166-20223166 on Build GRCh38
rs112845420 Chr.13: 20222670-20222670 on Build GRCh38
rs868769358 Chr.13: 20222858-20222858 on Build GRCh38
rs35002004 Chr.13: 20222992-20222992 on Build GRCh38
rs747371119 Chr.13: 20222739-20222739 on Build GRCh38
rs765775497 Chr.13: 20222928-20222928 on Build GRCh38
rs772181578 Chr.13: 20222925-20222925 on Build GRCh38
rs377061702 Chr.13: 20222967-20222967 on Build GRCh38
rs979580620 Chr.13: 20223153-20223153 on Build GRCh38
rs185491286 Chr.13: 20223137-20223137 on Build GRCh38
rs972769330 Chr.13: 20223096-20223096 on Build GRCh38
rs761027934 Chr.13: 20223091-20223091 on Build GRCh38
rs188399223 Chr.13: 20223161-20223161 on Build GRCh38
rs960114319 Chr.13: 20222841-20222841 on Build GRCh38
rs764901929 Chr.13: 20222783-20222783 on Build GRCh38
rs750748318 Chr.13: 20222898-20222898 on Build GRCh38
rs577832858 Chr.13: 20222795-20222795 on Build GRCh38
rs751774202 Chr.13: 20223130-20223130 on Build GRCh38
rs768935828 Chr.13: 20223037-20223038 on Build GRCh38
rs35277762 Chr.13: 20223005-20223005 on Build GRCh38
rs141752846 Chr.13: 20222850-20222850 on Build GRCh38
rs890203655 Chr.13: 20222954-20222954 on Build GRCh38
rs777810168 Chr.13: 20223030-20223030 on Build GRCh38
rs34298814 Chr.13: 20222961-20222962 on Build GRCh38
rs748464515 Chr.13: 20222917-20222917 on Build GRCh38
rs775700786 Chr.13: 20222752-20222752 on Build GRCh38
rs773719693 Chr.13: 20222968-20222968 on Build GRCh38
rs398124237 Chr.13: 20222791-20222792 on Build GRCh38
rs777064837 Chr.13: 20223137-20223137 on Build GRCh38
rs760079142 Chr.13: 20223110-20223110 on Build GRCh38
rs745965086 Chr.13: 20222746-20222746 on Build GRCh38
rs567122461 Chr.13: 20222696-20222696 on Build GRCh38
rs780513857 Chr.13: 20222912-20222912 on Build GRCh38
rs749332449 Chr.13: 20222819-20222819 on Build GRCh38
rs766305993 Chr.13: 20222709-20222709 on Build GRCh38
rs754458181 Chr.13: 20222725-20222725 on Build GRCh38
rs200353369 Chr.13: 20223088-20223088 on Build GRCh38
rs866050531 Chr.13: 20222790-20222790 on Build GRCh38
rs772157377 Chr.13: 20222827-20222828 on Build GRCh38
rs765237598 Chr.13: 20222864-20222864 on Build GRCh38
rs747722057 Chr.13: 20223155-20223155 on Build GRCh38
rs576686538 Chr.13: 20223035-20223035 on Build GRCh38
rs775528528 Chr.13: 20222964-20222964 on Build GRCh38
rs774580284 Chr.13: 20222831-20222831 on Build GRCh38
rs760853231 Chr.13: 20223006-20223016 on Build GRCh38
rs200674715 Chr.13: 20222874-20222874 on Build GRCh38
rs752253778 Chr.13: 20222786-20222786 on Build GRCh38
rs760594015 Chr.13: 20222708-20222708 on Build GRCh38
rs779458847 Chr.13: 20222746-20222747 on Build GRCh38
rs149845267 Chr.13: 20222715-20222715 on Build GRCh38
rs193292569 Chr.13: 20223123-20223123 on Build GRCh38
rs752690015 Chr.13: 20223024-20223024 on Build GRCh38
rs547428597 Chr.13: 20222863-20222863 on Build GRCh38
rs1027253577 Chr.13: 20223004-20223004 on Build GRCh38
rs751130707 Chr.13: 20222991-20222991 on Build GRCh38
rs758984964 Chr.13: 20222780-20222780 on Build GRCh38
rs890181881 Chr.13: 20222651-20222651 on Build GRCh38

View Full Product Details