Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49308753-49309262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4324
Entrez Gene ID
100422979
Gene Name
microRNA 4324
Gene Aliases
mir-4324
UniGene
Gene Chromosome Location
Chr.19: 49308797-49308868 on Build GRCh38
Transcripts
NR_036209.1 NM_014037.2 XM_005258820.2 XM_006723168.3 XM_011526859.2 XM_011526860.2 XM_011526861.2 XM_017026712.1

Gene Symbol
SLC6A16
Entrez Gene ID
28968
Gene Name
solute carrier family 6 member 16
Gene Aliases
NT5, NTT5
UniGene
Hs.130949
Gene Chromosome Location
Chr.19: 49289632-49332320 on Build GRCh38
Transcripts
NR_036209.1 NM_014037.2 XM_005258820.2 XM_006723168.3 XM_011526859.2 XM_011526860.2 XM_011526861.2 XM_017026712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4430692 c.1200G>A p.A400A Chr.19: 49308905-49308905 on Build GRCh38
3536787 c.1067C>T p.S356F Chr.19: 49309038-49309038 on Build GRCh38
3835365 c.1073C>T p.A358V Chr.19: 49309032-49309032 on Build GRCh38
4727668 c.992C>T p.S331L Chr.19: 49309113-49309113 on Build GRCh38
4727667 c.1035A>C p.Q345H Chr.19: 49309070-49309070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777986453 Chr.19: 49308985-49308985 on Build GRCh38
rs975836367 Chr.19: 49308788-49308788 on Build GRCh38
rs369751841 Chr.19: 49309036-49309036 on Build GRCh38
rs770462441 Chr.19: 49308993-49308993 on Build GRCh38
rs768550440 Chr.19: 49309074-49309074 on Build GRCh38
rs760402213 Chr.19: 49308910-49308910 on Build GRCh38
rs938017931 Chr.19: 49309217-49309217 on Build GRCh38
rs780549807 Chr.19: 49308779-49308779 on Build GRCh38
rs748482068 Chr.19: 49308891-49308891 on Build GRCh38
rs756557962 Chr.19: 49309143-49309143 on Build GRCh38
rs1026458331 Chr.19: 49309196-49309196 on Build GRCh38
rs751710599 Chr.19: 49309113-49309113 on Build GRCh38
rs1056601994 Chr.19: 49308878-49308878 on Build GRCh38
rs769205784 Chr.19: 49308792-49308792 on Build GRCh38
rs772627748 Chr.19: 49308812-49308812 on Build GRCh38
rs772628575 Chr.19: 49308868-49308868 on Build GRCh38
rs747335168 Chr.19: 49308783-49308783 on Build GRCh38
rs994000227 Chr.19: 49309175-49309175 on Build GRCh38
rs555259313 Chr.19: 49308902-49308902 on Build GRCh38
rs775963719 Chr.19: 49308984-49308984 on Build GRCh38
rs766233355 Chr.19: 49308818-49308818 on Build GRCh38
rs770532413 Chr.19: 49308900-49308900 on Build GRCh38
rs752461895 Chr.19: 49308847-49308847 on Build GRCh38
rs980050290 Chr.19: 49309208-49309208 on Build GRCh38
rs952299791 Chr.19: 49308901-49308901 on Build GRCh38
rs762468953 Chr.19: 49308817-49308817 on Build GRCh38
rs750721228 Chr.19: 49308918-49308918 on Build GRCh38
rs750479856 Chr.19: 49308864-49308864 on Build GRCh38
rs766471703 Chr.19: 49309112-49309112 on Build GRCh38
rs775289520 Chr.19: 49308907-49308907 on Build GRCh38
rs186859566 Chr.19: 49308866-49308866 on Build GRCh38
rs192423180 Chr.19: 49309141-49309141 on Build GRCh38
rs544732904 Chr.19: 49309014-49309014 on Build GRCh38
rs769346410 Chr.19: 49308893-49308893 on Build GRCh38
rs751657782 Chr.19: 49308965-49308965 on Build GRCh38
rs570510329 Chr.19: 49308976-49308976 on Build GRCh38
rs534129369 Chr.19: 49308906-49308906 on Build GRCh38
rs747572736 Chr.19: 49308875-49308875 on Build GRCh38
rs748888653 Chr.19: 49308896-49308896 on Build GRCh38
rs758848438 Chr.19: 49308775-49308775 on Build GRCh38
rs945440506 Chr.19: 49309231-49309231 on Build GRCh38
rs772855872 Chr.19: 49308895-49308895 on Build GRCh38
rs753314295 Chr.19: 49309138-49309138 on Build GRCh38
rs755571383 Chr.19: 49308968-49308968 on Build GRCh38
rs550970166 Chr.19: 49308854-49308854 on Build GRCh38
rs1056466612 Chr.19: 49309219-49309219 on Build GRCh38
rs975700340 Chr.19: 49309068-49309068 on Build GRCh38
rs199795374 Chr.19: 49309033-49309033 on Build GRCh38
rs947265293 Chr.19: 49309015-49309015 on Build GRCh38
rs983709788 Chr.19: 49309159-49309159 on Build GRCh38
rs765453084 Chr.19: 49308856-49308856 on Build GRCh38
rs769705453 Chr.19: 49309108-49309108 on Build GRCh38
rs549532104 Chr.19: 49309126-49309126 on Build GRCh38
rs777122216 Chr.19: 49308798-49308798 on Build GRCh38
rs756569943 Chr.19: 49308977-49308977 on Build GRCh38
rs138717616 Chr.19: 49308889-49308889 on Build GRCh38
rs759404533 Chr.19: 49309080-49309081 on Build GRCh38
rs755520021 Chr.19: 49308888-49308888 on Build GRCh38
rs905184108 Chr.19: 49309095-49309095 on Build GRCh38
rs745336274 Chr.19: 49309002-49309002 on Build GRCh38
rs761417160 Chr.19: 49309075-49309075 on Build GRCh38
rs2278405 Chr.19: 49309146-49309146 on Build GRCh38
rs747851962 Chr.19: 49308799-49308799 on Build GRCh38
rs941952878 Chr.19: 49309202-49309202 on Build GRCh38
rs79283132 Chr.19: 49309150-49309150 on Build GRCh38
rs749020068 Chr.19: 49308987-49308987 on Build GRCh38
rs148862704 Chr.19: 49308938-49308938 on Build GRCh38
rs927192812 Chr.19: 49309209-49309209 on Build GRCh38
rs759457721 Chr.19: 49308825-49308825 on Build GRCh38
rs923084602 Chr.19: 49308942-49308942 on Build GRCh38
rs941935826 Chr.19: 49308863-49308863 on Build GRCh38
rs573486901 Chr.19: 49308905-49308905 on Build GRCh38
rs765379330 Chr.19: 49308916-49308916 on Build GRCh38
rs896579751 Chr.19: 49309220-49309220 on Build GRCh38
rs767703362 Chr.19: 49309128-49309128 on Build GRCh38
rs143434861 Chr.19: 49309098-49309098 on Build GRCh38
rs779989558 Chr.19: 49308867-49308867 on Build GRCh38
rs778429898 Chr.19: 49308998-49308998 on Build GRCh38
rs267605583 Chr.19: 49309029-49309029 on Build GRCh38
rs776155130 Chr.19: 49308902-49308903 on Build GRCh38
rs746362642 Chr.19: 49308832-49308832 on Build GRCh38
rs767353494 Chr.19: 49308831-49308831 on Build GRCh38
rs909009433 Chr.19: 49309200-49309200 on Build GRCh38
rs760598577 Chr.19: 49309050-49309050 on Build GRCh38
rs865953393 Chr.19: 49309225-49309225 on Build GRCh38
rs775336685 Chr.19: 49309042-49309042 on Build GRCh38
rs756959132 Chr.19: 49309148-49309148 on Build GRCh38
rs775208263 Chr.19: 49309142-49309142 on Build GRCh38
rs779503344 Chr.19: 49308807-49308807 on Build GRCh38
rs879237333 Chr.19: 49308821-49308821 on Build GRCh38
rs778244729 Chr.19: 49309145-49309145 on Build GRCh38
rs766528845 Chr.19: 49308951-49308951 on Build GRCh38
rs968707198 Chr.19: 49308949-49308949 on Build GRCh38

View Full Product Details