Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44286586-44287169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF235
Entrez Gene ID
9310
Gene Name
zinc finger protein 235
Gene Aliases
ANF270, HZF6, ZFP93, ZNF270
UniGene
Hs.613424
Gene Chromosome Location
Chr.19: 44286348-44309093 on Build GRCh38
Transcripts
NM_004234.4 XM_005259412.4

SNP Detail

SNP ID Chromosome Location
rs527651016 Chr.19: 44286935-44286935 on Build GRCh38
rs899681325 Chr.19: 44286963-44286963 on Build GRCh38
rs992132512 Chr.19: 44287078-44287078 on Build GRCh38
rs552005961 Chr.19: 44286862-44286862 on Build GRCh38
rs987872138 Chr.19: 44286693-44286693 on Build GRCh38
rs193234315 Chr.19: 44286918-44286918 on Build GRCh38
rs1008273858 Chr.19: 44286923-44286923 on Build GRCh38
rs987157026 Chr.19: 44286870-44286870 on Build GRCh38
rs1002500273 Chr.19: 44286744-44286744 on Build GRCh38
rs567451549 Chr.19: 44287010-44287010 on Build GRCh38
rs547599865 Chr.19: 44286964-44286964 on Build GRCh38
rs184608645 Chr.19: 44287061-44287061 on Build GRCh38
rs937555141 Chr.19: 44287077-44287077 on Build GRCh38
rs189390772 Chr.19: 44287064-44287064 on Build GRCh38
rs777889298 Chr.19: 44286864-44286864 on Build GRCh38
rs920165133 Chr.19: 44286888-44286888 on Build GRCh38
rs533811752 Chr.19: 44287100-44287100 on Build GRCh38
rs913694912 Chr.19: 44286729-44286729 on Build GRCh38
rs1020761234 Chr.19: 44287028-44287028 on Build GRCh38
rs368510500 Chr.19: 44287035-44287035 on Build GRCh38
rs957878994 Chr.19: 44286780-44286780 on Build GRCh38
rs548810744 Chr.19: 44287039-44287039 on Build GRCh38
rs987841430 Chr.19: 44287004-44287004 on Build GRCh38
rs910181367 Chr.19: 44286877-44286877 on Build GRCh38
rs536332440 Chr.19: 44287021-44287021 on Build GRCh38
rs758452186 Chr.19: 44286845-44286845 on Build GRCh38
rs975357375 Chr.19: 44286633-44286633 on Build GRCh38
rs796518487 Chr.19: 44287143-44287143 on Build GRCh38
rs964268025 Chr.19: 44286629-44286629 on Build GRCh38
rs954442244 Chr.19: 44286860-44286860 on Build GRCh38
rs924916139 Chr.19: 44286638-44286638 on Build GRCh38
rs144668322 Chr.19: 44286621-44286621 on Build GRCh38
rs942984983 Chr.19: 44286879-44286879 on Build GRCh38
rs562550337 Chr.19: 44287020-44287020 on Build GRCh38
rs957690410 Chr.19: 44286991-44286991 on Build GRCh38
rs190126832 Chr.19: 44286667-44286667 on Build GRCh38
rs1002190339 Chr.19: 44287003-44287003 on Build GRCh38
rs931335987 Chr.19: 44286869-44286869 on Build GRCh38
rs1047219035 Chr.19: 44286930-44286930 on Build GRCh38
rs532092937 Chr.19: 44286850-44286850 on Build GRCh38
rs954492535 Chr.19: 44287038-44287038 on Build GRCh38
rs987957313 Chr.19: 44286786-44286786 on Build GRCh38
rs563061835 Chr.19: 44286806-44286806 on Build GRCh38
rs920218891 Chr.19: 44287136-44287136 on Build GRCh38
rs931305060 Chr.19: 44287144-44287144 on Build GRCh38
rs148517881 Chr.19: 44286755-44286755 on Build GRCh38
rs752373725 Chr.19: 44287063-44287063 on Build GRCh38
rs10420428 Chr.19: 44287082-44287082 on Build GRCh38
rs1045901095 Chr.19: 44286764-44286764 on Build GRCh38
rs557995541 Chr.19: 44287074-44287074 on Build GRCh38
rs1032287910 Chr.19: 44286965-44286965 on Build GRCh38
rs905119075 Chr.19: 44286647-44286647 on Build GRCh38
rs1046404833 Chr.19: 44286974-44286974 on Build GRCh38
rs937567006 Chr.19: 44286844-44286844 on Build GRCh38
rs905166199 Chr.19: 44286997-44286997 on Build GRCh38
rs1020642025 Chr.19: 44287007-44287007 on Build GRCh38
rs747220484 Chr.19: 44286942-44286942 on Build GRCh38
rs976095596 Chr.19: 44286880-44286880 on Build GRCh38
rs1020749917 Chr.19: 44286818-44286818 on Build GRCh38

View Full Product Details