Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169227764-169228346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929523
Entrez Gene ID
101929523
Gene Name
uncharacterized LOC101929523
Gene Aliases
-
UniGene
Hs.442696
Gene Chromosome Location
Chr.6: 169213212-169239565 on Build GRCh38
Transcripts
NR_134621.1 NM_003247.3

Gene Symbol
THBS2
Entrez Gene ID
7058
Gene Name
thrombospondin 2
Gene Aliases
TSP2
UniGene
Hs.371147
Gene Chromosome Location
Chr.6: 169215780-169254114 on Build GRCh38
Transcripts
NR_134621.1 NM_003247.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3622953 c.2404G>A p.D802N Chr.6: 169228137-169228137 on Build GRCh38
4733516 c.2333A>G p.N778S Chr.6: 169228208-169228208 on Build GRCh38
4733515 c.2369delC p.T790fs*75 Chr.6: 169228172-169228172 on Build GRCh38
6801116 c.2401G>A p.V801M Chr.6: 169228140-169228140 on Build GRCh38
1169070 c.2344G>A p.V782M Chr.6: 169228197-169228197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780718730 Chr.6: 169227810-169227810 on Build GRCh38
rs200191335 Chr.6: 169228218-169228218 on Build GRCh38
rs144009376 Chr.6: 169227989-169227989 on Build GRCh38
rs398049011 Chr.6: 169228089-169228089 on Build GRCh38
rs140209905 Chr.6: 169227942-169227942 on Build GRCh38
rs781584021 Chr.6: 169228308-169228308 on Build GRCh38
rs1036502898 Chr.6: 169227889-169227889 on Build GRCh38
rs548582850 Chr.6: 169227827-169227827 on Build GRCh38
rs756879651 Chr.6: 169228074-169228075 on Build GRCh38
rs768534061 Chr.6: 169227808-169227808 on Build GRCh38
rs766227731 Chr.6: 169228242-169228242 on Build GRCh38
rs192667734 Chr.6: 169228010-169228010 on Build GRCh38
rs548642248 Chr.6: 169228098-169228098 on Build GRCh38
rs966084778 Chr.6: 169228228-169228228 on Build GRCh38
rs746628894 Chr.6: 169228279-169228279 on Build GRCh38
rs528291070 Chr.6: 169228050-169228050 on Build GRCh38
rs777473240 Chr.6: 169228160-169228160 on Build GRCh38
rs552663357 Chr.6: 169227910-169227910 on Build GRCh38
rs770261346 Chr.6: 169228194-169228194 on Build GRCh38
rs564367414 Chr.6: 169228036-169228036 on Build GRCh38
rs66527130 Chr.6: 169228075-169228076 on Build GRCh38
rs773492528 Chr.6: 169228083-169228083 on Build GRCh38
rs761099567 Chr.6: 169228091-169228091 on Build GRCh38
rs757871737 Chr.6: 169228271-169228271 on Build GRCh38
rs780717145 Chr.6: 169228075-169228077 on Build GRCh38
rs778993260 Chr.6: 169228254-169228254 on Build GRCh38
rs530139073 Chr.6: 169227824-169227824 on Build GRCh38
rs575542546 Chr.6: 169228009-169228009 on Build GRCh38
rs9406327 Chr.6: 169227854-169227854 on Build GRCh38
rs960790634 Chr.6: 169227807-169227807 on Build GRCh38
rs370814319 Chr.6: 169228055-169228055 on Build GRCh38
rs142440409 Chr.6: 169227856-169227856 on Build GRCh38
rs1010082250 Chr.6: 169227809-169227809 on Build GRCh38
rs530887135 Chr.6: 169228197-169228197 on Build GRCh38
rs373105188 Chr.6: 169228295-169228295 on Build GRCh38
rs201343909 Chr.6: 169228123-169228123 on Build GRCh38
rs1056442915 Chr.6: 169227979-169227979 on Build GRCh38
rs777149891 Chr.6: 169228185-169228185 on Build GRCh38
rs771690963 Chr.6: 169228156-169228156 on Build GRCh38
rs753664960 Chr.6: 169228243-169228243 on Build GRCh38
rs1041592470 Chr.6: 169228163-169228163 on Build GRCh38
rs142094192 Chr.6: 169228270-169228270 on Build GRCh38
rs771257805 Chr.6: 169228177-169228177 on Build GRCh38
rs983797212 Chr.6: 169228013-169228015 on Build GRCh38
rs9406326 Chr.6: 169227804-169227804 on Build GRCh38
rs1018886059 Chr.6: 169228192-169228192 on Build GRCh38
rs1012364283 Chr.6: 169227869-169227869 on Build GRCh38
rs1002466469 Chr.6: 169227967-169227967 on Build GRCh38
rs1041269393 Chr.6: 169227820-169227820 on Build GRCh38
rs747351403 Chr.6: 169228164-169228164 on Build GRCh38
rs768742783 Chr.6: 169228325-169228325 on Build GRCh38
rs754559562 Chr.6: 169228140-169228140 on Build GRCh38
rs756608054 Chr.6: 169228075-169228078 on Build GRCh38
rs530203655 Chr.6: 169228092-169228092 on Build GRCh38
rs11284439 Chr.6: 169228075-169228075 on Build GRCh38
rs945896282 Chr.6: 169228102-169228102 on Build GRCh38
rs541451382 Chr.6: 169227966-169227966 on Build GRCh38
rs765950962 Chr.6: 169228030-169228030 on Build GRCh38
rs374399600 Chr.6: 169228249-169228249 on Build GRCh38
rs750272321 Chr.6: 169227928-169227928 on Build GRCh38
rs367576679 Chr.6: 169228276-169228276 on Build GRCh38
rs1021448337 Chr.6: 169228020-169228020 on Build GRCh38
rs187489761 Chr.6: 169227833-169227833 on Build GRCh38
rs200614383 Chr.6: 169228095-169228095 on Build GRCh38
rs111289787 Chr.6: 169227803-169227803 on Build GRCh38
rs368102843 Chr.6: 169228217-169228217 on Build GRCh38
rs910924435 Chr.6: 169228019-169228019 on Build GRCh38
rs750823912 Chr.6: 169228076-169228076 on Build GRCh38
rs747666963 Chr.6: 169228152-169228152 on Build GRCh38
rs762954399 Chr.6: 169228319-169228319 on Build GRCh38
rs184532797 Chr.6: 169228302-169228302 on Build GRCh38
rs753787752 Chr.6: 169228138-169228138 on Build GRCh38
rs569945503 Chr.6: 169228135-169228135 on Build GRCh38
rs552603239 Chr.6: 169228198-169228198 on Build GRCh38
rs6903835 Chr.6: 169227863-169227863 on Build GRCh38
rs970950970 Chr.6: 169227914-169227914 on Build GRCh38
rs755006363 Chr.6: 169228253-169228253 on Build GRCh38
rs570960089 Chr.6: 169227857-169227857 on Build GRCh38
rs919038311 Chr.6: 169227900-169227900 on Build GRCh38
rs905406462 Chr.6: 169227954-169227954 on Build GRCh38
rs535029137 Chr.6: 169228305-169228306 on Build GRCh38
rs746197811 Chr.6: 169228311-169228311 on Build GRCh38
rs147580962 Chr.6: 169228150-169228150 on Build GRCh38
rs952933273 Chr.6: 169228327-169228327 on Build GRCh38
rs759985256 Chr.6: 169228191-169228191 on Build GRCh38
rs967167368 Chr.6: 169227819-169227819 on Build GRCh38
rs374248057 Chr.6: 169228214-169228214 on Build GRCh38
rs377648669 Chr.6: 169228096-169228096 on Build GRCh38
rs200586551 Chr.6: 169228292-169228292 on Build GRCh38
rs763130514 Chr.6: 169228101-169228101 on Build GRCh38
rs567847941 Chr.6: 169228312-169228312 on Build GRCh38
rs755923430 Chr.6: 169228133-169228133 on Build GRCh38
rs559915902 Chr.6: 169228075-169228075 on Build GRCh38
rs944448754 Chr.6: 169227874-169227874 on Build GRCh38
rs113924621 Chr.6: 169228284-169228284 on Build GRCh38
rs754468894 Chr.6: 169228139-169228139 on Build GRCh38
rs764344581 Chr.6: 169228108-169228108 on Build GRCh38

View Full Product Details