Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44513163-44513629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPC1L1
Entrez Gene ID
29881
Gene Name
NPC1 like intracellular cholesterol transporter 1
Gene Aliases
NPC11L1
UniGene
Hs.567486
Gene Chromosome Location
Chr.7: 44512440-44541650 on Build GRCh38
Transcripts
NM_001101648.1 NM_013389.2 XM_011515326.2 XM_011515328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
338846 c.3925G>T p.E1309* Chr.7: 44513602-44513602 on Build GRCh38
1488569 c.3988G>C p.D1330H Chr.7: 44513539-44513539 on Build GRCh38
3381941 c.4006C>T p.H1336Y Chr.7: 44513521-44513521 on Build GRCh38
6485409 c.3999T>C p.Y1333Y Chr.7: 44513528-44513528 on Build GRCh38
119818 c.4011C>G p.S1337R Chr.7: 44513516-44513516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012888370 Chr.7: 44513378-44513378 on Build GRCh38
rs560915807 Chr.7: 44513536-44513536 on Build GRCh38
rs1011391773 Chr.7: 44513273-44513273 on Build GRCh38
rs217436 Chr.7: 44513317-44513317 on Build GRCh38
rs114488279 Chr.7: 44513540-44513540 on Build GRCh38
rs781219627 Chr.7: 44513596-44513596 on Build GRCh38
rs749674646 Chr.7: 44513503-44513503 on Build GRCh38
rs776843211 Chr.7: 44513396-44513396 on Build GRCh38
rs748475245 Chr.7: 44513397-44513397 on Build GRCh38
rs217435 Chr.7: 44513605-44513605 on Build GRCh38
rs924327330 Chr.7: 44513186-44513186 on Build GRCh38
rs545245568 Chr.7: 44513326-44513326 on Build GRCh38
rs777984273 Chr.7: 44513499-44513499 on Build GRCh38
rs1021273317 Chr.7: 44513563-44513563 on Build GRCh38
rs1002659936 Chr.7: 44513462-44513462 on Build GRCh38
rs375351864 Chr.7: 44513569-44513569 on Build GRCh38
rs764908618 Chr.7: 44513433-44513433 on Build GRCh38
rs763799879 Chr.7: 44513416-44513416 on Build GRCh38
rs763022264 Chr.7: 44513292-44513292 on Build GRCh38
rs770080176 Chr.7: 44513401-44513401 on Build GRCh38
rs761605724 Chr.7: 44513427-44513427 on Build GRCh38
rs770097198 Chr.7: 44513491-44513491 on Build GRCh38
rs943231529 Chr.7: 44513251-44513251 on Build GRCh38
rs763362144 Chr.7: 44513405-44513405 on Build GRCh38
rs750128847 Chr.7: 44513556-44513556 on Build GRCh38
rs1006176622 Chr.7: 44513357-44513357 on Build GRCh38
rs115805898 Chr.7: 44513585-44513585 on Build GRCh38
rs748350539 Chr.7: 44513481-44513481 on Build GRCh38
rs369777905 Chr.7: 44513583-44513583 on Build GRCh38
rs755084966 Chr.7: 44513458-44513458 on Build GRCh38
rs200195472 Chr.7: 44513439-44513439 on Build GRCh38
rs145396059 Chr.7: 44513341-44513341 on Build GRCh38
rs776319918 Chr.7: 44513545-44513545 on Build GRCh38
rs753567415 Chr.7: 44513419-44513419 on Build GRCh38
rs990104921 Chr.7: 44513214-44513214 on Build GRCh38
rs559858415 Chr.7: 44513315-44513315 on Build GRCh38
rs914494851 Chr.7: 44513247-44513247 on Build GRCh38
rs1037628500 Chr.7: 44513364-44513364 on Build GRCh38
rs1048266582 Chr.7: 44513330-44513330 on Build GRCh38
rs1031834716 Chr.7: 44513468-44513468 on Build GRCh38
rs746212091 Chr.7: 44513203-44513203 on Build GRCh38
rs775025237 Chr.7: 44513513-44513513 on Build GRCh38
rs371572587 Chr.7: 44513553-44513553 on Build GRCh38
rs770061436 Chr.7: 44513275-44513275 on Build GRCh38
rs766964251 Chr.7: 44513408-44513408 on Build GRCh38
rs981061313 Chr.7: 44513312-44513312 on Build GRCh38
rs367742890 Chr.7: 44513465-44513465 on Build GRCh38
rs1050310404 Chr.7: 44513520-44513520 on Build GRCh38
rs781347126 Chr.7: 44513467-44513467 on Build GRCh38
rs907514500 Chr.7: 44513420-44513420 on Build GRCh38
rs771271957 Chr.7: 44513508-44513508 on Build GRCh38
rs764999826 Chr.7: 44513554-44513554 on Build GRCh38
rs773677245 Chr.7: 44513402-44513402 on Build GRCh38
rs549885303 Chr.7: 44513597-44513597 on Build GRCh38
rs771348078 Chr.7: 44513610-44513610 on Build GRCh38
rs368864761 Chr.7: 44513270-44513270 on Build GRCh38
rs923074982 Chr.7: 44513287-44513287 on Build GRCh38
rs775779020 Chr.7: 44513278-44513278 on Build GRCh38
rs895414312 Chr.7: 44513375-44513375 on Build GRCh38
rs754960232 Chr.7: 44513592-44513592 on Build GRCh38
rs372864437 Chr.7: 44513413-44513413 on Build GRCh38
rs766188111 Chr.7: 44513581-44513581 on Build GRCh38
rs867880561 Chr.7: 44513222-44513222 on Build GRCh38
rs758255762 Chr.7: 44513444-44513444 on Build GRCh38
rs751572189 Chr.7: 44513457-44513457 on Build GRCh38
rs762127492 Chr.7: 44513329-44513329 on Build GRCh38
rs760722175 Chr.7: 44513363-44513363 on Build GRCh38
rs764252457 Chr.7: 44513316-44513316 on Build GRCh38
rs143889287 Chr.7: 44513598-44513598 on Build GRCh38
rs527948546 Chr.7: 44513392-44513392 on Build GRCh38
rs548075117 Chr.7: 44513550-44513550 on Build GRCh38

View Full Product Details