Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117099382-117099866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST7
Entrez Gene ID
7982
Gene Name
suppression of tumorigenicity 7
Gene Aliases
ETS7q, FAM4A, FAM4A1, HELG, RAY1, SEN4, TSG7
UniGene
Hs.368131
Gene Chromosome Location
Chr.7: 116953327-117230023 on Build GRCh38
Transcripts
NM_018412.3 NM_021908.2

Gene Symbol
ST7-AS2
Entrez Gene ID
93654
Gene Name
ST7 antisense RNA 2
Gene Aliases
ST7AS2, ST7OT2
UniGene
Hs.628891
Gene Chromosome Location
Chr.7: 117072072-117145592 on Build GRCh38
Transcripts
NM_018412.3 NM_021908.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7093221 c.28A>C p.M10L Chr.7: 117099767-117099767 on Build GRCh38
5796057 c.50C>T p.P17L Chr.7: 117099789-117099789 on Build GRCh38
7093220 c.157A>C p.M53L Chr.7: 117099767-117099767 on Build GRCh38
1647969 c.219A>T p.I73I Chr.7: 117099829-117099829 on Build GRCh38
7447644 c.95G>A p.G32E Chr.7: 117099834-117099834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759010682 Chr.7: 117099835-117099835 on Build GRCh38
rs1031166445 Chr.7: 117099716-117099716 on Build GRCh38
rs779872873 Chr.7: 117099732-117099732 on Build GRCh38
rs968254173 Chr.7: 117099652-117099652 on Build GRCh38
rs953739704 Chr.7: 117099537-117099537 on Build GRCh38
rs1036282711 Chr.7: 117099495-117099495 on Build GRCh38
rs917152246 Chr.7: 117099598-117099598 on Build GRCh38
rs948162671 Chr.7: 117099412-117099412 on Build GRCh38
rs150614816 Chr.7: 117099448-117099448 on Build GRCh38
rs139716982 Chr.7: 117099494-117099494 on Build GRCh38
rs1000063649 Chr.7: 117099678-117099678 on Build GRCh38
rs772281894 Chr.7: 117099749-117099750 on Build GRCh38
rs901794089 Chr.7: 117099508-117099508 on Build GRCh38
rs80169857 Chr.7: 117099650-117099650 on Build GRCh38
rs768636943 Chr.7: 117099766-117099766 on Build GRCh38
rs988686045 Chr.7: 117099580-117099580 on Build GRCh38
rs950031633 Chr.7: 117099599-117099599 on Build GRCh38
rs969330792 Chr.7: 117099790-117099790 on Build GRCh38
rs1011351422 Chr.7: 117099628-117099628 on Build GRCh38
rs978527693 Chr.7: 117099538-117099538 on Build GRCh38
rs372140830 Chr.7: 117099571-117099571 on Build GRCh38
rs763757342 Chr.7: 117099569-117099569 on Build GRCh38
rs773622593 Chr.7: 117099819-117099819 on Build GRCh38
rs368520848 Chr.7: 117099781-117099781 on Build GRCh38
rs961283677 Chr.7: 117099759-117099760 on Build GRCh38
rs749327257 Chr.7: 117099746-117099746 on Build GRCh38
rs998249349 Chr.7: 117099450-117099450 on Build GRCh38
rs750881338 Chr.7: 117099639-117099639 on Build GRCh38
rs1049111476 Chr.7: 117099414-117099414 on Build GRCh38
rs992351149 Chr.7: 117099764-117099764 on Build GRCh38
rs1016243920 Chr.7: 117099409-117099409 on Build GRCh38
rs376706985 Chr.7: 117099663-117099663 on Build GRCh38
rs535823473 Chr.7: 117099480-117099480 on Build GRCh38
rs982481620 Chr.7: 117099635-117099635 on Build GRCh38
rs772419572 Chr.7: 117099799-117099799 on Build GRCh38
rs530179945 Chr.7: 117099747-117099751 on Build GRCh38
rs984848240 Chr.7: 117099841-117099841 on Build GRCh38
rs567991907 Chr.7: 117099823-117099823 on Build GRCh38
rs570882202 Chr.7: 117099433-117099433 on Build GRCh38
rs774606952 Chr.7: 117099778-117099778 on Build GRCh38

View Full Product Details