Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 15108217-15108703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RERG
Entrez Gene ID
85004
Gene Name
RAS like estrogen regulated growth inhibitor
Gene Aliases
-
UniGene
Hs.199487
Gene Chromosome Location
Chr.12: 15107782-15221803 on Build GRCh38
Transcripts
NM_001190726.1 NM_032918.2 XM_017020121.1

SNP Detail

SNP ID Chromosome Location
rs930956290 Chr.12: 15108515-15108515 on Build GRCh38
rs953220580 Chr.12: 15108599-15108599 on Build GRCh38
rs528955915 Chr.12: 15108479-15108479 on Build GRCh38
rs1041659899 Chr.12: 15108374-15108374 on Build GRCh38
rs973502826 Chr.12: 15108545-15108545 on Build GRCh38
rs878952904 Chr.12: 15108336-15108336 on Build GRCh38
rs543039274 Chr.12: 15108427-15108427 on Build GRCh38
rs2080476 Chr.12: 15108246-15108246 on Build GRCh38
rs755846922 Chr.12: 15108611-15108613 on Build GRCh38
rs187282348 Chr.12: 15108352-15108352 on Build GRCh38
rs193258270 Chr.12: 15108461-15108461 on Build GRCh38
rs17834986 Chr.12: 15108503-15108503 on Build GRCh38
rs889330868 Chr.12: 15108544-15108544 on Build GRCh38
rs901995272 Chr.12: 15108569-15108569 on Build GRCh38
rs879380597 Chr.12: 15108247-15108247 on Build GRCh38
rs926535830 Chr.12: 15108407-15108407 on Build GRCh38
rs569606936 Chr.12: 15108583-15108583 on Build GRCh38
rs139122572 Chr.12: 15108506-15108506 on Build GRCh38
rs911688540 Chr.12: 15108669-15108669 on Build GRCh38
rs985897205 Chr.12: 15108618-15108618 on Build GRCh38
rs746931796 Chr.12: 15108309-15108309 on Build GRCh38
rs563523633 Chr.12: 15108485-15108485 on Build GRCh38
rs1023727527 Chr.12: 15108492-15108492 on Build GRCh38
rs374347532 Chr.12: 15108366-15108366 on Build GRCh38
rs749619374 Chr.12: 15108296-15108299 on Build GRCh38
rs1020993225 Chr.12: 15108658-15108658 on Build GRCh38
rs961735515 Chr.12: 15108525-15108525 on Build GRCh38
rs1012401939 Chr.12: 15108486-15108486 on Build GRCh38
rs16910619 Chr.12: 15108359-15108359 on Build GRCh38
rs2080475 Chr.12: 15108283-15108283 on Build GRCh38
rs1008243782 Chr.12: 15108561-15108561 on Build GRCh38
rs1053871313 Chr.12: 15108480-15108480 on Build GRCh38
rs968003429 Chr.12: 15108372-15108372 on Build GRCh38
rs1019082624 Chr.12: 15108568-15108568 on Build GRCh38
rs531382586 Chr.12: 15108405-15108405 on Build GRCh38
rs937938936 Chr.12: 15108465-15108465 on Build GRCh38
rs1050716518 Chr.12: 15108327-15108327 on Build GRCh38
rs779977446 Chr.12: 15108237-15108237 on Build GRCh38
rs7301809 Chr.12: 15108564-15108564 on Build GRCh38
rs551309906 Chr.12: 15108508-15108508 on Build GRCh38
rs1049430147 Chr.12: 15108521-15108521 on Build GRCh38

View Full Product Details