Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135445837-135446435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCANP1
Entrez Gene ID
140947
Gene Name
dendritic cell associated nuclear protein
Gene Aliases
C5orf20, DCNP1
UniGene
Hs.152477
Gene Chromosome Location
Chr.5: 135444214-135447348 on Build GRCh38
Transcripts
NM_130848.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7330005 c.690G>A p.L230L Chr.5: 135446419-135446419 on Build GRCh38
3608620 c.714G>A p.R238R Chr.5: 135446395-135446395 on Build GRCh38
3608621 c.686C>T p.P229L Chr.5: 135446423-135446423 on Build GRCh38
5527772 c.674C>T p.S225F Chr.5: 135446435-135446435 on Build GRCh38
5811646 c.700A>T p.S234C Chr.5: 135446409-135446409 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1006852064 Chr.5: 135446206-135446206 on Build GRCh38
rs180721424 Chr.5: 135446351-135446351 on Build GRCh38
rs1042328073 Chr.5: 135446286-135446286 on Build GRCh38
rs781303653 Chr.5: 135446345-135446345 on Build GRCh38
rs778421248 Chr.5: 135446008-135446008 on Build GRCh38
rs981638106 Chr.5: 135446010-135446010 on Build GRCh38
rs867558313 Chr.5: 135445876-135445876 on Build GRCh38
rs779879834 Chr.5: 135446161-135446161 on Build GRCh38
rs766592910 Chr.5: 135445885-135445885 on Build GRCh38
rs904385953 Chr.5: 135446130-135446130 on Build GRCh38
rs572439025 Chr.5: 135445914-135445914 on Build GRCh38
rs532981201 Chr.5: 135446160-135446160 on Build GRCh38
rs562659265 Chr.5: 135446135-135446135 on Build GRCh38
rs1041719631 Chr.5: 135446052-135446052 on Build GRCh38
rs572376932 Chr.5: 135446322-135446322 on Build GRCh38
rs997565354 Chr.5: 135446111-135446111 on Build GRCh38
rs535831984 Chr.5: 135446227-135446227 on Build GRCh38
rs555982123 Chr.5: 135446278-135446278 on Build GRCh38
rs868342315 Chr.5: 135446192-135446192 on Build GRCh38
rs567606009 Chr.5: 135446066-135446066 on Build GRCh38
rs367644701 Chr.5: 135446370-135446370 on Build GRCh38
rs889013930 Chr.5: 135446374-135446374 on Build GRCh38
rs538832057 Chr.5: 135446301-135446301 on Build GRCh38
rs531798677 Chr.5: 135446026-135446026 on Build GRCh38
rs778137686 Chr.5: 135446334-135446334 on Build GRCh38
rs893376678 Chr.5: 135446038-135446038 on Build GRCh38
rs1020050873 Chr.5: 135446223-135446223 on Build GRCh38
rs1006648834 Chr.5: 135446164-135446164 on Build GRCh38
rs17168355 Chr.5: 135446302-135446302 on Build GRCh38
rs139456966 Chr.5: 135446230-135446230 on Build GRCh38
rs560968296 Chr.5: 135445970-135445970 on Build GRCh38
rs950992139 Chr.5: 135446220-135446220 on Build GRCh38
rs548962185 Chr.5: 135446245-135446245 on Build GRCh38
rs756086139 Chr.5: 135446395-135446395 on Build GRCh38
rs35462904 Chr.5: 135446251-135446252 on Build GRCh38
rs115328649 Chr.5: 135446043-135446043 on Build GRCh38
rs1056467716 Chr.5: 135446029-135446029 on Build GRCh38
rs903300452 Chr.5: 135446055-135446055 on Build GRCh38
rs991656407 Chr.5: 135446246-135446246 on Build GRCh38
rs749960228 Chr.5: 135446014-135446014 on Build GRCh38
rs60881382 Chr.5: 135446285-135446285 on Build GRCh38
rs377423605 Chr.5: 135445987-135445987 on Build GRCh38
rs757516402 Chr.5: 135446343-135446343 on Build GRCh38
rs972872355 Chr.5: 135445886-135445886 on Build GRCh38
rs781331124 Chr.5: 135446393-135446393 on Build GRCh38
rs537898941 Chr.5: 135446271-135446271 on Build GRCh38
rs867040882 Chr.5: 135446253-135446253 on Build GRCh38
rs1030334204 Chr.5: 135446148-135446148 on Build GRCh38
rs771910221 Chr.5: 135446401-135446401 on Build GRCh38
rs376103324 Chr.5: 135446352-135446352 on Build GRCh38
rs954109373 Chr.5: 135445881-135445881 on Build GRCh38
rs370525727 Chr.5: 135446112-135446112 on Build GRCh38
rs768764449 Chr.5: 135446399-135446399 on Build GRCh38
rs747182587 Chr.5: 135446338-135446338 on Build GRCh38
rs746004472 Chr.5: 135446347-135446347 on Build GRCh38
rs905519199 Chr.5: 135445979-135445979 on Build GRCh38
rs371746306 Chr.5: 135446191-135446191 on Build GRCh38
rs761536447 Chr.5: 135446367-135446367 on Build GRCh38
rs769835476 Chr.5: 135446307-135446307 on Build GRCh38
rs780214750 Chr.5: 135446396-135446396 on Build GRCh38
rs370425129 Chr.5: 135446361-135446361 on Build GRCh38
rs115662828 Chr.5: 135446364-135446364 on Build GRCh38
rs768158734 Chr.5: 135446209-135446209 on Build GRCh38
rs751723252 Chr.5: 135445895-135445895 on Build GRCh38
rs772669054 Chr.5: 135446403-135446403 on Build GRCh38
rs918757039 Chr.5: 135446266-135446266 on Build GRCh38
rs566760598 Chr.5: 135446221-135446221 on Build GRCh38
rs759839686 Chr.5: 135445935-135445935 on Build GRCh38
rs17168357 Chr.5: 135446323-135446323 on Build GRCh38
rs150331492 Chr.5: 135445983-135445983 on Build GRCh38
rs567158390 Chr.5: 135446251-135446251 on Build GRCh38
rs937451621 Chr.5: 135446028-135446028 on Build GRCh38
rs146257733 Chr.5: 135446165-135446165 on Build GRCh38
rs149928130 Chr.5: 135446385-135446385 on Build GRCh38
rs370314333 Chr.5: 135446381-135446381 on Build GRCh38
rs760045869 Chr.5: 135446377-135446377 on Build GRCh38
rs778844390 Chr.5: 135446400-135446400 on Build GRCh38
rs768533105 Chr.5: 135446363-135446363 on Build GRCh38
rs189112976 Chr.5: 135445949-135445949 on Build GRCh38
rs1000516626 Chr.5: 135446158-135446158 on Build GRCh38
rs1020064397 Chr.5: 135446081-135446081 on Build GRCh38
rs751958565 Chr.5: 135446390-135446390 on Build GRCh38
rs937609714 Chr.5: 135446316-135446316 on Build GRCh38
rs147422825 Chr.5: 135446386-135446386 on Build GRCh38
rs184602640 Chr.5: 135445894-135445894 on Build GRCh38
rs918354792 Chr.5: 135445973-135445973 on Build GRCh38
rs867889944 Chr.5: 135445925-135445925 on Build GRCh38
rs760406830 Chr.5: 135446414-135446414 on Build GRCh38
rs578111576 Chr.5: 135445882-135445882 on Build GRCh38
rs564416603 Chr.5: 135446198-135446198 on Build GRCh38
rs771686783 Chr.5: 135446368-135446368 on Build GRCh38
rs752982081 Chr.5: 135445964-135445964 on Build GRCh38
rs141544376 Chr.5: 135445884-135445884 on Build GRCh38
rs139885262 Chr.5: 135446397-135446397 on Build GRCh38

View Full Product Details