Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26123237-26123759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H2BC
Entrez Gene ID
8347
Gene Name
histone cluster 1, H2bc
Gene Aliases
H2B.1, H2B/l, H2BFL, dJ221C16.3
UniGene
Hs.658713
Gene Chromosome Location
Chr.6: 26123467-26123904 on Build GRCh38
Transcripts
NM_003526.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5869890 c.214G>A p.E72K Chr.6: 26123691-26123691 on Build GRCh38
7428970 c.375_376insC p.K126fs*>2 Chr.6: 26123529-26123530 on Build GRCh38
451014 c.374C>T p.S125F Chr.6: 26123531-26123531 on Build GRCh38
6106131 c.331G>C p.A111P Chr.6: 26123574-26123574 on Build GRCh38
1621280 c.324C>G p.A108A Chr.6: 26123581-26123581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762631658 Chr.6: 26123627-26123627 on Build GRCh38
rs925605948 Chr.6: 26123310-26123310 on Build GRCh38
rs189878433 Chr.6: 26123430-26123430 on Build GRCh38
rs747654721 Chr.6: 26123699-26123699 on Build GRCh38
rs762001780 Chr.6: 26123501-26123501 on Build GRCh38
rs768236825 Chr.6: 26123687-26123687 on Build GRCh38
rs532539925 Chr.6: 26123289-26123289 on Build GRCh38
rs755866103 Chr.6: 26123602-26123602 on Build GRCh38
rs761241511 Chr.6: 26123629-26123629 on Build GRCh38
rs773052549 Chr.6: 26123633-26123633 on Build GRCh38
rs942484360 Chr.6: 26123672-26123672 on Build GRCh38
rs912788889 Chr.6: 26123400-26123400 on Build GRCh38
rs964232123 Chr.6: 26123491-26123491 on Build GRCh38
rs116104991 Chr.6: 26123404-26123404 on Build GRCh38
rs28365923 Chr.6: 26123421-26123421 on Build GRCh38
rs780153724 Chr.6: 26123520-26123520 on Build GRCh38
rs981853844 Chr.6: 26123692-26123692 on Build GRCh38
rs376391280 Chr.6: 26123575-26123575 on Build GRCh38
rs775702965 Chr.6: 26123478-26123478 on Build GRCh38
rs776118532 Chr.6: 26123496-26123496 on Build GRCh38
rs145816298 Chr.6: 26123626-26123626 on Build GRCh38
rs994450319 Chr.6: 26123386-26123386 on Build GRCh38
rs768148798 Chr.6: 26123628-26123628 on Build GRCh38
rs763981422 Chr.6: 26123500-26123500 on Build GRCh38
rs1033141669 Chr.6: 26123607-26123607 on Build GRCh38
rs561633884 Chr.6: 26123366-26123366 on Build GRCh38
rs773685186 Chr.6: 26123548-26123548 on Build GRCh38
rs1051182738 Chr.6: 26123338-26123338 on Build GRCh38
rs1009423988 Chr.6: 26123453-26123453 on Build GRCh38
rs198822 Chr.6: 26123311-26123311 on Build GRCh38
rs920230730 Chr.6: 26123502-26123502 on Build GRCh38
rs1002014735 Chr.6: 26123329-26123329 on Build GRCh38
rs776510779 Chr.6: 26123710-26123710 on Build GRCh38
rs967450808 Chr.6: 26123469-26123469 on Build GRCh38
rs767976076 Chr.6: 26123495-26123495 on Build GRCh38
rs139980610 Chr.6: 26123584-26123584 on Build GRCh38
rs570471579 Chr.6: 26123393-26123393 on Build GRCh38
rs1025900277 Chr.6: 26123539-26123539 on Build GRCh38
rs994544053 Chr.6: 26123530-26123530 on Build GRCh38
rs762227039 Chr.6: 26123494-26123494 on Build GRCh38
rs905123367 Chr.6: 26123325-26123325 on Build GRCh38
rs760964813 Chr.6: 26123551-26123551 on Build GRCh38
rs908309877 Chr.6: 26123416-26123416 on Build GRCh38
rs765519959 Chr.6: 26123738-26123738 on Build GRCh38
rs1020405449 Chr.6: 26123353-26123353 on Build GRCh38
rs374653821 Chr.6: 26123483-26123483 on Build GRCh38
rs775000741 Chr.6: 26123486-26123486 on Build GRCh38
rs952974062 Chr.6: 26123517-26123517 on Build GRCh38
rs766828029 Chr.6: 26123664-26123664 on Build GRCh38
rs773177860 Chr.6: 26123706-26123706 on Build GRCh38
rs896772556 Chr.6: 26123572-26123572 on Build GRCh38
rs942786595 Chr.6: 26123518-26123518 on Build GRCh38
rs957062398 Chr.6: 26123610-26123610 on Build GRCh38
rs762403828 Chr.6: 26123577-26123577 on Build GRCh38
rs757608021 Chr.6: 26123668-26123668 on Build GRCh38
rs935635998 Chr.6: 26123471-26123471 on Build GRCh38
rs59995466 Chr.6: 26123319-26123319 on Build GRCh38
rs772573839 Chr.6: 26123620-26123620 on Build GRCh38
rs918787389 Chr.6: 26123261-26123261 on Build GRCh38
rs549225831 Chr.6: 26123420-26123420 on Build GRCh38
rs1016976039 Chr.6: 26123381-26123381 on Build GRCh38
rs569932521 Chr.6: 26123601-26123601 on Build GRCh38
rs891638991 Chr.6: 26123333-26123333 on Build GRCh38
rs749798602 Chr.6: 26123480-26123480 on Build GRCh38
rs763639919 Chr.6: 26123582-26123582 on Build GRCh38
rs765570979 Chr.6: 26123659-26123659 on Build GRCh38
rs563473258 Chr.6: 26123481-26123481 on Build GRCh38
rs781077239 Chr.6: 26123513-26123513 on Build GRCh38
rs778150476 Chr.6: 26123621-26123621 on Build GRCh38
rs889612538 Chr.6: 26123509-26123509 on Build GRCh38
rs769425574 Chr.6: 26123566-26123566 on Build GRCh38
rs948395242 Chr.6: 26123302-26123302 on Build GRCh38
rs765632001 Chr.6: 26123563-26123563 on Build GRCh38
rs180677864 Chr.6: 26123438-26123438 on Build GRCh38
rs764641345 Chr.6: 26123498-26123498 on Build GRCh38
rs181595196 Chr.6: 26123429-26123429 on Build GRCh38
rs146690970 Chr.6: 26123731-26123731 on Build GRCh38
rs957441405 Chr.6: 26123337-26123337 on Build GRCh38
rs115740542 Chr.6: 26123274-26123274 on Build GRCh38
rs898472099 Chr.6: 26123385-26123385 on Build GRCh38
rs748922417 Chr.6: 26123683-26123683 on Build GRCh38
rs758883693 Chr.6: 26123614-26123614 on Build GRCh38
rs527319274 Chr.6: 26123373-26123373 on Build GRCh38
rs933782821 Chr.6: 26123335-26123335 on Build GRCh38
rs548119883 Chr.6: 26123330-26123330 on Build GRCh38
rs748652014 Chr.6: 26123617-26123617 on Build GRCh38
rs992606749 Chr.6: 26123279-26123279 on Build GRCh38
rs200921073 Chr.6: 26123541-26123541 on Build GRCh38
rs549941215 Chr.6: 26123709-26123709 on Build GRCh38
rs1039864431 Chr.6: 26123278-26123278 on Build GRCh38
rs778296122 Chr.6: 26123661-26123661 on Build GRCh38
rs750602400 Chr.6: 26123514-26123514 on Build GRCh38
rs879250962 Chr.6: 26123656-26123656 on Build GRCh38
rs772320204 Chr.6: 26123545-26123545 on Build GRCh38
rs545608798 Chr.6: 26123277-26123277 on Build GRCh38
rs1038473209 Chr.6: 26123678-26123678 on Build GRCh38
rs747363195 Chr.6: 26123623-26123623 on Build GRCh38
rs754134252 Chr.6: 26123613-26123613 on Build GRCh38
rs547345212 Chr.6: 26123384-26123384 on Build GRCh38
rs1024879357 Chr.6: 26123322-26123322 on Build GRCh38
rs539481947 Chr.6: 26123398-26123398 on Build GRCh38
rs931533278 Chr.6: 26123412-26123412 on Build GRCh38
rs557027706 Chr.6: 26123463-26123463 on Build GRCh38
rs370776308 Chr.6: 26123734-26123734 on Build GRCh38
rs773995989 Chr.6: 26123689-26123689 on Build GRCh38
rs893626140 Chr.6: 26123452-26123452 on Build GRCh38
rs754109959 Chr.6: 26123651-26123651 on Build GRCh38
rs996881212 Chr.6: 26123650-26123650 on Build GRCh38
rs752759673 Chr.6: 26123660-26123660 on Build GRCh38
rs1023779787 Chr.6: 26123586-26123586 on Build GRCh38
rs185059741 Chr.6: 26123515-26123515 on Build GRCh38
rs979770848 Chr.6: 26123305-26123305 on Build GRCh38
rs748547124 Chr.6: 26123663-26123670 on Build GRCh38
rs879122516 Chr.6: 26123676-26123676 on Build GRCh38
rs1019691643 Chr.6: 26123632-26123632 on Build GRCh38
rs889914875 Chr.6: 26123342-26123342 on Build GRCh38
rs779542235 Chr.6: 26123527-26123527 on Build GRCh38
rs371600527 Chr.6: 26123653-26123653 on Build GRCh38
rs372665363 Chr.6: 26123510-26123510 on Build GRCh38
rs749607225 Chr.6: 26123726-26123726 on Build GRCh38
rs766317808 Chr.6: 26123647-26123647 on Build GRCh38
rs1023748742 Chr.6: 26123414-26123414 on Build GRCh38
rs781600734 Chr.6: 26123681-26123681 on Build GRCh38
rs189531424 Chr.6: 26123428-26123428 on Build GRCh38
rs140650086 Chr.6: 26123611-26123611 on Build GRCh38
rs770745106 Chr.6: 26123387-26123387 on Build GRCh38
rs1020771752 Chr.6: 26123462-26123462 on Build GRCh38
rs149841603 Chr.6: 26123590-26123590 on Build GRCh38
rs769727333 Chr.6: 26123473-26123473 on Build GRCh38
rs386698215 Chr.6: 26123645-26123646 on Build GRCh38
rs543558651 Chr.6: 26123456-26123456 on Build GRCh38
rs368906030 Chr.6: 26123485-26123485 on Build GRCh38
rs986699042 Chr.6: 26123619-26123619 on Build GRCh38
rs926382331 Chr.6: 26123695-26123695 on Build GRCh38
rs760597993 Chr.6: 26123707-26123707 on Build GRCh38
rs759299723 Chr.6: 26123497-26123497 on Build GRCh38
rs566810063 Chr.6: 26123315-26123315 on Build GRCh38
rs1054092714 Chr.6: 26123599-26123599 on Build GRCh38
rs779527827 Chr.6: 26123288-26123291 on Build GRCh38
rs748707089 Chr.6: 26123533-26123533 on Build GRCh38
rs198821 Chr.6: 26123401-26123401 on Build GRCh38
rs771502786 Chr.6: 26123701-26123701 on Build GRCh38
rs749312462 Chr.6: 26123521-26123521 on Build GRCh38
rs779463146 Chr.6: 26123503-26123517 on Build GRCh38
rs146880453 Chr.6: 26123593-26123593 on Build GRCh38
rs760468183 Chr.6: 26123644-26123644 on Build GRCh38
rs752887874 Chr.6: 26123740-26123740 on Build GRCh38
rs368414402 Chr.6: 26123523-26123523 on Build GRCh38
rs767421864 Chr.6: 26123631-26123631 on Build GRCh38
rs537609928 Chr.6: 26123422-26123422 on Build GRCh38
rs199919825 Chr.6: 26123598-26123598 on Build GRCh38
rs920097145 Chr.6: 26123410-26123410 on Build GRCh38
rs886767367 Chr.6: 26123312-26123312 on Build GRCh38

View Full Product Details