Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140334270-140334869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC37A3
Entrez Gene ID
84255
Gene Name
solute carrier family 37 member 3
Gene Aliases
-
UniGene
Hs.446021
Gene Chromosome Location
Chr.7: 140333752-140404352 on Build GRCh38
Transcripts
NM_001287498.1 NM_032295.3 NM_207113.2 XM_011516626.2 XM_011516627.2 XM_017012712.1 XM_017012713.1 XM_017012714.1 XM_017012715.1 XM_017012716.1 XM_017012717.1

SNP Detail

SNP ID Chromosome Location
rs1004301765 Chr.7: 140334780-140334780 on Build GRCh38
rs777557567 Chr.7: 140334678-140334678 on Build GRCh38
rs570724649 Chr.7: 140334651-140334651 on Build GRCh38
rs34580240 Chr.7: 140334554-140334555 on Build GRCh38
rs945165891 Chr.7: 140334622-140334622 on Build GRCh38
rs912834114 Chr.7: 140334307-140334307 on Build GRCh38
rs73737572 Chr.7: 140334373-140334373 on Build GRCh38
rs988101610 Chr.7: 140334470-140334470 on Build GRCh38
rs1002131613 Chr.7: 140334493-140334493 on Build GRCh38
rs957737123 Chr.7: 140334608-140334608 on Build GRCh38
rs754124631 Chr.7: 140334567-140334567 on Build GRCh38
rs1028554096 Chr.7: 140334833-140334833 on Build GRCh38
rs1010021729 Chr.7: 140334338-140334338 on Build GRCh38
rs751415172 Chr.7: 140334610-140334610 on Build GRCh38
rs954436677 Chr.7: 140334586-140334586 on Build GRCh38
rs998409909 Chr.7: 140334786-140334786 on Build GRCh38
rs894462347 Chr.7: 140334634-140334634 on Build GRCh38
rs1033825577 Chr.7: 140334419-140334419 on Build GRCh38
rs765307298 Chr.7: 140334609-140334609 on Build GRCh38
rs368157037 Chr.7: 140334293-140334293 on Build GRCh38
rs1128589 Chr.7: 140334494-140334494 on Build GRCh38
rs192560001 Chr.7: 140334628-140334628 on Build GRCh38
rs529748572 Chr.7: 140334558-140334558 on Build GRCh38
rs922951665 Chr.7: 140334585-140334585 on Build GRCh38
rs190768835 Chr.7: 140334317-140334317 on Build GRCh38
rs887166862 Chr.7: 140334700-140334700 on Build GRCh38
rs1015830706 Chr.7: 140334704-140334704 on Build GRCh38
rs576353642 Chr.7: 140334483-140334483 on Build GRCh38
rs754810057 Chr.7: 140334580-140334580 on Build GRCh38
rs115617434 Chr.7: 140334798-140334798 on Build GRCh38
rs1019797137 Chr.7: 140334591-140334591 on Build GRCh38
rs1055086716 Chr.7: 140334633-140334633 on Build GRCh38
rs920931435 Chr.7: 140334354-140334354 on Build GRCh38
rs894514543 Chr.7: 140334507-140334507 on Build GRCh38
rs975644450 Chr.7: 140334605-140334605 on Build GRCh38
rs546463337 Chr.7: 140334466-140334466 on Build GRCh38
rs957686708 Chr.7: 140334839-140334839 on Build GRCh38
rs995588285 Chr.7: 140334547-140334547 on Build GRCh38
rs1006174483 Chr.7: 140334688-140334688 on Build GRCh38
rs767729208 Chr.7: 140334391-140334391 on Build GRCh38
rs760090206 Chr.7: 140334363-140334363 on Build GRCh38
rs538143489 Chr.7: 140334691-140334691 on Build GRCh38
rs971728962 Chr.7: 140334583-140334583 on Build GRCh38
rs575623558 Chr.7: 140334454-140334454 on Build GRCh38
rs114389520 Chr.7: 140334606-140334606 on Build GRCh38
rs150834422 Chr.7: 140334349-140334349 on Build GRCh38
rs1055225454 Chr.7: 140334639-140334639 on Build GRCh38
rs747947354 Chr.7: 140334632-140334632 on Build GRCh38
rs183296039 Chr.7: 140334472-140334472 on Build GRCh38
rs187776590 Chr.7: 140334627-140334627 on Build GRCh38
rs1046244443 Chr.7: 140334469-140334469 on Build GRCh38
rs868044419 Chr.7: 140334435-140334435 on Build GRCh38
rs140989996 Chr.7: 140334370-140334370 on Build GRCh38
rs985844118 Chr.7: 140334598-140334598 on Build GRCh38
rs867650820 Chr.7: 140334804-140334804 on Build GRCh38
rs77864406 Chr.7: 140334487-140334487 on Build GRCh38
rs552172083 Chr.7: 140334650-140334650 on Build GRCh38
rs572390678 Chr.7: 140334464-140334464 on Build GRCh38
rs936559835 Chr.7: 140334314-140334314 on Build GRCh38
rs10244212 Chr.7: 140334822-140334822 on Build GRCh38
rs965668775 Chr.7: 140334313-140334313 on Build GRCh38
rs865826608 Chr.7: 140334497-140334497 on Build GRCh38
rs887270458 Chr.7: 140334561-140334561 on Build GRCh38
rs937918696 Chr.7: 140334631-140334631 on Build GRCh38
rs892458021 Chr.7: 140334332-140334332 on Build GRCh38
rs965974555 Chr.7: 140334600-140334600 on Build GRCh38
rs748958564 Chr.7: 140334797-140334797 on Build GRCh38
rs73472731 Chr.7: 140334428-140334428 on Build GRCh38
rs1048438702 Chr.7: 140334665-140334665 on Build GRCh38
rs961899980 Chr.7: 140334705-140334705 on Build GRCh38

View Full Product Details