Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 67062047-67062598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS5
Entrez Gene ID
10987
Gene Name
COP9 signalosome subunit 5
Gene Aliases
CSN5, JAB1, MOV-34, SGN5
UniGene
Hs.491912
Gene Chromosome Location
Chr.8: 67043079-67062327 on Build GRCh38
Transcripts
NM_006837.2 XM_005251305.3 XM_006716474.2 XM_006716477.2 XM_011517598.1 XM_011517599.1 XM_011517600.1 XM_011517601.1 XM_011517602.1 XM_011517607.1 XM_017013847.1 XM_017013848.1 XM_017013849.1 XM_017013850.1 XM_017013852.1 XM_017013854.1 XM_017013855.1

Gene Symbol
CSPP1
Entrez Gene ID
79848
Gene Name
centrosome and spindle pole associated protein 1
Gene Aliases
CSPP, JBTS21
UniGene
Hs.370147
Gene Chromosome Location
Chr.8: 67062417-67196614 on Build GRCh38
Transcripts
NM_006837.2 XM_005251305.3 XM_006716474.2 XM_006716477.2 XM_011517598.1 XM_011517599.1 XM_011517600.1 XM_011517601.1 XM_011517602.1 XM_011517607.1 XM_017013847.1 XM_017013848.1 XM_017013849.1 XM_017013850.1 XM_017013852.1 XM_017013854.1 XM_017013855.1

SNP Detail

SNP ID Chromosome Location
rs997495668 Chr.8: 67062199-67062199 on Build GRCh38
rs762158187 Chr.8: 67062066-67062066 on Build GRCh38
rs777127654 Chr.8: 67062111-67062111 on Build GRCh38
rs1026322536 Chr.8: 67062207-67062207 on Build GRCh38
rs748293612 Chr.8: 67062186-67062186 on Build GRCh38
rs539795114 Chr.8: 67062465-67062466 on Build GRCh38
rs763335511 Chr.8: 67062078-67062078 on Build GRCh38
rs763449931 Chr.8: 67062268-67062271 on Build GRCh38
rs560728613 Chr.8: 67062548-67062548 on Build GRCh38
rs187713414 Chr.8: 67062180-67062180 on Build GRCh38
rs914182963 Chr.8: 67062228-67062228 on Build GRCh38
rs748155189 Chr.8: 67062214-67062214 on Build GRCh38
rs944189370 Chr.8: 67062331-67062331 on Build GRCh38
rs766867960 Chr.8: 67062086-67062086 on Build GRCh38
rs1055026523 Chr.8: 67062103-67062103 on Build GRCh38
rs745817917 Chr.8: 67062144-67062144 on Build GRCh38
rs371438535 Chr.8: 67062292-67062292 on Build GRCh38
rs779202205 Chr.8: 67062105-67062106 on Build GRCh38
rs780256588 Chr.8: 67062492-67062492 on Build GRCh38
rs775479530 Chr.8: 67062178-67062178 on Build GRCh38
rs752081809 Chr.8: 67062091-67062091 on Build GRCh38
rs748871311 Chr.8: 67062112-67062112 on Build GRCh38
rs1016507560 Chr.8: 67062291-67062294 on Build GRCh38
rs990960501 Chr.8: 67062317-67062317 on Build GRCh38
rs576072707 Chr.8: 67062453-67062453 on Build GRCh38
rs968762924 Chr.8: 67062556-67062556 on Build GRCh38
rs977686478 Chr.8: 67062352-67062352 on Build GRCh38
rs754985151 Chr.8: 67062101-67062102 on Build GRCh38
rs1002695477 Chr.8: 67062451-67062451 on Build GRCh38
rs756299636 Chr.8: 67062194-67062196 on Build GRCh38
rs370993239 Chr.8: 67062470-67062471 on Build GRCh38
rs934913781 Chr.8: 67062359-67062359 on Build GRCh38
rs970468501 Chr.8: 67062346-67062346 on Build GRCh38
rs145880569 Chr.8: 67062485-67062485 on Build GRCh38
rs759727821 Chr.8: 67062477-67062477 on Build GRCh38
rs901942012 Chr.8: 67062189-67062189 on Build GRCh38
rs1024764745 Chr.8: 67062526-67062526 on Build GRCh38
rs536329763 Chr.8: 67062380-67062380 on Build GRCh38
rs767381640 Chr.8: 67062088-67062089 on Build GRCh38
rs756751737 Chr.8: 67062120-67062120 on Build GRCh38
rs750426531 Chr.8: 67062091-67062092 on Build GRCh38
rs1047751882 Chr.8: 67062435-67062435 on Build GRCh38
rs568715812 Chr.8: 67062376-67062376 on Build GRCh38
rs1012056627 Chr.8: 67062523-67062523 on Build GRCh38
rs1013047219 Chr.8: 67062514-67062514 on Build GRCh38
rs958446395 Chr.8: 67062509-67062509 on Build GRCh38
rs376148887 Chr.8: 67062158-67062158 on Build GRCh38
rs558500596 Chr.8: 67062310-67062310 on Build GRCh38
rs765215722 Chr.8: 67062069-67062069 on Build GRCh38
rs888413157 Chr.8: 67062382-67062382 on Build GRCh38
rs1039629361 Chr.8: 67062406-67062406 on Build GRCh38
rs897685703 Chr.8: 67062446-67062446 on Build GRCh38
rs1021303724 Chr.8: 67062520-67062520 on Build GRCh38
rs755277044 Chr.8: 67062092-67062092 on Build GRCh38
rs771984240 Chr.8: 67062165-67062165 on Build GRCh38
rs532388929 Chr.8: 67062126-67062126 on Build GRCh38
rs548484282 Chr.8: 67062355-67062355 on Build GRCh38
rs767873086 Chr.8: 67062108-67062108 on Build GRCh38
rs1006152836 Chr.8: 67062408-67062408 on Build GRCh38
rs541364145 Chr.8: 67062517-67062517 on Build GRCh38
rs950578341 Chr.8: 67062216-67062216 on Build GRCh38
rs934923704 Chr.8: 67062102-67062102 on Build GRCh38
rs969004862 Chr.8: 67062258-67062258 on Build GRCh38
rs192344550 Chr.8: 67062433-67062433 on Build GRCh38
rs572611728 Chr.8: 67062068-67062068 on Build GRCh38
rs772889556 Chr.8: 67062373-67062373 on Build GRCh38
rs563329721 Chr.8: 67062110-67062110 on Build GRCh38
rs183016088 Chr.8: 67062500-67062500 on Build GRCh38
rs761774214 Chr.8: 67062087-67062088 on Build GRCh38
rs1041138452 Chr.8: 67062340-67062340 on Build GRCh38
rs543335947 Chr.8: 67062428-67062428 on Build GRCh38
rs774608446 Chr.8: 67062443-67062443 on Build GRCh38
rs572242203 Chr.8: 67062503-67062503 on Build GRCh38

View Full Product Details