Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38992086-38992600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3B
Entrez Gene ID
9582
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3B
Gene Aliases
A3B, APOBEC1L, ARCD3, ARP4, DJ742C19.2, PHRBNL, bK150C2.2
UniGene
Hs.226307
Gene Chromosome Location
Chr.22: 38982399-38992779 on Build GRCh38
Transcripts
NM_001270411.1 NM_004900.4 NR_104187.1

Gene Symbol
APOBEC3B-AS1
Entrez Gene ID
100874530
Gene Name
APOBEC3B antisense RNA 1
Gene Aliases
-
UniGene
Hs.626951
Gene Chromosome Location
Chr.22: 38991559-38998220 on Build GRCh38
Transcripts
NM_001270411.1 NM_004900.4 NR_104187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1034213 c.1120C>T p.R374W Chr.22: 38992135-38992135 on Build GRCh38
1416299 c.1121G>A p.R374Q Chr.22: 38992136-38992136 on Build GRCh38
4794928 c.1115G>A p.R372K Chr.22: 38992130-38992130 on Build GRCh38
6587146 c.1110T>A p.S370R Chr.22: 38992125-38992125 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770971928 Chr.22: 38992258-38992258 on Build GRCh38
rs1029833686 Chr.22: 38992327-38992327 on Build GRCh38
rs199837717 Chr.22: 38992130-38992130 on Build GRCh38
rs147717094 Chr.22: 38992132-38992132 on Build GRCh38
rs781129393 Chr.22: 38992135-38992135 on Build GRCh38
rs762284983 Chr.22: 38992408-38992408 on Build GRCh38
rs369092318 Chr.22: 38992112-38992112 on Build GRCh38
rs749808743 Chr.22: 38992524-38992524 on Build GRCh38
rs759754257 Chr.22: 38992169-38992169 on Build GRCh38
rs111603647 Chr.22: 38992461-38992461 on Build GRCh38
rs34383929 Chr.22: 38992125-38992126 on Build GRCh38
rs544458527 Chr.22: 38992404-38992404 on Build GRCh38
rs778831011 Chr.22: 38992262-38992262 on Build GRCh38
rs558255728 Chr.22: 38992319-38992319 on Build GRCh38
rs376840843 Chr.22: 38992180-38992180 on Build GRCh38
rs752341655 Chr.22: 38992355-38992355 on Build GRCh38
rs5757414 Chr.22: 38992211-38992211 on Build GRCh38
rs765789641 Chr.22: 38992187-38992187 on Build GRCh38
rs754388978 Chr.22: 38992116-38992116 on Build GRCh38
rs770139814 Chr.22: 38992512-38992512 on Build GRCh38
rs757039018 Chr.22: 38992462-38992462 on Build GRCh38
rs761800683 Chr.22: 38992136-38992141 on Build GRCh38
rs186931724 Chr.22: 38992562-38992562 on Build GRCh38
rs767143186 Chr.22: 38992353-38992353 on Build GRCh38
rs754228071 Chr.22: 38992183-38992183 on Build GRCh38
rs777426702 Chr.22: 38992367-38992367 on Build GRCh38
rs778864263 Chr.22: 38992392-38992392 on Build GRCh38
rs747131466 Chr.22: 38992487-38992487 on Build GRCh38
rs755907840 Chr.22: 38992210-38992210 on Build GRCh38
rs767230480 Chr.22: 38992143-38992144 on Build GRCh38
rs767770397 Chr.22: 38992171-38992171 on Build GRCh38
rs781424422 Chr.22: 38992493-38992493 on Build GRCh38
rs1025490709 Chr.22: 38992254-38992254 on Build GRCh38
rs769830596 Chr.22: 38992137-38992137 on Build GRCh38
rs760888672 Chr.22: 38992278-38992278 on Build GRCh38
rs764548809 Chr.22: 38992181-38992181 on Build GRCh38
rs750300713 Chr.22: 38992479-38992479 on Build GRCh38
rs747995529 Chr.22: 38992136-38992136 on Build GRCh38
rs781039331 Chr.22: 38992201-38992201 on Build GRCh38
rs759670284 Chr.22: 38992243-38992243 on Build GRCh38
rs751147075 Chr.22: 38992193-38992193 on Build GRCh38
rs775647396 Chr.22: 38992268-38992268 on Build GRCh38
rs757803839 Chr.22: 38992126-38992126 on Build GRCh38
rs763859391 Chr.22: 38992449-38992449 on Build GRCh38
rs780099788 Chr.22: 38992485-38992485 on Build GRCh38
rs772985111 Chr.22: 38992141-38992141 on Build GRCh38
rs748348695 Chr.22: 38992503-38992503 on Build GRCh38
rs34960225 Chr.22: 38992188-38992189 on Build GRCh38
rs756585285 Chr.22: 38992111-38992111 on Build GRCh38
rs745999319 Chr.22: 38992267-38992267 on Build GRCh38
rs753130435 Chr.22: 38992108-38992108 on Build GRCh38
rs202013249 Chr.22: 38992444-38992444 on Build GRCh38
rs762222315 Chr.22: 38992324-38992324 on Build GRCh38
rs755854444 Chr.22: 38992363-38992363 on Build GRCh38
rs753712055 Chr.22: 38992377-38992377 on Build GRCh38
rs994418281 Chr.22: 38992243-38992243 on Build GRCh38
rs878895525 Chr.22: 38992517-38992517 on Build GRCh38
rs753458291 Chr.22: 38992454-38992454 on Build GRCh38
rs768841404 Chr.22: 38992302-38992302 on Build GRCh38
rs368755246 Chr.22: 38992403-38992403 on Build GRCh38
rs142527797 Chr.22: 38992200-38992200 on Build GRCh38
rs1029391683 Chr.22: 38992541-38992541 on Build GRCh38
rs770074224 Chr.22: 38992415-38992415 on Build GRCh38
rs377246003 Chr.22: 38992532-38992532 on Build GRCh38
rs773790412 Chr.22: 38992416-38992416 on Build GRCh38
rs539419274 Chr.22: 38992317-38992317 on Build GRCh38
rs375638163 Chr.22: 38992161-38992161 on Build GRCh38
rs562667270 Chr.22: 38992434-38992434 on Build GRCh38
rs780118261 Chr.22: 38992401-38992401 on Build GRCh38
rs755862935 Chr.22: 38992505-38992507 on Build GRCh38
rs746580811 Chr.22: 38992131-38992131 on Build GRCh38
rs757123760 Chr.22: 38992379-38992379 on Build GRCh38
rs1005422317 Chr.22: 38992494-38992494 on Build GRCh38
rs200130268 Chr.22: 38992158-38992158 on Build GRCh38
rs373909956 Chr.22: 38992176-38992176 on Build GRCh38
rs112565950 Chr.22: 38992172-38992172 on Build GRCh38
rs771606143 Chr.22: 38992531-38992531 on Build GRCh38
rs1017888373 Chr.22: 38992277-38992277 on Build GRCh38
rs765813520 Chr.22: 38992334-38992334 on Build GRCh38
rs745840583 Chr.22: 38992393-38992393 on Build GRCh38
rs755132520 Chr.22: 38992489-38992489 on Build GRCh38
rs767122998 Chr.22: 38992424-38992424 on Build GRCh38
rs576561916 Chr.22: 38992400-38992400 on Build GRCh38
rs542090684 Chr.22: 38992477-38992477 on Build GRCh38
rs773772191 Chr.22: 38992523-38992523 on Build GRCh38
rs563848168 Chr.22: 38992574-38992574 on Build GRCh38
rs572392139 Chr.22: 38992224-38992224 on Build GRCh38
rs878921978 Chr.22: 38992198-38992198 on Build GRCh38
rs150925968 Chr.22: 38992481-38992481 on Build GRCh38
rs551840824 Chr.22: 38992572-38992572 on Build GRCh38
rs749253938 Chr.22: 38992150-38992150 on Build GRCh38
rs201246417 Chr.22: 38992184-38992184 on Build GRCh38
rs964089396 Chr.22: 38992281-38992281 on Build GRCh38
rs375488552 Chr.22: 38992114-38992114 on Build GRCh38
rs759066851 Chr.22: 38992343-38992343 on Build GRCh38
rs5757413 Chr.22: 38992202-38992202 on Build GRCh38
rs751090823 Chr.22: 38992337-38992337 on Build GRCh38
rs750178560 Chr.22: 38992419-38992423 on Build GRCh38
rs888343571 Chr.22: 38992259-38992259 on Build GRCh38
rs760188006 Chr.22: 38992536-38992536 on Build GRCh38

View Full Product Details