Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33285935-33286493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB1
Entrez Gene ID
92747
Gene Name
BPI fold containing family B member 1
Gene Aliases
C20orf114, LPLUNC1
UniGene
Hs.65551
Gene Chromosome Location
Chr.20: 33283135-33309878 on Build GRCh38
Transcripts
NM_033197.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4396346 c.10C>T p.P4S Chr.20: 33286083-33286083 on Build GRCh38
724032 c.23C>G p.T8S Chr.20: 33286096-33286096 on Build GRCh38
4752186 c.7G>T p.G3C Chr.20: 33286080-33286080 on Build GRCh38
7254942 c.50C>T p.T17I Chr.20: 33286123-33286123 on Build GRCh38
1615466 c.65C>A p.T22N Chr.20: 33286138-33286138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987936141 Chr.20: 33286367-33286367 on Build GRCh38
rs753968938 Chr.20: 33286216-33286216 on Build GRCh38
rs997082098 Chr.20: 33286000-33286000 on Build GRCh38
rs376586352 Chr.20: 33286206-33286206 on Build GRCh38
rs943801093 Chr.20: 33286373-33286373 on Build GRCh38
rs769324424 Chr.20: 33286183-33286183 on Build GRCh38
rs780918876 Chr.20: 33286177-33286177 on Build GRCh38
rs1027279856 Chr.20: 33286195-33286195 on Build GRCh38
rs371071403 Chr.20: 33286088-33286088 on Build GRCh38
rs140181326 Chr.20: 33286188-33286188 on Build GRCh38
rs767734834 Chr.20: 33286067-33286067 on Build GRCh38
rs760482226 Chr.20: 33286083-33286083 on Build GRCh38
rs1048503270 Chr.20: 33286449-33286449 on Build GRCh38
rs758158036 Chr.20: 33286231-33286231 on Build GRCh38
rs979534907 Chr.20: 33285971-33285971 on Build GRCh38
rs922352724 Chr.20: 33286326-33286326 on Build GRCh38
rs925257416 Chr.20: 33286317-33286317 on Build GRCh38
rs753769903 Chr.20: 33286084-33286084 on Build GRCh38
rs929804885 Chr.20: 33286458-33286458 on Build GRCh38
rs370873212 Chr.20: 33286147-33286147 on Build GRCh38
rs766101557 Chr.20: 33286166-33286166 on Build GRCh38
rs777435344 Chr.20: 33286099-33286099 on Build GRCh38
rs576532539 Chr.20: 33286242-33286242 on Build GRCh38
rs1056578045 Chr.20: 33286138-33286138 on Build GRCh38
rs534385884 Chr.20: 33286438-33286438 on Build GRCh38
rs970107436 Chr.20: 33286282-33286282 on Build GRCh38
rs1052210841 Chr.20: 33286149-33286149 on Build GRCh38
rs373309112 Chr.20: 33286205-33286205 on Build GRCh38
rs955786172 Chr.20: 33286013-33286013 on Build GRCh38
rs551241629 Chr.20: 33286059-33286059 on Build GRCh38
rs564560020 Chr.20: 33285959-33285959 on Build GRCh38
rs374464078 Chr.20: 33285974-33285974 on Build GRCh38
rs3827028 Chr.20: 33286451-33286451 on Build GRCh38
rs746658596 Chr.20: 33286163-33286163 on Build GRCh38
rs374492385 Chr.20: 33286033-33286033 on Build GRCh38
rs758159591 Chr.20: 33286089-33286089 on Build GRCh38
rs34388907 Chr.20: 33286085-33286085 on Build GRCh38
rs373622002 Chr.20: 33286167-33286167 on Build GRCh38
rs535791914 Chr.20: 33286051-33286051 on Build GRCh38
rs770374449 Chr.20: 33286045-33286045 on Build GRCh38
rs3746393 Chr.20: 33286015-33286015 on Build GRCh38
rs746487883 Chr.20: 33286044-33286044 on Build GRCh38
rs896584324 Chr.20: 33286141-33286141 on Build GRCh38
rs772566676 Chr.20: 33286039-33286039 on Build GRCh38
rs915890164 Chr.20: 33286060-33286060 on Build GRCh38
rs952935950 Chr.20: 33286016-33286016 on Build GRCh38
rs200987598 Chr.20: 33286234-33286234 on Build GRCh38
rs910930628 Chr.20: 33286428-33286428 on Build GRCh38
rs941262291 Chr.20: 33286046-33286046 on Build GRCh38
rs538711984 Chr.20: 33286323-33286323 on Build GRCh38
rs759457236 Chr.20: 33286248-33286248 on Build GRCh38
rs892222447 Chr.20: 33286162-33286162 on Build GRCh38
rs571625790 Chr.20: 33286276-33286276 on Build GRCh38
rs913711804 Chr.20: 33286368-33286368 on Build GRCh38
rs370824180 Chr.20: 33286212-33286212 on Build GRCh38
rs759439787 Chr.20: 33286225-33286225 on Build GRCh38
rs957913980 Chr.20: 33286347-33286347 on Build GRCh38
rs1040826052 Chr.20: 33286401-33286401 on Build GRCh38
rs368557203 Chr.20: 33286069-33286069 on Build GRCh38
rs761907533 Chr.20: 33286065-33286065 on Build GRCh38
rs370908525 Chr.20: 33286027-33286027 on Build GRCh38
rs549804910 Chr.20: 33286058-33286058 on Build GRCh38
rs768796667 Chr.20: 33285970-33285970 on Build GRCh38
rs749788143 Chr.20: 33286179-33286179 on Build GRCh38
rs1056712414 Chr.20: 33286461-33286461 on Build GRCh38
rs139597118 Chr.20: 33286079-33286079 on Build GRCh38
rs992898946 Chr.20: 33286262-33286262 on Build GRCh38
rs988948531 Chr.20: 33286307-33286307 on Build GRCh38
rs751222675 Chr.20: 33286235-33286235 on Build GRCh38
rs992832329 Chr.20: 33286052-33286052 on Build GRCh38
rs146312989 Chr.20: 33285960-33285960 on Build GRCh38
rs900084515 Chr.20: 33285996-33285996 on Build GRCh38
rs373034089 Chr.20: 33286232-33286232 on Build GRCh38
rs765119449 Chr.20: 33286227-33286227 on Build GRCh38
rs979166610 Chr.20: 33286283-33286283 on Build GRCh38
rs921013874 Chr.20: 33286429-33286429 on Build GRCh38
rs756921259 Chr.20: 33286240-33286240 on Build GRCh38
rs936547140 Chr.20: 33286098-33286098 on Build GRCh38
rs756160952 Chr.20: 33286080-33286080 on Build GRCh38
rs375302420 Chr.20: 33286229-33286229 on Build GRCh38
rs372545262 Chr.20: 33286038-33286038 on Build GRCh38
rs762184843 Chr.20: 33286189-33286189 on Build GRCh38
rs763561469 Chr.20: 33286057-33286057 on Build GRCh38
rs960130680 Chr.20: 33286258-33286258 on Build GRCh38

View Full Product Details