Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 94528128-94528709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASD1
Entrez Gene ID
64921
Gene Name
CAS1 domain containing 1
Gene Aliases
C7orf12, NBLA04196
UniGene
Hs.260041
Gene Chromosome Location
Chr.7: 94509858-94633465 on Build GRCh38
Transcripts
NM_022900.4 XM_006716092.3 XM_006716093.3 XM_011516495.2 XM_017012546.1 XM_017012547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6582711 c.424G>A p.G142S Chr.7: 94528215-94528215 on Build GRCh38
6270976 c.440G>T p.C147F Chr.7: 94528231-94528231 on Build GRCh38
6404594 c.436C>A p.Q146K Chr.7: 94528227-94528227 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs964943741 Chr.7: 94528217-94528217 on Build GRCh38
rs200497452 Chr.7: 94528281-94528281 on Build GRCh38
rs752802503 Chr.7: 94528261-94528261 on Build GRCh38
rs757653085 Chr.7: 94528471-94528471 on Build GRCh38
rs917591889 Chr.7: 94528456-94528456 on Build GRCh38
rs956835486 Chr.7: 94528670-94528670 on Build GRCh38
rs764101241 Chr.7: 94528274-94528276 on Build GRCh38
rs1018911598 Chr.7: 94528398-94528398 on Build GRCh38
rs1016784705 Chr.7: 94528441-94528441 on Build GRCh38
rs544625159 Chr.7: 94528275-94528276 on Build GRCh38
rs904000019 Chr.7: 94528338-94528338 on Build GRCh38
rs751613754 Chr.7: 94528285-94528310 on Build GRCh38
rs180960704 Chr.7: 94528616-94528616 on Build GRCh38
rs558785260 Chr.7: 94528488-94528488 on Build GRCh38
rs553131283 Chr.7: 94528433-94528433 on Build GRCh38
rs759885071 Chr.7: 94528273-94528273 on Build GRCh38
rs373498217 Chr.7: 94528416-94528416 on Build GRCh38
rs201576828 Chr.7: 94528205-94528205 on Build GRCh38
rs966509916 Chr.7: 94528598-94528598 on Build GRCh38
rs912894882 Chr.7: 94528566-94528566 on Build GRCh38
rs199658239 Chr.7: 94528287-94528287 on Build GRCh38
rs398005478 Chr.7: 94528286-94528286 on Build GRCh38
rs113451184 Chr.7: 94528667-94528667 on Build GRCh38
rs766025090 Chr.7: 94528251-94528254 on Build GRCh38
rs1042825520 Chr.7: 94528332-94528332 on Build GRCh38
rs761585954 Chr.7: 94528235-94528235 on Build GRCh38
rs1055313607 Chr.7: 94528357-94528357 on Build GRCh38
rs974957215 Chr.7: 94528464-94528464 on Build GRCh38
rs762908815 Chr.7: 94528240-94528240 on Build GRCh38
rs978520696 Chr.7: 94528605-94528605 on Build GRCh38
rs150000138 Chr.7: 94528214-94528214 on Build GRCh38
rs749663816 Chr.7: 94528156-94528156 on Build GRCh38
rs1029403682 Chr.7: 94528619-94528619 on Build GRCh38
rs546766758 Chr.7: 94528263-94528263 on Build GRCh38
rs760597823 Chr.7: 94528518-94528518 on Build GRCh38
rs758842742 Chr.7: 94528273-94528274 on Build GRCh38
rs968376326 Chr.7: 94528577-94528577 on Build GRCh38
rs886908422 Chr.7: 94528564-94528564 on Build GRCh38
rs563087535 Chr.7: 94528663-94528665 on Build GRCh38
rs886474081 Chr.7: 94528423-94528423 on Build GRCh38
rs561355641 Chr.7: 94528274-94528275 on Build GRCh38
rs768325942 Chr.7: 94528204-94528204 on Build GRCh38
rs979633944 Chr.7: 94528422-94528422 on Build GRCh38
rs776135983 Chr.7: 94528291-94528291 on Build GRCh38
rs202044183 Chr.7: 94528286-94528286 on Build GRCh38
rs760150216 Chr.7: 94528184-94528184 on Build GRCh38
rs575539610 Chr.7: 94528553-94528553 on Build GRCh38
rs1013848974 Chr.7: 94528401-94528401 on Build GRCh38
rs368247868 Chr.7: 94528181-94528181 on Build GRCh38
rs778739901 Chr.7: 94528384-94528384 on Build GRCh38
rs560425276 Chr.7: 94528268-94528268 on Build GRCh38
rs572120623 Chr.7: 94528275-94528275 on Build GRCh38
rs1055663786 Chr.7: 94528682-94528682 on Build GRCh38
rs954592683 Chr.7: 94528520-94528520 on Build GRCh38
rs779377513 Chr.7: 94528271-94528271 on Build GRCh38
rs538864069 Chr.7: 94528484-94528484 on Build GRCh38
rs780467864 Chr.7: 94528282-94528282 on Build GRCh38
rs923210671 Chr.7: 94528273-94528273 on Build GRCh38
rs200785605 Chr.7: 94528293-94528293 on Build GRCh38
rs968329601 Chr.7: 94528647-94528647 on Build GRCh38
rs950842870 Chr.7: 94528360-94528360 on Build GRCh38
rs537409318 Chr.7: 94528348-94528348 on Build GRCh38
rs896119352 Chr.7: 94528272-94528272 on Build GRCh38
rs189320156 Chr.7: 94528403-94528403 on Build GRCh38
rs947769837 Chr.7: 94528481-94528481 on Build GRCh38
rs57770781 Chr.7: 94528274-94528274 on Build GRCh38
rs764958730 Chr.7: 94528236-94528236 on Build GRCh38
rs776913330 Chr.7: 94528260-94528261 on Build GRCh38
rs922233382 Chr.7: 94528404-94528404 on Build GRCh38
rs370364007 Chr.7: 94528265-94528265 on Build GRCh38
rs746237439 Chr.7: 94528280-94528280 on Build GRCh38
rs573074656 Chr.7: 94528446-94528446 on Build GRCh38
rs779433776 Chr.7: 94528172-94528172 on Build GRCh38
rs772731095 Chr.7: 94528162-94528166 on Build GRCh38
rs535524797 Chr.7: 94528646-94528646 on Build GRCh38
rs202237133 Chr.7: 94528179-94528179 on Build GRCh38
rs569188387 Chr.7: 94528328-94528328 on Build GRCh38
rs1018963728 Chr.7: 94528588-94528588 on Build GRCh38
rs1045110838 Chr.7: 94528521-94528521 on Build GRCh38
rs995381348 Chr.7: 94528550-94528550 on Build GRCh38
rs898448188 Chr.7: 94528545-94528545 on Build GRCh38
rs927005381 Chr.7: 94528632-94528632 on Build GRCh38
rs766251640 Chr.7: 94528245-94528245 on Build GRCh38
rs760277196 Chr.7: 94528228-94528229 on Build GRCh38
rs778178163 Chr.7: 94528155-94528155 on Build GRCh38
rs912906388 Chr.7: 94528326-94528326 on Build GRCh38
rs183579226 Chr.7: 94528395-94528395 on Build GRCh38
rs552469790 Chr.7: 94528311-94528311 on Build GRCh38

View Full Product Details