Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148865144-148865642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA3
Entrez Gene ID
1359
Gene Name
carboxypeptidase A3
Gene Aliases
MC-CPA
UniGene
Hs.646
Gene Chromosome Location
Chr.3: 148865256-148897091 on Build GRCh38
Transcripts
NM_001870.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7181626 c.40C>T p.L14F Chr.3: 148865347-148865347 on Build GRCh38
5612098 c.59G>T p.R20L Chr.3: 148865366-148865366 on Build GRCh38
7153149 c.83G>A p.R28H Chr.3: 148865487-148865487 on Build GRCh38
310296 c.49G>T p.A17S Chr.3: 148865356-148865356 on Build GRCh38
33023 c.100G>T p.E34* Chr.3: 148865504-148865504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77558349 Chr.3: 148865602-148865602 on Build GRCh38
rs1021999459 Chr.3: 148865450-148865450 on Build GRCh38
rs766746996 Chr.3: 148865369-148865369 on Build GRCh38
rs955579210 Chr.3: 148865448-148865448 on Build GRCh38
rs143377024 Chr.3: 148865326-148865326 on Build GRCh38
rs372797881 Chr.3: 148865469-148865469 on Build GRCh38
rs145785078 Chr.3: 148865508-148865508 on Build GRCh38
rs955865368 Chr.3: 148865401-148865401 on Build GRCh38
rs751830747 Chr.3: 148865318-148865318 on Build GRCh38
rs1031572739 Chr.3: 148865187-148865187 on Build GRCh38
rs752693946 Chr.3: 148865363-148865363 on Build GRCh38
rs779182356 Chr.3: 148865475-148865477 on Build GRCh38
rs143382113 Chr.3: 148865173-148865173 on Build GRCh38
rs561390621 Chr.3: 148865507-148865507 on Build GRCh38
rs758772748 Chr.3: 148865309-148865309 on Build GRCh38
rs200942451 Chr.3: 148865379-148865379 on Build GRCh38
rs770909885 Chr.3: 148865491-148865491 on Build GRCh38
rs766144929 Chr.3: 148865542-148865542 on Build GRCh38
rs879203021 Chr.3: 148865172-148865173 on Build GRCh38
rs1007670073 Chr.3: 148865382-148865382 on Build GRCh38
rs375410605 Chr.3: 148865522-148865522 on Build GRCh38
rs757971830 Chr.3: 148865396-148865396 on Build GRCh38
rs746775216 Chr.3: 148865351-148865351 on Build GRCh38
rs987063956 Chr.3: 148865455-148865455 on Build GRCh38
rs774345537 Chr.3: 148865496-148865496 on Build GRCh38
rs537389511 Chr.3: 148865307-148865307 on Build GRCh38
rs776985477 Chr.3: 148865361-148865361 on Build GRCh38
rs960757636 Chr.3: 148865490-148865490 on Build GRCh38
rs7635943 Chr.3: 148865377-148865377 on Build GRCh38
rs779670062 Chr.3: 148865476-148865476 on Build GRCh38
rs760095645 Chr.3: 148865445-148865445 on Build GRCh38
rs112769670 Chr.3: 148865455-148865455 on Build GRCh38
rs760635072 Chr.3: 148865240-148865240 on Build GRCh38
rs113598575 Chr.3: 148865184-148865184 on Build GRCh38
rs754158896 Chr.3: 148865283-148865283 on Build GRCh38
rs565544752 Chr.3: 148865259-148865259 on Build GRCh38
rs575412448 Chr.3: 148865462-148865462 on Build GRCh38
rs370509092 Chr.3: 148865364-148865364 on Build GRCh38
rs761469664 Chr.3: 148865345-148865345 on Build GRCh38
rs747006517 Chr.3: 148865399-148865399 on Build GRCh38
rs781309145 Chr.3: 148865487-148865487 on Build GRCh38
rs766913060 Chr.3: 148865315-148865315 on Build GRCh38
rs761007394 Chr.3: 148865606-148865606 on Build GRCh38
rs796927275 Chr.3: 148865445-148865445 on Build GRCh38
rs375313654 Chr.3: 148865393-148865393 on Build GRCh38
rs140527015 Chr.3: 148865488-148865488 on Build GRCh38
rs747959165 Chr.3: 148865425-148865425 on Build GRCh38
rs753680171 Chr.3: 148865282-148865282 on Build GRCh38
rs751125624 Chr.3: 148865479-148865479 on Build GRCh38
rs745400658 Chr.3: 148865341-148865341 on Build GRCh38
rs761950959 Chr.3: 148865290-148865290 on Build GRCh38
rs754567654 Chr.3: 148865547-148865547 on Build GRCh38
rs111765977 Chr.3: 148865365-148865365 on Build GRCh38
rs919938603 Chr.3: 148865516-148865516 on Build GRCh38
rs201203054 Chr.3: 148865395-148865395 on Build GRCh38
rs775093925 Chr.3: 148865276-148865276 on Build GRCh38
rs763969113 Chr.3: 148865277-148865277 on Build GRCh38
rs199499869 Chr.3: 148865350-148865350 on Build GRCh38
rs771842039 Chr.3: 148865347-148865347 on Build GRCh38
rs767938478 Chr.3: 148865386-148865386 on Build GRCh38
rs779126871 Chr.3: 148865598-148865598 on Build GRCh38
rs769866932 Chr.3: 148865427-148865427 on Build GRCh38
rs771124891 Chr.3: 148865587-148865587 on Build GRCh38
rs781244498 Chr.3: 148865563-148865563 on Build GRCh38
rs1038754481 Chr.3: 148865511-148865511 on Build GRCh38
rs199942135 Chr.3: 148865548-148865548 on Build GRCh38
rs777689720 Chr.3: 148865583-148865583 on Build GRCh38
rs756682539 Chr.3: 148865444-148865445 on Build GRCh38
rs79975401 Chr.3: 148865584-148865584 on Build GRCh38
rs1052857510 Chr.3: 148865617-148865617 on Build GRCh38
rs148382472 Chr.3: 148865366-148865366 on Build GRCh38
rs778348410 Chr.3: 148865336-148865336 on Build GRCh38
rs759623323 Chr.3: 148865498-148865498 on Build GRCh38
rs975939886 Chr.3: 148865186-148865186 on Build GRCh38
rs180983067 Chr.3: 148865467-148865467 on Build GRCh38
rs571456711 Chr.3: 148865202-148865202 on Build GRCh38
rs777834773 Chr.3: 148865489-148865489 on Build GRCh38
rs909481574 Chr.3: 148865568-148865568 on Build GRCh38
rs750043139 Chr.3: 148865540-148865540 on Build GRCh38
rs768028360 Chr.3: 148865447-148865447 on Build GRCh38
rs980877095 Chr.3: 148865255-148865255 on Build GRCh38
rs919394305 Chr.3: 148865520-148865520 on Build GRCh38
rs373285078 Chr.3: 148865261-148865261 on Build GRCh38
rs569817031 Chr.3: 148865509-148865509 on Build GRCh38
rs755847608 Chr.3: 148865570-148865570 on Build GRCh38
rs768608818 Chr.3: 148865355-148865355 on Build GRCh38
rs990043319 Chr.3: 148865434-148865434 on Build GRCh38
rs750672324 Chr.3: 148865486-148865486 on Build GRCh38
rs751066330 Chr.3: 148865546-148865546 on Build GRCh38
rs752730291 Chr.3: 148865567-148865567 on Build GRCh38
rs764581661 Chr.3: 148865528-148865528 on Build GRCh38
rs200734672 Chr.3: 148865433-148865433 on Build GRCh38
rs757631082 Chr.3: 148865456-148865456 on Build GRCh38
rs116801366 Chr.3: 148865400-148865400 on Build GRCh38
rs370306676 Chr.3: 148865398-148865398 on Build GRCh38
rs145828911 Chr.3: 148865504-148865504 on Build GRCh38
rs551560835 Chr.3: 148865192-148865192 on Build GRCh38
rs553051404 Chr.3: 148865387-148865387 on Build GRCh38
rs773505321 Chr.3: 148865432-148865432 on Build GRCh38
rs547086849 Chr.3: 148865169-148865169 on Build GRCh38
rs755305712 Chr.3: 148865385-148865385 on Build GRCh38
rs771084177 Chr.3: 148865437-148865437 on Build GRCh38
rs762601870 Chr.3: 148865541-148865541 on Build GRCh38
rs757838642 Chr.3: 148865475-148865475 on Build GRCh38
rs776004966 Chr.3: 148865519-148865519 on Build GRCh38
rs781387404 Chr.3: 148865298-148865300 on Build GRCh38
rs200479513 Chr.3: 148865492-148865492 on Build GRCh38
rs111593746 Chr.3: 148865311-148865311 on Build GRCh38
rs189731775 Chr.3: 148865191-148865191 on Build GRCh38
rs758483100 Chr.3: 148865545-148865545 on Build GRCh38
rs780216618 Chr.3: 148865551-148865551 on Build GRCh38
rs184829198 Chr.3: 148865175-148865175 on Build GRCh38
rs777382950 Chr.3: 148865322-148865322 on Build GRCh38
rs776508646 Chr.3: 148865412-148865412 on Build GRCh38
rs934208479 Chr.3: 148865225-148865225 on Build GRCh38
rs867422674 Chr.3: 148865247-148865247 on Build GRCh38
rs943029313 Chr.3: 148865577-148865577 on Build GRCh38
rs925052066 Chr.3: 148865182-148865183 on Build GRCh38

View Full Product Details