Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23935225-23935736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf58
Entrez Gene ID
254158
Gene Name
chromosome X open reading frame 58
Gene Aliases
-
UniGene
Hs.351265
Gene Chromosome Location
Chr.X: 23907932-23939520 on Build GRCh38
Transcripts
NM_001169574.1 NM_152761.2 XM_011545473.2 XM_011545474.2

SNP Detail

SNP ID Chromosome Location
rs777169420 Chr.X: 23935448-23935449 on Build GRCh38
rs913564796 Chr.X: 23935555-23935555 on Build GRCh38
rs896439315 Chr.X: 23935691-23935691 on Build GRCh38
rs775086373 Chr.X: 23935344-23935344 on Build GRCh38
rs913595378 Chr.X: 23935554-23935554 on Build GRCh38
rs770343335 Chr.X: 23935297-23935297 on Build GRCh38
rs764164047 Chr.X: 23935374-23935374 on Build GRCh38
rs946354866 Chr.X: 23935561-23935561 on Build GRCh38
rs888842781 Chr.X: 23935688-23935688 on Build GRCh38
rs375708749 Chr.X: 23935263-23935263 on Build GRCh38
rs570262107 Chr.X: 23935406-23935406 on Build GRCh38
rs371158194 Chr.X: 23935294-23935294 on Build GRCh38
rs1026306543 Chr.X: 23935338-23935338 on Build GRCh38
rs200347693 Chr.X: 23935402-23935402 on Build GRCh38
rs888978682 Chr.X: 23935267-23935267 on Build GRCh38
rs751809793 Chr.X: 23935390-23935390 on Build GRCh38
rs112872519 Chr.X: 23935511-23935511 on Build GRCh38
rs755097364 Chr.X: 23935398-23935398 on Build GRCh38
rs904911426 Chr.X: 23935581-23935581 on Build GRCh38
rs750333278 Chr.X: 23935668-23935668 on Build GRCh38
rs760843941 Chr.X: 23935416-23935416 on Build GRCh38
rs764259936 Chr.X: 23935494-23935494 on Build GRCh38
rs760908965 Chr.X: 23935407-23935407 on Build GRCh38
rs529143167 Chr.X: 23935559-23935559 on Build GRCh38
rs761720174 Chr.X: 23935380-23935380 on Build GRCh38
rs764898494 Chr.X: 23935381-23935381 on Build GRCh38
rs377514334 Chr.X: 23935285-23935285 on Build GRCh38
rs1056309202 Chr.X: 23935614-23935614 on Build GRCh38
rs753800410 Chr.X: 23935375-23935375 on Build GRCh38
rs771619651 Chr.X: 23935343-23935343 on Build GRCh38
rs771179658 Chr.X: 23935319-23935319 on Build GRCh38
rs189246159 Chr.X: 23935523-23935523 on Build GRCh38
rs776351923 Chr.X: 23935576-23935576 on Build GRCh38
rs761572265 Chr.X: 23935521-23935521 on Build GRCh38
rs150830689 Chr.X: 23935372-23935372 on Build GRCh38
rs147217789 Chr.X: 23935280-23935280 on Build GRCh38
rs374205248 Chr.X: 23935318-23935318 on Build GRCh38
rs937855842 Chr.X: 23935592-23935592 on Build GRCh38
rs766946956 Chr.X: 23935366-23935367 on Build GRCh38
rs771360225 Chr.X: 23935438-23935438 on Build GRCh38
rs140614869 Chr.X: 23935337-23935337 on Build GRCh38
rs779082537 Chr.X: 23935469-23935469 on Build GRCh38
rs946448605 Chr.X: 23935275-23935275 on Build GRCh38
rs1053779192 Chr.X: 23935546-23935546 on Build GRCh38
rs144578801 Chr.X: 23935339-23935339 on Build GRCh38
rs761581763 Chr.X: 23935637-23935637 on Build GRCh38
rs778302960 Chr.X: 23935408-23935408 on Build GRCh38
rs928889942 Chr.X: 23935513-23935513 on Build GRCh38
rs778248163 Chr.X: 23935301-23935301 on Build GRCh38

View Full Product Details