Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180199768-180200310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ23867
Entrez Gene ID
200058
Gene Name
uncharacterized protein FLJ23867
Gene Aliases
-
UniGene
Hs.719176
Gene Chromosome Location
Chr.1: 180198009-180200725 on Build GRCh38
Transcripts
NR_026900.1

SNP Detail

SNP ID Chromosome Location
rs914735274 Chr.1: 180199891-180199891 on Build GRCh38
rs1051702066 Chr.1: 180200201-180200201 on Build GRCh38
rs977652107 Chr.1: 180200087-180200087 on Build GRCh38
rs914764911 Chr.1: 180200062-180200062 on Build GRCh38
rs28524589 Chr.1: 180199800-180199800 on Build GRCh38
rs990504385 Chr.1: 180200101-180200101 on Build GRCh38
rs780620533 Chr.1: 180199848-180199848 on Build GRCh38
rs918750485 Chr.1: 180199842-180199842 on Build GRCh38
rs565381314 Chr.1: 180199902-180199902 on Build GRCh38
rs34204805 Chr.1: 180199850-180199850 on Build GRCh38
rs548919265 Chr.1: 180200144-180200144 on Build GRCh38
rs533570205 Chr.1: 180200184-180200184 on Build GRCh38
rs56211231 Chr.1: 180199792-180199792 on Build GRCh38
rs928863351 Chr.1: 180200091-180200091 on Build GRCh38
rs533823148 Chr.1: 180199956-180199956 on Build GRCh38
rs966501608 Chr.1: 180200065-180200065 on Build GRCh38
rs7513063 Chr.1: 180200270-180200270 on Build GRCh38
rs573717784 Chr.1: 180199982-180199982 on Build GRCh38
rs958161773 Chr.1: 180200100-180200100 on Build GRCh38
rs11126 Chr.1: 180200195-180200195 on Build GRCh38
rs538065052 Chr.1: 180199811-180199811 on Build GRCh38
rs901297934 Chr.1: 180200016-180200016 on Build GRCh38
rs188137711 Chr.1: 180200135-180200135 on Build GRCh38
rs910266813 Chr.1: 180199825-180199825 on Build GRCh38
rs1023718050 Chr.1: 180200108-180200108 on Build GRCh38
rs191092649 Chr.1: 180200154-180200154 on Build GRCh38
rs990749527 Chr.1: 180199868-180199868 on Build GRCh38
rs560199085 Chr.1: 180200213-180200213 on Build GRCh38
rs557295144 Chr.1: 180199955-180199955 on Build GRCh38
rs141527330 Chr.1: 180200008-180200008 on Build GRCh38
rs34870356 Chr.1: 180200025-180200025 on Build GRCh38
rs1055895796 Chr.1: 180199945-180199945 on Build GRCh38
rs916341659 Chr.1: 180199801-180199801 on Build GRCh38
rs942143523 Chr.1: 180200159-180200159 on Build GRCh38
rs775445440 Chr.1: 180200282-180200282 on Build GRCh38
rs557633321 Chr.1: 180199893-180199893 on Build GRCh38
rs10913945 Chr.1: 180199973-180199973 on Build GRCh38
rs958058726 Chr.1: 180200042-180200042 on Build GRCh38
rs765102479 Chr.1: 180200030-180200031 on Build GRCh38
rs765888029 Chr.1: 180200226-180200226 on Build GRCh38
rs1032274209 Chr.1: 180200079-180200079 on Build GRCh38
rs938651841 Chr.1: 180199929-180199929 on Build GRCh38
rs183642780 Chr.1: 180200194-180200194 on Build GRCh38
rs367814685 Chr.1: 180199923-180199923 on Build GRCh38
rs1030213225 Chr.1: 180200020-180200020 on Build GRCh38
rs12145255 Chr.1: 180200232-180200232 on Build GRCh38
rs999292394 Chr.1: 180200078-180200078 on Build GRCh38
rs901157705 Chr.1: 180200225-180200225 on Build GRCh38
rs967480274 Chr.1: 180200067-180200067 on Build GRCh38
rs1048659274 Chr.1: 180199943-180199943 on Build GRCh38
rs11414836 Chr.1: 180199795-180199796 on Build GRCh38
rs970536276 Chr.1: 180200129-180200129 on Build GRCh38
rs369415905 Chr.1: 180200054-180200054 on Build GRCh38
rs985879331 Chr.1: 180199824-180199824 on Build GRCh38
rs1017253994 Chr.1: 180200030-180200030 on Build GRCh38
rs1006287073 Chr.1: 180199969-180199969 on Build GRCh38
rs779517925 Chr.1: 180199987-180199987 on Build GRCh38
rs954356328 Chr.1: 180199806-180199806 on Build GRCh38
rs894527303 Chr.1: 180199957-180199957 on Build GRCh38
rs1037786649 Chr.1: 180200172-180200172 on Build GRCh38
rs981844144 Chr.1: 180200130-180200130 on Build GRCh38
rs910141198 Chr.1: 180200186-180200186 on Build GRCh38
rs548936310 Chr.1: 180200238-180200238 on Build GRCh38
rs184842745 Chr.1: 180199919-180199919 on Build GRCh38
rs776393964 Chr.1: 180200023-180200023 on Build GRCh38
rs989240059 Chr.1: 180200060-180200060 on Build GRCh38

View Full Product Details