Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122190019-122190515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC43
Entrez Gene ID
254050
Gene Name
leucine rich repeat containing 43
Gene Aliases
-
UniGene
Hs.374856
Gene Chromosome Location
Chr.12: 122167719-122203471 on Build GRCh38
Transcripts
NM_001098519.1 NM_152759.4 XM_017019124.1 XM_017019125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4699097 c.673G>A p.V225I Chr.12: 122190140-122190140 on Build GRCh38
5953954 c.739C>T p.L247F Chr.12: 122190206-122190206 on Build GRCh38
3457274 c.859C>A p.P287T Chr.12: 122190326-122190326 on Build GRCh38
5503977 c.132G>T p.L44L Chr.12: 122190154-122190154 on Build GRCh38
6015211 c.882G>A p.R294R Chr.12: 122190349-122190349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768007696 Chr.12: 122190269-122190269 on Build GRCh38
rs748450769 Chr.12: 122190133-122190133 on Build GRCh38
rs112371618 Chr.12: 122190104-122190104 on Build GRCh38
rs773893539 Chr.12: 122190277-122190277 on Build GRCh38
rs953190545 Chr.12: 122190046-122190046 on Build GRCh38
rs952812974 Chr.12: 122190357-122190357 on Build GRCh38
rs767346072 Chr.12: 122190171-122190171 on Build GRCh38
rs757139607 Chr.12: 122190098-122190098 on Build GRCh38
rs765390232 Chr.12: 122190258-122190258 on Build GRCh38
rs531649458 Chr.12: 122190057-122190057 on Build GRCh38
rs779153966 Chr.12: 122190178-122190194 on Build GRCh38
rs949616042 Chr.12: 122190102-122190102 on Build GRCh38
rs763320584 Chr.12: 122190256-122190256 on Build GRCh38
rs201401336 Chr.12: 122190320-122190320 on Build GRCh38
rs762590408 Chr.12: 122190162-122190162 on Build GRCh38
rs777288860 Chr.12: 122190394-122190394 on Build GRCh38
rs779263665 Chr.12: 122190113-122190113 on Build GRCh38
rs550088928 Chr.12: 122190434-122190434 on Build GRCh38
rs370975101 Chr.12: 122190253-122190253 on Build GRCh38
rs138172345 Chr.12: 122190278-122190278 on Build GRCh38
rs1008626186 Chr.12: 122190397-122190397 on Build GRCh38
rs558151255 Chr.12: 122190195-122190195 on Build GRCh38
rs762145418 Chr.12: 122190369-122190369 on Build GRCh38
rs368277585 Chr.12: 122190266-122190266 on Build GRCh38
rs553810521 Chr.12: 122190156-122190156 on Build GRCh38
rs758651882 Chr.12: 122190206-122190206 on Build GRCh38
rs749421822 Chr.12: 122190111-122190111 on Build GRCh38
rs751280291 Chr.12: 122190085-122190085 on Build GRCh38
rs888868659 Chr.12: 122190379-122190379 on Build GRCh38
rs781711648 Chr.12: 122190315-122190315 on Build GRCh38
rs746572209 Chr.12: 122190396-122190396 on Build GRCh38
rs757006329 Chr.12: 122190409-122190409 on Build GRCh38
rs754946423 Chr.12: 122190257-122190257 on Build GRCh38
rs752356299 Chr.12: 122190077-122190077 on Build GRCh38
rs780556066 Chr.12: 122190391-122190391 on Build GRCh38
rs200715996 Chr.12: 122190110-122190110 on Build GRCh38
rs768805163 Chr.12: 122190420-122190420 on Build GRCh38
rs199684502 Chr.12: 122190419-122190419 on Build GRCh38
rs969894557 Chr.12: 122190457-122190457 on Build GRCh38
rs760404558 Chr.12: 122190280-122190280 on Build GRCh38
rs369492934 Chr.12: 122190367-122190367 on Build GRCh38
rs569976296 Chr.12: 122190281-122190281 on Build GRCh38
rs770539350 Chr.12: 122190404-122190404 on Build GRCh38
rs962061237 Chr.12: 122190482-122190482 on Build GRCh38
rs372232440 Chr.12: 122190348-122190348 on Build GRCh38
rs749438454 Chr.12: 122190077-122190077 on Build GRCh38
rs746696882 Chr.12: 122190347-122190347 on Build GRCh38
rs772910492 Chr.12: 122190279-122190279 on Build GRCh38
rs750315190 Chr.12: 122190186-122190186 on Build GRCh38
rs963904602 Chr.12: 122190173-122190173 on Build GRCh38
rs760533188 Chr.12: 122190245-122190245 on Build GRCh38
rs765134366 Chr.12: 122190298-122190298 on Build GRCh38
rs761453750 Chr.12: 122190167-122190167 on Build GRCh38
rs199892245 Chr.12: 122190218-122190218 on Build GRCh38
rs377167993 Chr.12: 122190152-122190152 on Build GRCh38
rs376714571 Chr.12: 122190199-122190199 on Build GRCh38
rs77473460 Chr.12: 122190260-122190260 on Build GRCh38
rs767637298 Chr.12: 122190302-122190302 on Build GRCh38
rs201800249 Chr.12: 122190330-122190330 on Build GRCh38
rs369912763 Chr.12: 122190169-122190169 on Build GRCh38
rs747684897 Chr.12: 122190337-122190337 on Build GRCh38
rs776512672 Chr.12: 122190293-122190293 on Build GRCh38
rs899301827 Chr.12: 122190216-122190216 on Build GRCh38
rs11322813 Chr.12: 122190482-122190482 on Build GRCh38
rs776386897 Chr.12: 122190351-122190351 on Build GRCh38
rs766347124 Chr.12: 122190188-122190188 on Build GRCh38
rs547210187 Chr.12: 122190449-122190449 on Build GRCh38
rs780976458 Chr.12: 122190101-122190101 on Build GRCh38
rs761555443 Chr.12: 122190226-122190226 on Build GRCh38
rs369545625 Chr.12: 122190209-122190209 on Build GRCh38
rs4758651 Chr.12: 122190211-122190211 on Build GRCh38
rs769489309 Chr.12: 122190410-122190410 on Build GRCh38
rs775427359 Chr.12: 122190411-122190411 on Build GRCh38
rs1011731415 Chr.12: 122190136-122190136 on Build GRCh38
rs777340085 Chr.12: 122190345-122190345 on Build GRCh38
rs369998737 Chr.12: 122190219-122190219 on Build GRCh38
rs371846622 Chr.12: 122190311-122190311 on Build GRCh38
rs957302653 Chr.12: 122190448-122190448 on Build GRCh38
rs747372452 Chr.12: 122190264-122190264 on Build GRCh38
rs772454725 Chr.12: 122190052-122190052 on Build GRCh38
rs753913249 Chr.12: 122190191-122190191 on Build GRCh38
rs764397269 Chr.12: 122190305-122190305 on Build GRCh38
rs77598875 Chr.12: 122190224-122190224 on Build GRCh38
rs780887755 Chr.12: 122190408-122190408 on Build GRCh38
rs200480977 Chr.12: 122190294-122190294 on Build GRCh38
rs926767185 Chr.12: 122190391-122190392 on Build GRCh38
rs772500076 Chr.12: 122190135-122190135 on Build GRCh38
rs199533998 Chr.12: 122190192-122190192 on Build GRCh38
rs773828491 Chr.12: 122190053-122190053 on Build GRCh38
rs750798913 Chr.12: 122190385-122190385 on Build GRCh38
rs967236240 Chr.12: 122190068-122190068 on Build GRCh38
rs757438318 Chr.12: 122190105-122190111 on Build GRCh38
rs748205408 Chr.12: 122190060-122190060 on Build GRCh38
rs1022758136 Chr.12: 122190456-122190456 on Build GRCh38
rs376552948 Chr.12: 122190352-122190352 on Build GRCh38
rs565799221 Chr.12: 122190474-122190474 on Build GRCh38
rs533359635 Chr.12: 122190263-122190263 on Build GRCh38
rs181314079 Chr.12: 122190163-122190163 on Build GRCh38
rs757864111 Chr.12: 122190393-122190393 on Build GRCh38
rs775347816 Chr.12: 122190365-122190365 on Build GRCh38
rs373355361 Chr.12: 122190392-122190392 on Build GRCh38
rs751280120 Chr.12: 122190164-122190164 on Build GRCh38
rs1036643136 Chr.12: 122190340-122190340 on Build GRCh38
rs756027291 Chr.12: 122190187-122190187 on Build GRCh38
rs771826883 Chr.12: 122190469-122190469 on Build GRCh38
rs772961895 Chr.12: 122190243-122190243 on Build GRCh38
rs892822416 Chr.12: 122190125-122190125 on Build GRCh38
rs1050606223 Chr.12: 122190371-122190371 on Build GRCh38
rs376554290 Chr.12: 122190319-122190319 on Build GRCh38
rs372790097 Chr.12: 122190210-122190210 on Build GRCh38
rs751841325 Chr.12: 122190310-122190310 on Build GRCh38
rs773525458 Chr.12: 122190139-122190139 on Build GRCh38
rs561656002 Chr.12: 122190435-122190435 on Build GRCh38
rs1020056907 Chr.12: 122190138-122190138 on Build GRCh38
rs146275848 Chr.12: 122190268-122190268 on Build GRCh38
rs768838381 Chr.12: 122190112-122190112 on Build GRCh38
rs764270416 Chr.12: 122190368-122190368 on Build GRCh38
rs188524855 Chr.12: 122190140-122190140 on Build GRCh38
rs11060094 Chr.12: 122190233-122190233 on Build GRCh38
rs902860094 Chr.12: 122190438-122190438 on Build GRCh38
rs529006567 Chr.12: 122190446-122190446 on Build GRCh38
rs746769603 Chr.12: 122190308-122190309 on Build GRCh38
rs549688310 Chr.12: 122190103-122190103 on Build GRCh38
rs756496943 Chr.12: 122190390-122190390 on Build GRCh38
rs781739519 Chr.12: 122190267-122190267 on Build GRCh38
rs767895869 Chr.12: 122190374-122190374 on Build GRCh38
rs1008096430 Chr.12: 122190144-122190144 on Build GRCh38

View Full Product Details