Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108958544-108959093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MORC1
Entrez Gene ID
27136
Gene Name
MORC family CW-type zinc finger 1
Gene Aliases
CT33, MORC, ZCW6
UniGene
Hs.591296
Gene Chromosome Location
Chr.3: 108958239-109118146 on Build GRCh38
Transcripts
NM_014429.3 XM_005247362.2 XM_011512691.1 XM_011512692.1 XM_011512693.1 XM_011512694.1 XM_011512696.2 XM_011512697.2 XM_017006170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4002369 c.2946G>T p.S982S Chr.3: 108958974-108958974 on Build GRCh38
3585069 c.2915G>A p.R972K Chr.3: 108959005-108959005 on Build GRCh38
3380327 c.2873T>C p.F958S Chr.3: 108959047-108959047 on Build GRCh38
5624521 c.2829T>G p.T943T Chr.3: 108959091-108959091 on Build GRCh38
1036118 c.2946G>A p.S982S Chr.3: 108958974-108958974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780367204 Chr.3: 108958966-108958966 on Build GRCh38
rs920924345 Chr.3: 108958767-108958767 on Build GRCh38
rs892724548 Chr.3: 108958839-108958845 on Build GRCh38
rs572994021 Chr.3: 108958593-108958593 on Build GRCh38
rs776257775 Chr.3: 108959025-108959025 on Build GRCh38
rs770484120 Chr.3: 108959021-108959021 on Build GRCh38
rs559359796 Chr.3: 108958902-108958902 on Build GRCh38
rs767765321 Chr.3: 108958982-108958982 on Build GRCh38
rs150620007 Chr.3: 108958677-108958677 on Build GRCh38
rs115684294 Chr.3: 108958668-108958668 on Build GRCh38
rs1022267749 Chr.3: 108958863-108958863 on Build GRCh38
rs990389667 Chr.3: 108958847-108958847 on Build GRCh38
rs745663923 Chr.3: 108958714-108958714 on Build GRCh38
rs772586042 Chr.3: 108958986-108958986 on Build GRCh38
rs1009703258 Chr.3: 108958897-108958897 on Build GRCh38
rs16855035 Chr.3: 108958975-108958975 on Build GRCh38
rs941981001 Chr.3: 108959000-108959000 on Build GRCh38
rs1043930305 Chr.3: 108958900-108958900 on Build GRCh38
rs533634908 Chr.3: 108959001-108959001 on Build GRCh38
rs961014392 Chr.3: 108958611-108958612 on Build GRCh38
rs866580468 Chr.3: 108959058-108959058 on Build GRCh38
rs541120499 Chr.3: 108958857-108958857 on Build GRCh38
rs750474678 Chr.3: 108958958-108958958 on Build GRCh38
rs886431625 Chr.3: 108958654-108958654 on Build GRCh38
rs992747148 Chr.3: 108958638-108958638 on Build GRCh38
rs1002568495 Chr.3: 108958768-108958768 on Build GRCh38
rs751247747 Chr.3: 108958936-108958937 on Build GRCh38
rs2197737 Chr.3: 108958974-108958974 on Build GRCh38
rs201701166 Chr.3: 108959061-108959061 on Build GRCh38
rs775306498 Chr.3: 108959049-108959049 on Build GRCh38
rs923390764 Chr.3: 108958904-108958904 on Build GRCh38
rs1004173409 Chr.3: 108958663-108958663 on Build GRCh38
rs370610500 Chr.3: 108958959-108958959 on Build GRCh38
rs532001734 Chr.3: 108958591-108958592 on Build GRCh38
rs1013139592 Chr.3: 108958705-108958705 on Build GRCh38
rs769688342 Chr.3: 108958784-108958784 on Build GRCh38
rs763175019 Chr.3: 108958796-108958796 on Build GRCh38
rs775536773 Chr.3: 108958795-108958795 on Build GRCh38
rs763960275 Chr.3: 108958929-108958929 on Build GRCh38
rs964927791 Chr.3: 108958892-108958892 on Build GRCh38
rs968132046 Chr.3: 108958585-108958585 on Build GRCh38
rs115773666 Chr.3: 108959056-108959056 on Build GRCh38
rs941130997 Chr.3: 108958717-108958717 on Build GRCh38
rs898367607 Chr.3: 108959008-108959008 on Build GRCh38
rs150100342 Chr.3: 108959046-108959046 on Build GRCh38
rs1022349843 Chr.3: 108958583-108958583 on Build GRCh38
rs746451682 Chr.3: 108959005-108959005 on Build GRCh38
rs78212601 Chr.3: 108959063-108959063 on Build GRCh38
rs201034263 Chr.3: 108958979-108958979 on Build GRCh38
rs971520243 Chr.3: 108958730-108958730 on Build GRCh38
rs185341759 Chr.3: 108958636-108958636 on Build GRCh38
rs373141779 Chr.3: 108959041-108959041 on Build GRCh38

View Full Product Details