Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138115114-138115637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME5
Entrez Gene ID
8382
Gene Name
NME/NM23 family member 5
Gene Aliases
NM23-H5, NM23H5, RSPH23
UniGene
Hs.730856
Gene Chromosome Location
Chr.5: 138115172-138139443 on Build GRCh38
Transcripts
NM_003551.2 XM_006714716.3 XM_017009945.1

SNP Detail

SNP ID Chromosome Location
rs921730777 Chr.5: 138115471-138115471 on Build GRCh38
rs760275236 Chr.5: 138115350-138115350 on Build GRCh38
rs186364134 Chr.5: 138115361-138115361 on Build GRCh38
rs971019496 Chr.5: 138115257-138115257 on Build GRCh38
rs899368343 Chr.5: 138115235-138115235 on Build GRCh38
rs1004657594 Chr.5: 138115180-138115180 on Build GRCh38
rs62383493 Chr.5: 138115292-138115292 on Build GRCh38
rs996508597 Chr.5: 138115588-138115588 on Build GRCh38
rs866881000 Chr.5: 138115339-138115339 on Build GRCh38
rs964435112 Chr.5: 138115452-138115452 on Build GRCh38
rs956694469 Chr.5: 138115326-138115326 on Build GRCh38
rs1012420030 Chr.5: 138115239-138115239 on Build GRCh38
rs933242574 Chr.5: 138115523-138115523 on Build GRCh38
rs903522701 Chr.5: 138115553-138115553 on Build GRCh38
rs1030986744 Chr.5: 138115311-138115311 on Build GRCh38
rs913403021 Chr.5: 138115470-138115470 on Build GRCh38
rs548360823 Chr.5: 138115492-138115492 on Build GRCh38
rs576743328 Chr.5: 138115225-138115225 on Build GRCh38
rs530778995 Chr.5: 138115530-138115530 on Build GRCh38
rs545934056 Chr.5: 138115243-138115243 on Build GRCh38
rs181221057 Chr.5: 138115189-138115189 on Build GRCh38
rs190185957 Chr.5: 138115517-138115517 on Build GRCh38
rs1048930 Chr.5: 138115582-138115582 on Build GRCh38
rs970052445 Chr.5: 138115348-138115348 on Build GRCh38
rs780531195 Chr.5: 138115332-138115332 on Build GRCh38
rs142638270 Chr.5: 138115469-138115469 on Build GRCh38
rs559654401 Chr.5: 138115347-138115347 on Build GRCh38
rs762305675 Chr.5: 138115560-138115560 on Build GRCh38
rs1056746790 Chr.5: 138115383-138115383 on Build GRCh38
rs1043457463 Chr.5: 138115552-138115552 on Build GRCh38
rs938827713 Chr.5: 138115413-138115413 on Build GRCh38

View Full Product Details