Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37968625-37969133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2F
Entrez Gene ID
5435
Gene Name
RNA polymerase II subunit F
Gene Aliases
HRBP14.4, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPC15
UniGene
Hs.436578
Gene Chromosome Location
Chr.22: 37953663-38041915 on Build GRCh38
Transcripts
NM_001301129.1 NM_001301130.1 NM_001301131.1 NM_021974.4 NR_125371.1 XM_017028832.1

SNP Detail

SNP ID Chromosome Location
rs989224097 Chr.22: 37968729-37968729 on Build GRCh38
rs955315687 Chr.22: 37968921-37968921 on Build GRCh38
rs1020687341 Chr.22: 37969054-37969054 on Build GRCh38
rs996711363 Chr.22: 37968657-37968657 on Build GRCh38
rs901091701 Chr.22: 37968956-37968956 on Build GRCh38
rs997049897 Chr.22: 37968977-37968977 on Build GRCh38
rs900974199 Chr.22: 37968656-37968656 on Build GRCh38
rs867107073 Chr.22: 37968769-37968769 on Build GRCh38
rs139880 Chr.22: 37968924-37968924 on Build GRCh38
rs893679781 Chr.22: 37969042-37969042 on Build GRCh38
rs572488829 Chr.22: 37968997-37968997 on Build GRCh38
rs570976518 Chr.22: 37968778-37968778 on Build GRCh38
rs3026642 Chr.22: 37968682-37968682 on Build GRCh38
rs934409019 Chr.22: 37968951-37968951 on Build GRCh38
rs1034738372 Chr.22: 37968932-37968932 on Build GRCh38
rs554169918 Chr.22: 37968976-37968976 on Build GRCh38
rs1041338100 Chr.22: 37968940-37968940 on Build GRCh38
rs770677934 Chr.22: 37969086-37969086 on Build GRCh38
rs745322611 Chr.22: 37968800-37968800 on Build GRCh38
rs183489647 Chr.22: 37968692-37968692 on Build GRCh38
rs1027988182 Chr.22: 37968668-37968668 on Build GRCh38
rs780566541 Chr.22: 37968909-37968909 on Build GRCh38
rs748380051 Chr.22: 37969023-37969023 on Build GRCh38
rs552891279 Chr.22: 37968753-37968753 on Build GRCh38
rs950773259 Chr.22: 37968736-37968736 on Build GRCh38
rs757848952 Chr.22: 37968899-37968899 on Build GRCh38
rs3026643 Chr.22: 37968816-37968816 on Build GRCh38
rs966748671 Chr.22: 37969055-37969055 on Build GRCh38
rs566871088 Chr.22: 37968701-37968701 on Build GRCh38
rs3026644 Chr.22: 37968826-37968826 on Build GRCh38
rs934440217 Chr.22: 37968646-37968646 on Build GRCh38
rs892831687 Chr.22: 37968655-37968656 on Build GRCh38
rs777171484 Chr.22: 37968998-37968998 on Build GRCh38
rs971579485 Chr.22: 37968922-37968922 on Build GRCh38
rs907433289 Chr.22: 37969036-37969036 on Build GRCh38
rs538386856 Chr.22: 37968795-37968795 on Build GRCh38
rs948359280 Chr.22: 37969010-37969010 on Build GRCh38
rs753467922 Chr.22: 37968737-37968737 on Build GRCh38
rs1047925044 Chr.22: 37969053-37969053 on Build GRCh38
rs558788315 Chr.22: 37969051-37969051 on Build GRCh38
rs767433207 Chr.22: 37968655-37968655 on Build GRCh38
rs981816951 Chr.22: 37969074-37969074 on Build GRCh38
rs1003016550 Chr.22: 37968687-37968687 on Build GRCh38
rs920799657 Chr.22: 37968878-37968878 on Build GRCh38
rs982757386 Chr.22: 37968806-37968806 on Build GRCh38
rs924481602 Chr.22: 37968944-37968944 on Build GRCh38
rs930714761 Chr.22: 37968879-37968879 on Build GRCh38
rs540151969 Chr.22: 37969033-37969033 on Build GRCh38
rs528022546 Chr.22: 37968728-37968728 on Build GRCh38
rs769039011 Chr.22: 37968970-37968970 on Build GRCh38
rs990995993 Chr.22: 37968831-37968831 on Build GRCh38

View Full Product Details