Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129738305-129738794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTGES
Entrez Gene ID
9536
Gene Name
prostaglandin E synthase
Gene Aliases
MGST-IV, MGST1-L1, MGST1L1, MPGES, PGES, PIG12, PP102, PP1294, TP53I12, mPGES-1
UniGene
Hs.146688
Gene Chromosome Location
Chr.9: 129738336-129777579 on Build GRCh38
Transcripts
NM_004878.4 XM_011519210.2

SNP Detail

SNP ID Chromosome Location
rs1048558957 Chr.9: 129738730-129738730 on Build GRCh38
rs200442832 Chr.9: 129738572-129738572 on Build GRCh38
rs3892537 Chr.9: 129738628-129738628 on Build GRCh38
rs1043934249 Chr.9: 129738461-129738461 on Build GRCh38
rs190173379 Chr.9: 129738332-129738332 on Build GRCh38
rs992096285 Chr.9: 129738640-129738640 on Build GRCh38
rs200795148 Chr.9: 129738347-129738347 on Build GRCh38
rs900127528 Chr.9: 129738712-129738712 on Build GRCh38
rs914634210 Chr.9: 129738632-129738632 on Build GRCh38
rs201326372 Chr.9: 129738552-129738552 on Build GRCh38
rs202149383 Chr.9: 129738770-129738770 on Build GRCh38
rs199663488 Chr.9: 129738680-129738680 on Build GRCh38
rs1035704463 Chr.9: 129738621-129738621 on Build GRCh38
rs201196781 Chr.9: 129738670-129738670 on Build GRCh38
rs200024254 Chr.9: 129738420-129738420 on Build GRCh38
rs200862723 Chr.9: 129738760-129738760 on Build GRCh38
rs201493719 Chr.9: 129738410-129738410 on Build GRCh38
rs912246671 Chr.9: 129738699-129738699 on Build GRCh38
rs770219825 Chr.9: 129738411-129738416 on Build GRCh38
rs375073395 Chr.9: 129738411-129738414 on Build GRCh38
rs200641487 Chr.9: 129738397-129738397 on Build GRCh38
rs754396822 Chr.9: 129738413-129738416 on Build GRCh38
rs201150451 Chr.9: 129738583-129738583 on Build GRCh38
rs201106450 Chr.9: 129738525-129738525 on Build GRCh38
rs1801978 Chr.9: 129738569-129738569 on Build GRCh38
rs199660000 Chr.9: 129738340-129738340 on Build GRCh38
rs890928256 Chr.9: 129738436-129738436 on Build GRCh38
rs947847446 Chr.9: 129738457-129738457 on Build GRCh38
rs374501779 Chr.9: 129738412-129738412 on Build GRCh38
rs200541878 Chr.9: 129738330-129738330 on Build GRCh38
rs995717947 Chr.9: 129738574-129738574 on Build GRCh38
rs201991407 Chr.9: 129738456-129738456 on Build GRCh38
rs199887997 Chr.9: 129738747-129738747 on Build GRCh38
rs201303826 Chr.9: 129738356-129738356 on Build GRCh38
rs962524993 Chr.9: 129738503-129738503 on Build GRCh38
rs201720877 Chr.9: 129738703-129738703 on Build GRCh38
rs199604354 Chr.9: 129738596-129738596 on Build GRCh38
rs202058937 Chr.9: 129738540-129738540 on Build GRCh38
rs139907561 Chr.9: 129738448-129738449 on Build GRCh38
rs996158737 Chr.9: 129738716-129738716 on Build GRCh38
rs1039140620 Chr.9: 129738675-129738675 on Build GRCh38
rs1027559274 Chr.9: 129738575-129738575 on Build GRCh38
rs45455892 Chr.9: 129738693-129738693 on Build GRCh38
rs199812659 Chr.9: 129738385-129738385 on Build GRCh38
rs900073866 Chr.9: 129738573-129738573 on Build GRCh38
rs969282389 Chr.9: 129738672-129738672 on Build GRCh38
rs45542831 Chr.9: 129738547-129738547 on Build GRCh38
rs57195369 Chr.9: 129738458-129738459 on Build GRCh38
rs200136040 Chr.9: 129738496-129738496 on Build GRCh38
rs201628510 Chr.9: 129738692-129738692 on Build GRCh38
rs201147953 Chr.9: 129738455-129738455 on Build GRCh38
rs376567249 Chr.9: 129738587-129738587 on Build GRCh38
rs71385459 Chr.9: 129738453-129738458 on Build GRCh38
rs200894291 Chr.9: 129738414-129738414 on Build GRCh38
rs1029605670 Chr.9: 129738379-129738379 on Build GRCh38
rs1015743038 Chr.9: 129738501-129738501 on Build GRCh38
rs992043866 Chr.9: 129738355-129738356 on Build GRCh38
rs199906204 Chr.9: 129738412-129738412 on Build GRCh38
rs202048693 Chr.9: 129738416-129738416 on Build GRCh38
rs574731851 Chr.9: 129738627-129738627 on Build GRCh38
rs572539131 Chr.9: 129738398-129738398 on Build GRCh38
rs45544737 Chr.9: 129738701-129738701 on Build GRCh38
rs200800282 Chr.9: 129738681-129738681 on Build GRCh38
rs995224139 Chr.9: 129738526-129738526 on Build GRCh38
rs1052106881 Chr.9: 129738531-129738531 on Build GRCh38

View Full Product Details