Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 185091308-185091814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF2
Entrez Gene ID
6045
Gene Name
ring finger protein 2
Gene Aliases
BAP-1, BAP1, DING, HIPI3, RING1B, RING2
UniGene
Hs.591490
Gene Chromosome Location
Chr.1: 185045419-185102608 on Build GRCh38
Transcripts
NM_007212.3 XM_005245413.3 XM_011509851.2 XM_011509852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6060411 c.157A>T p.I53F Chr.1: 185091648-185091648 on Build GRCh38
2098073 c.210delT p.C72fs*23 Chr.1: 185091701-185091701 on Build GRCh38
5623749 c.103G>T p.G35C Chr.1: 185091594-185091594 on Build GRCh38
424789 c.133C>T p.L45L Chr.1: 185091624-185091624 on Build GRCh38
6835371 c.125C>T p.P42L Chr.1: 185091616-185091616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530142997 Chr.1: 185091387-185091387 on Build GRCh38
rs371064337 Chr.1: 185091767-185091767 on Build GRCh38
rs750656273 Chr.1: 185091745-185091745 on Build GRCh38
rs376791167 Chr.1: 185091783-185091783 on Build GRCh38
rs746600470 Chr.1: 185091568-185091568 on Build GRCh38
rs756486168 Chr.1: 185091784-185091784 on Build GRCh38
rs1026130754 Chr.1: 185091479-185091479 on Build GRCh38
rs369869181 Chr.1: 185091774-185091774 on Build GRCh38
rs369972542 Chr.1: 185091671-185091671 on Build GRCh38
rs748422837 Chr.1: 185091524-185091524 on Build GRCh38
rs570179831 Chr.1: 185091753-185091753 on Build GRCh38
rs763609823 Chr.1: 185091648-185091648 on Build GRCh38
rs974825834 Chr.1: 185091765-185091765 on Build GRCh38
rs1010290140 Chr.1: 185091396-185091396 on Build GRCh38
rs868029311 Chr.1: 185091425-185091425 on Build GRCh38
rs200385213 Chr.1: 185091777-185091777 on Build GRCh38
rs879151829 Chr.1: 185091469-185091469 on Build GRCh38
rs781363917 Chr.1: 185091585-185091585 on Build GRCh38
rs548848546 Chr.1: 185091422-185091422 on Build GRCh38
rs765003126 Chr.1: 185091761-185091761 on Build GRCh38
rs776058949 Chr.1: 185091647-185091647 on Build GRCh38
rs780292156 Chr.1: 185091785-185091785 on Build GRCh38
rs865983679 Chr.1: 185091709-185091709 on Build GRCh38
rs950626399 Chr.1: 185091508-185091508 on Build GRCh38
rs777280395 Chr.1: 185091538-185091538 on Build GRCh38
rs775024802 Chr.1: 185091432-185091432 on Build GRCh38
rs937147960 Chr.1: 185091788-185091788 on Build GRCh38
rs907959808 Chr.1: 185091710-185091710 on Build GRCh38
rs1009058704 Chr.1: 185091412-185091412 on Build GRCh38
rs767761145 Chr.1: 185091736-185091736 on Build GRCh38
rs545644574 Chr.1: 185091618-185091618 on Build GRCh38
rs140977509 Chr.1: 185091760-185091760 on Build GRCh38
rs373023574 Chr.1: 185091776-185091776 on Build GRCh38
rs981229538 Chr.1: 185091475-185091475 on Build GRCh38
rs144442502 Chr.1: 185091665-185091665 on Build GRCh38
rs1012665269 Chr.1: 185091402-185091402 on Build GRCh38
rs768795162 Chr.1: 185091653-185091653 on Build GRCh38
rs971234525 Chr.1: 185091464-185091464 on Build GRCh38
rs567091916 Chr.1: 185091424-185091424 on Build GRCh38
rs1041456706 Chr.1: 185091401-185091401 on Build GRCh38
rs770094200 Chr.1: 185091635-185091635 on Build GRCh38
rs892822239 Chr.1: 185091387-185091389 on Build GRCh38
rs145415168 Chr.1: 185091341-185091341 on Build GRCh38
rs895114299 Chr.1: 185091399-185091399 on Build GRCh38
rs996424478 Chr.1: 185091490-185091490 on Build GRCh38
rs770602645 Chr.1: 185091572-185091572 on Build GRCh38
rs372665414 Chr.1: 185091588-185091588 on Build GRCh38
rs758338389 Chr.1: 185091770-185091770 on Build GRCh38
rs751093217 Chr.1: 185091775-185091775 on Build GRCh38

View Full Product Details